Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43321270-43321791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
API5
Entrez Gene ID
8539
Gene Name
apoptosis inhibitor 5
Gene Aliases
AAC-11, AAC11
UniGene
Hs.435771
Gene Chromosome Location
Chr.11: 43311955-43344530 on Build GRCh38
Transcripts
NM_001142930.1 NM_001142931.1 NM_001243747.1 NM_006595.3 NR_024625.1 XM_006718359.3 XM_017018464.1 XM_017018465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183213 c.359C>T p.A120V Chr.11: 43321444-43321444 on Build GRCh38
6602161 c.366A>G p.L122L Chr.11: 43321451-43321451 on Build GRCh38
6602160 c.366A>G p.L122L Chr.11: 43321451-43321451 on Build GRCh38
3869146 c.345C>T p.N115N Chr.11: 43321430-43321430 on Build GRCh38
6602162 c.366A>G p.L122L Chr.11: 43321451-43321451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368627391 Chr.11: 43321509-43321509 on Build GRCh38
rs992778161 Chr.11: 43321547-43321547 on Build GRCh38
rs531733147 Chr.11: 43321586-43321586 on Build GRCh38
rs5743220 Chr.11: 43321337-43321337 on Build GRCh38
rs1020390471 Chr.11: 43321724-43321724 on Build GRCh38
rs1022665020 Chr.11: 43321674-43321674 on Build GRCh38
rs930739978 Chr.11: 43321634-43321634 on Build GRCh38
rs5743221 Chr.11: 43321522-43321522 on Build GRCh38
rs1048409872 Chr.11: 43321611-43321611 on Build GRCh38
rs762961238 Chr.11: 43321406-43321406 on Build GRCh38
rs768539957 Chr.11: 43321389-43321389 on Build GRCh38
rs746959048 Chr.11: 43321381-43321381 on Build GRCh38
rs548036839 Chr.11: 43321719-43321719 on Build GRCh38
rs749154411 Chr.11: 43321704-43321704 on Build GRCh38
rs889290661 Chr.11: 43321679-43321679 on Build GRCh38
rs368247674 Chr.11: 43321373-43321373 on Build GRCh38
rs965047237 Chr.11: 43321354-43321354 on Build GRCh38
rs5743222 Chr.11: 43321707-43321707 on Build GRCh38
rs760294766 Chr.11: 43321744-43321744 on Build GRCh38
rs375400893 Chr.11: 43321375-43321375 on Build GRCh38
rs145077695 Chr.11: 43321441-43321441 on Build GRCh38
rs750546492 Chr.11: 43321327-43321327 on Build GRCh38
rs911948073 Chr.11: 43321470-43321470 on Build GRCh38
rs71488359 Chr.11: 43321376-43321376 on Build GRCh38
rs958568292 Chr.11: 43321764-43321765 on Build GRCh38
rs183525306 Chr.11: 43321556-43321556 on Build GRCh38
rs968756230 Chr.11: 43321683-43321683 on Build GRCh38
rs755844541 Chr.11: 43321631-43321631 on Build GRCh38
rs915546871 Chr.11: 43321299-43321299 on Build GRCh38
rs747299634 Chr.11: 43321536-43321536 on Build GRCh38
rs564382849 Chr.11: 43321562-43321562 on Build GRCh38
rs995132250 Chr.11: 43321591-43321591 on Build GRCh38
rs1039057534 Chr.11: 43321561-43321561 on Build GRCh38
rs759791687 Chr.11: 43321448-43321448 on Build GRCh38
rs992642353 Chr.11: 43321767-43321767 on Build GRCh38
rs944866765 Chr.11: 43321687-43321687 on Build GRCh38
rs934456669 Chr.11: 43321374-43321374 on Build GRCh38
rs753414483 Chr.11: 43321526-43321526 on Build GRCh38
rs747820811 Chr.11: 43321515-43321515 on Build GRCh38
rs767821087 Chr.11: 43321464-43321464 on Build GRCh38
rs189513622 Chr.11: 43321675-43321675 on Build GRCh38
rs987294166 Chr.11: 43321741-43321741 on Build GRCh38
rs771148577 Chr.11: 43321430-43321430 on Build GRCh38
rs892700092 Chr.11: 43321717-43321717 on Build GRCh38
rs1020055278 Chr.11: 43321379-43321379 on Build GRCh38
rs886606642 Chr.11: 43321644-43321644 on Build GRCh38
rs191191477 Chr.11: 43321298-43321298 on Build GRCh38
rs193018735 Chr.11: 43321759-43321759 on Build GRCh38
rs981987727 Chr.11: 43321645-43321645 on Build GRCh38
rs968857849 Chr.11: 43321421-43321421 on Build GRCh38
rs562419927 Chr.11: 43321647-43321647 on Build GRCh38
rs754398157 Chr.11: 43321320-43321320 on Build GRCh38
rs552730819 Chr.11: 43321370-43321370 on Build GRCh38
rs543421437 Chr.11: 43321646-43321646 on Build GRCh38
rs777607214 Chr.11: 43321658-43321658 on Build GRCh38
rs1009106135 Chr.11: 43321332-43321332 on Build GRCh38
rs753078724 Chr.11: 43321499-43321499 on Build GRCh38

View Full Product Details