Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44096061-44096558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXT2
Entrez Gene ID
2132
Gene Name
exostosin glycosyltransferase 2
Gene Aliases
SOTV, SSMS
UniGene
Hs.368404
Gene Chromosome Location
Chr.11: 44095549-44245430 on Build GRCh38
Transcripts
NM_000401.3 NM_001178083.1 NM_207122.1 XM_011519950.1 XM_011519951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019370 c.69+5G>A p.? Chr.11: 44096326-44096326 on Build GRCh38
6518876 c.7T>A p.C3S Chr.11: 44096259-44096259 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1048981523 Chr.11: 44096166-44096166 on Build GRCh38
rs778390973 Chr.11: 44096294-44096294 on Build GRCh38
rs769460577 Chr.11: 44096343-44096373 on Build GRCh38
rs1032612888 Chr.11: 44096305-44096305 on Build GRCh38
rs927800406 Chr.11: 44096229-44096229 on Build GRCh38
rs555447984 Chr.11: 44096527-44096527 on Build GRCh38
rs11037860 Chr.11: 44096348-44096348 on Build GRCh38
rs1036189808 Chr.11: 44096464-44096464 on Build GRCh38
rs971896368 Chr.11: 44096224-44096224 on Build GRCh38
rs931926355 Chr.11: 44096164-44096164 on Build GRCh38
rs534658369 Chr.11: 44096255-44096255 on Build GRCh38
rs780312380 Chr.11: 44096480-44096480 on Build GRCh38
rs887573884 Chr.11: 44096171-44096171 on Build GRCh38
rs1050826491 Chr.11: 44096402-44096402 on Build GRCh38
rs547551412 Chr.11: 44096525-44096525 on Build GRCh38
rs951208739 Chr.11: 44096337-44096367 on Build GRCh38
rs931334983 Chr.11: 44096384-44096384 on Build GRCh38
rs773135572 Chr.11: 44096341-44096341 on Build GRCh38
rs797007580 Chr.11: 44096126-44096126 on Build GRCh38
rs909148638 Chr.11: 44096415-44096415 on Build GRCh38
rs915872102 Chr.11: 44096222-44096222 on Build GRCh38
rs971398650 Chr.11: 44096485-44096485 on Build GRCh38
rs985090634 Chr.11: 44096413-44096413 on Build GRCh38
rs998962711 Chr.11: 44096170-44096176 on Build GRCh38
rs1032888653 Chr.11: 44096120-44096120 on Build GRCh38
rs563323146 Chr.11: 44096127-44096127 on Build GRCh38
rs912664568 Chr.11: 44096139-44096139 on Build GRCh38
rs1019692241 Chr.11: 44096337-44096337 on Build GRCh38
rs534390864 Chr.11: 44096373-44096373 on Build GRCh38
rs776783788 Chr.11: 44096162-44096162 on Build GRCh38
rs576067180 Chr.11: 44096516-44096516 on Build GRCh38
rs776139089 Chr.11: 44096217-44096217 on Build GRCh38
rs112082531 Chr.11: 44096326-44096326 on Build GRCh38
rs1001484410 Chr.11: 44096300-44096300 on Build GRCh38
rs940547680 Chr.11: 44096182-44096182 on Build GRCh38
rs902926143 Chr.11: 44096412-44096412 on Build GRCh38
rs1033251717 Chr.11: 44096188-44096188 on Build GRCh38
rs74980806 Chr.11: 44096509-44096509 on Build GRCh38
rs1005665541 Chr.11: 44096120-44096122 on Build GRCh38
rs988199397 Chr.11: 44096135-44096135 on Build GRCh38
rs979614904 Chr.11: 44096087-44096087 on Build GRCh38
rs944006795 Chr.11: 44096141-44096141 on Build GRCh38
rs144870408 Chr.11: 44096446-44096446 on Build GRCh38
rs975843693 Chr.11: 44096149-44096149 on Build GRCh38
rs566098004 Chr.11: 44096407-44096407 on Build GRCh38
rs956830475 Chr.11: 44096315-44096315 on Build GRCh38
rs982614569 Chr.11: 44096523-44096523 on Build GRCh38
rs543522647 Chr.11: 44096366-44096411 on Build GRCh38
rs904831952 Chr.11: 44096513-44096513 on Build GRCh38
rs764950894 Chr.11: 44096338-44096338 on Build GRCh38
rs1013135001 Chr.11: 44096473-44096474 on Build GRCh38
rs530511530 Chr.11: 44096203-44096203 on Build GRCh38
rs762836059 Chr.11: 44096346-44096346 on Build GRCh38
rs1026436564 Chr.11: 44096493-44096494 on Build GRCh38
rs941248621 Chr.11: 44096327-44096327 on Build GRCh38
rs1033135840 Chr.11: 44096414-44096414 on Build GRCh38
rs981421936 Chr.11: 44096493-44096493 on Build GRCh38
rs547982536 Chr.11: 44096395-44096395 on Build GRCh38
rs921231578 Chr.11: 44096383-44096383 on Build GRCh38
rs991936248 Chr.11: 44096206-44096206 on Build GRCh38
rs997914505 Chr.11: 44096137-44096137 on Build GRCh38
rs1046923672 Chr.11: 44096318-44096318 on Build GRCh38
rs1023753999 Chr.11: 44096262-44096262 on Build GRCh38
rs949119484 Chr.11: 44096496-44096496 on Build GRCh38
rs981495657 Chr.11: 44096225-44096225 on Build GRCh38

View Full Product Details