Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46603230-46603740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HARBI1
Entrez Gene ID
283254
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77
UniGene
Hs.714463
Gene Chromosome Location
Chr.11: 46603027-46617651 on Build GRCh38
Transcripts
NM_173811.3 XM_011520025.1 XM_011520026.2 XM_011520027.1 XM_011520028.2 XM_011520029.2 XM_017017602.1 XM_017017603.1 XM_017017604.1 XM_017017605.1 XM_017017606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033037 c.965C>T p.P322L Chr.11: 46603615-46603615 on Build GRCh38
6132759 c.983A>G p.H328R Chr.11: 46603597-46603597 on Build GRCh38
7206152 c.990G>T p.E330D Chr.11: 46603590-46603590 on Build GRCh38
3359195 c.906C>T p.S302S Chr.11: 46603674-46603674 on Build GRCh38
6992738 c.989A>T p.E330V Chr.11: 46603591-46603591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372811398 Chr.11: 46603629-46603629 on Build GRCh38
rs1032362382 Chr.11: 46603474-46603474 on Build GRCh38
rs556584273 Chr.11: 46603479-46603479 on Build GRCh38
rs1028917259 Chr.11: 46603510-46603510 on Build GRCh38
rs185640614 Chr.11: 46603641-46603641 on Build GRCh38
rs752403768 Chr.11: 46603379-46603379 on Build GRCh38
rs755986675 Chr.11: 46603519-46603519 on Build GRCh38
rs941653213 Chr.11: 46603262-46603262 on Build GRCh38
rs545753876 Chr.11: 46603342-46603342 on Build GRCh38
rs576847235 Chr.11: 46603291-46603291 on Build GRCh38
rs752364425 Chr.11: 46603670-46603670 on Build GRCh38
rs542270082 Chr.11: 46603309-46603309 on Build GRCh38
rs766607928 Chr.11: 46603562-46603562 on Build GRCh38
rs202159962 Chr.11: 46603501-46603501 on Build GRCh38
rs899225475 Chr.11: 46603360-46603360 on Build GRCh38
rs779875591 Chr.11: 46603699-46603699 on Build GRCh38
rs753419895 Chr.11: 46603679-46603679 on Build GRCh38
rs985897244 Chr.11: 46603637-46603637 on Build GRCh38
rs773103636 Chr.11: 46603717-46603717 on Build GRCh38
rs761869794 Chr.11: 46603550-46603550 on Build GRCh38
rs927065567 Chr.11: 46603650-46603650 on Build GRCh38
rs763967077 Chr.11: 46603673-46603673 on Build GRCh38
rs12362183 Chr.11: 46603294-46603294 on Build GRCh38
rs748317544 Chr.11: 46603708-46603708 on Build GRCh38
rs770082532 Chr.11: 46603710-46603710 on Build GRCh38
rs201248157 Chr.11: 46603607-46603607 on Build GRCh38
rs759963705 Chr.11: 46603644-46603644 on Build GRCh38
rs758160529 Chr.11: 46603698-46603698 on Build GRCh38
rs768059419 Chr.11: 46603661-46603661 on Build GRCh38
rs763462311 Chr.11: 46603677-46603677 on Build GRCh38
rs770866746 Chr.11: 46603675-46603675 on Build GRCh38
rs749157398 Chr.11: 46603602-46603602 on Build GRCh38
rs1037963684 Chr.11: 46603343-46603343 on Build GRCh38
rs369696306 Chr.11: 46603622-46603622 on Build GRCh38
rs757084968 Chr.11: 46603683-46603683 on Build GRCh38
rs529843288 Chr.11: 46603568-46603568 on Build GRCh38
rs371969577 Chr.11: 46603561-46603561 on Build GRCh38
rs1036294747 Chr.11: 46603274-46603274 on Build GRCh38
rs768871024 Chr.11: 46603525-46603525 on Build GRCh38
rs1009488361 Chr.11: 46603553-46603553 on Build GRCh38
rs778897133 Chr.11: 46603695-46603695 on Build GRCh38
rs745677263 Chr.11: 46603610-46603610 on Build GRCh38
rs765370645 Chr.11: 46603556-46603556 on Build GRCh38
rs140123366 Chr.11: 46603598-46603598 on Build GRCh38
rs968462363 Chr.11: 46603627-46603627 on Build GRCh38
rs745873713 Chr.11: 46603696-46603696 on Build GRCh38
rs776636016 Chr.11: 46603529-46603529 on Build GRCh38
rs926951605 Chr.11: 46603264-46603264 on Build GRCh38
rs976983246 Chr.11: 46603615-46603615 on Build GRCh38
rs762065869 Chr.11: 46603616-46603616 on Build GRCh38
rs908860839 Chr.11: 46603260-46603260 on Build GRCh38
rs35569076 Chr.11: 46603590-46603590 on Build GRCh38
rs11038934 Chr.11: 46603485-46603485 on Build GRCh38
rs377297419 Chr.11: 46603631-46603631 on Build GRCh38
rs770599417 Chr.11: 46603493-46603493 on Build GRCh38
rs940524835 Chr.11: 46603316-46603316 on Build GRCh38
rs143408296 Chr.11: 46603277-46603277 on Build GRCh38
rs373418691 Chr.11: 46603639-46603639 on Build GRCh38
rs752452401 Chr.11: 46603591-46603591 on Build GRCh38
rs201752855 Chr.11: 46603614-46603614 on Build GRCh38
rs562293059 Chr.11: 46603567-46603567 on Build GRCh38
rs777381633 Chr.11: 46603600-46603600 on Build GRCh38
rs771913231 Chr.11: 46603613-46603613 on Build GRCh38
rs1002226534 Chr.11: 46603471-46603471 on Build GRCh38
rs1046649576 Chr.11: 46603318-46603318 on Build GRCh38
rs905304112 Chr.11: 46603464-46603464 on Build GRCh38

View Full Product Details