Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46617190-46617681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1 NM_173811.3 XM_011520025.1 XM_011520026.2 XM_011520028.2 XM_011520029.2 XM_017017605.1

Gene Symbol
HARBI1
Entrez Gene ID
283254
Gene Name
harbinger transposase derived 1
Gene Aliases
C11orf77
UniGene
Hs.714463
Gene Chromosome Location
Chr.11: 46603027-46617651 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1 NM_173811.3 XM_011520025.1 XM_011520026.2 XM_011520028.2 XM_011520029.2 XM_017017605.1

SNP Detail

SNP ID Chromosome Location
rs919202027 Chr.11: 46617650-46617650 on Build GRCh38
rs569041443 Chr.11: 46617476-46617476 on Build GRCh38
rs999529107 Chr.11: 46617287-46617287 on Build GRCh38
rs1014516548 Chr.11: 46617260-46617260 on Build GRCh38
rs950507956 Chr.11: 46617581-46617581 on Build GRCh38
rs935432242 Chr.11: 46617648-46617648 on Build GRCh38
rs548247208 Chr.11: 46617546-46617546 on Build GRCh38
rs927886098 Chr.11: 46617232-46617232 on Build GRCh38
rs144543788 Chr.11: 46617336-46617337 on Build GRCh38
rs796473266 Chr.11: 46617337-46617338 on Build GRCh38
rs556773501 Chr.11: 46617631-46617631 on Build GRCh38
rs879625136 Chr.11: 46617402-46617402 on Build GRCh38
rs1007533472 Chr.11: 46617549-46617550 on Build GRCh38
rs537924218 Chr.11: 46617543-46617543 on Build GRCh38
rs544254279 Chr.11: 46617261-46617261 on Build GRCh38
rs895755519 Chr.11: 46617533-46617533 on Build GRCh38
rs920265809 Chr.11: 46617218-46617218 on Build GRCh38
rs367729861 Chr.11: 46617337-46617338 on Build GRCh38
rs560142974 Chr.11: 46617445-46617445 on Build GRCh38
rs759709291 Chr.11: 46617338-46617339 on Build GRCh38
rs1021236535 Chr.11: 46617567-46617567 on Build GRCh38
rs1007074011 Chr.11: 46617335-46617335 on Build GRCh38
rs762321504 Chr.11: 46617356-46617356 on Build GRCh38
rs944834793 Chr.11: 46617345-46617345 on Build GRCh38
rs1048149379 Chr.11: 46617231-46617231 on Build GRCh38
rs1034253854 Chr.11: 46617230-46617230 on Build GRCh38
rs533900267 Chr.11: 46617549-46617549 on Build GRCh38
rs758136693 Chr.11: 46617388-46617388 on Build GRCh38
rs1027647100 Chr.11: 46617289-46617289 on Build GRCh38
rs1014985113 Chr.11: 46617263-46617263 on Build GRCh38
rs953332144 Chr.11: 46617543-46617544 on Build GRCh38
rs1051122202 Chr.11: 46617514-46617514 on Build GRCh38
rs554450226 Chr.11: 46617550-46617550 on Build GRCh38
rs916498550 Chr.11: 46617638-46617638 on Build GRCh38
rs912163727 Chr.11: 46617342-46617342 on Build GRCh38
rs895640086 Chr.11: 46617237-46617237 on Build GRCh38
rs544558426 Chr.11: 46617547-46617548 on Build GRCh38
rs994234522 Chr.11: 46617234-46617234 on Build GRCh38
rs200022679 Chr.11: 46617547-46617547 on Build GRCh38
rs902576846 Chr.11: 46617483-46617483 on Build GRCh38
rs568128728 Chr.11: 46617548-46617548 on Build GRCh38
rs866761530 Chr.11: 46617410-46617410 on Build GRCh38
rs532385942 Chr.11: 46617454-46617454 on Build GRCh38
rs112588575 Chr.11: 46617548-46617549 on Build GRCh38
rs868631897 Chr.11: 46617325-46617325 on Build GRCh38
rs1043783617 Chr.11: 46617281-46617281 on Build GRCh38
rs968594240 Chr.11: 46617571-46617571 on Build GRCh38
rs966705149 Chr.11: 46617544-46617544 on Build GRCh38
rs191845906 Chr.11: 46617223-46617223 on Build GRCh38
rs565018957 Chr.11: 46617209-46617209 on Build GRCh38
rs962440465 Chr.11: 46617269-46617269 on Build GRCh38
rs576682604 Chr.11: 46617632-46617632 on Build GRCh38
rs935385628 Chr.11: 46617498-46617498 on Build GRCh38
rs1010996012 Chr.11: 46617264-46617264 on Build GRCh38
rs540059893 Chr.11: 46617603-46617603 on Build GRCh38
rs937037469 Chr.11: 46617509-46617509 on Build GRCh38
rs953395813 Chr.11: 46617318-46617318 on Build GRCh38
rs140143793 Chr.11: 46617361-46617361 on Build GRCh38
rs1055608478 Chr.11: 46617532-46617532 on Build GRCh38
rs983580939 Chr.11: 46617595-46617595 on Build GRCh38
rs1055006987 Chr.11: 46617441-46617441 on Build GRCh38
rs560889603 Chr.11: 46617357-46617357 on Build GRCh38
rs916425391 Chr.11: 46617455-46617455 on Build GRCh38
rs1014048781 Chr.11: 46617541-46617542 on Build GRCh38
rs939266010 Chr.11: 46617416-46617416 on Build GRCh38
rs868574052 Chr.11: 46617636-46617636 on Build GRCh38
rs995673971 Chr.11: 46617552-46617552 on Build GRCh38
rs149866474 Chr.11: 46617385-46617385 on Build GRCh38
rs925717915 Chr.11: 46617546-46617547 on Build GRCh38
rs946523530 Chr.11: 46617473-46617473 on Build GRCh38
rs973163535 Chr.11: 46617371-46617371 on Build GRCh38
rs894955718 Chr.11: 46617253-46617253 on Build GRCh38
rs958455639 Chr.11: 46617620-46617620 on Build GRCh38
rs904232113 Chr.11: 46617219-46617219 on Build GRCh38
rs1018401662 Chr.11: 46617338-46617338 on Build GRCh38
rs113299100 Chr.11: 46617337-46617340 on Build GRCh38
rs555848238 Chr.11: 46617340-46617341 on Build GRCh38
rs973088152 Chr.11: 46617217-46617217 on Build GRCh38
rs1015178546 Chr.11: 46617542-46617543 on Build GRCh38
rs946554628 Chr.11: 46617639-46617639 on Build GRCh38
rs1010216548 Chr.11: 46617566-46617566 on Build GRCh38
rs866062357 Chr.11: 46617540-46617540 on Build GRCh38
rs1027847469 Chr.11: 46617562-46617562 on Build GRCh38
rs902546389 Chr.11: 46617285-46617285 on Build GRCh38
rs923746745 Chr.11: 46617641-46617641 on Build GRCh38
rs1030357721 Chr.11: 46617298-46617298 on Build GRCh38
rs546883583 Chr.11: 46617397-46617397 on Build GRCh38
rs914054715 Chr.11: 46617545-46617546 on Build GRCh38
rs979588808 Chr.11: 46617577-46617577 on Build GRCh38
rs988446541 Chr.11: 46617545-46617545 on Build GRCh38
rs552074985 Chr.11: 46617474-46617474 on Build GRCh38
rs979274865 Chr.11: 46617640-46617640 on Build GRCh38
rs927589148 Chr.11: 46617409-46617409 on Build GRCh38
rs1054976436 Chr.11: 46617244-46617244 on Build GRCh38
rs898708616 Chr.11: 46617551-46617551 on Build GRCh38
rs939194210 Chr.11: 46617240-46617240 on Build GRCh38
rs183754521 Chr.11: 46617598-46617598 on Build GRCh38
rs904179466 Chr.11: 46617480-46617480 on Build GRCh38

View Full Product Details