Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46751031-46751531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKAP5
Entrez Gene ID
9793
Gene Name
cytoskeleton associated protein 5
Gene Aliases
CHTOG, MSPS, TOG, TOGp, ch-TOG
UniGene
Hs.201253
Gene Chromosome Location
Chr.11: 46743534-46846308 on Build GRCh38
Transcripts
NM_001008938.3 NM_014756.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871323 c.5378A>G p.H1793R Chr.11: 46751200-46751200 on Build GRCh38
4393080 c.5292A>G p.L1764L Chr.11: 46751376-46751376 on Build GRCh38
6245285 c.5293C>A p.H1765N Chr.11: 46751375-46751375 on Build GRCh38
1604582 c.5163T>C p.P1721P Chr.11: 46751505-46751505 on Build GRCh38
6452057 c.5347G>A p.D1783N Chr.11: 46751231-46751231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773861293 Chr.11: 46751123-46751123 on Build GRCh38
rs1023703426 Chr.11: 46751181-46751181 on Build GRCh38
rs749691034 Chr.11: 46751391-46751391 on Build GRCh38
rs778203793 Chr.11: 46751239-46751239 on Build GRCh38
rs753078108 Chr.11: 46751364-46751364 on Build GRCh38
rs201908100 Chr.11: 46751262-46751262 on Build GRCh38
rs751881686 Chr.11: 46751332-46751332 on Build GRCh38
rs550843243 Chr.11: 46751085-46751085 on Build GRCh38
rs745589422 Chr.11: 46751400-46751400 on Build GRCh38
rs781759452 Chr.11: 46751231-46751231 on Build GRCh38
rs757451357 Chr.11: 46751446-46751446 on Build GRCh38
rs931373074 Chr.11: 46751055-46751055 on Build GRCh38
rs766678557 Chr.11: 46751329-46751329 on Build GRCh38
rs771506386 Chr.11: 46751407-46751407 on Build GRCh38
rs567570709 Chr.11: 46751145-46751145 on Build GRCh38
rs141389940 Chr.11: 46751340-46751340 on Build GRCh38
rs774998766 Chr.11: 46751430-46751430 on Build GRCh38
rs368874792 Chr.11: 46751169-46751169 on Build GRCh38
rs530225094 Chr.11: 46751187-46751187 on Build GRCh38
rs759012439 Chr.11: 46751288-46751288 on Build GRCh38
rs1023108863 Chr.11: 46751432-46751432 on Build GRCh38
rs931366287 Chr.11: 46751381-46751381 on Build GRCh38
rs755093929 Chr.11: 46751333-46751333 on Build GRCh38
rs763046677 Chr.11: 46751325-46751325 on Build GRCh38
rs758663989 Chr.11: 46751191-46751191 on Build GRCh38
rs373170724 Chr.11: 46751295-46751295 on Build GRCh38
rs375405792 Chr.11: 46751213-46751213 on Build GRCh38
rs986983190 Chr.11: 46751263-46751263 on Build GRCh38
rs759416998 Chr.11: 46751135-46751135 on Build GRCh38
rs377283816 Chr.11: 46751301-46751301 on Build GRCh38
rs143431133 Chr.11: 46751466-46751466 on Build GRCh38
rs776473892 Chr.11: 46751463-46751463 on Build GRCh38
rs768671080 Chr.11: 46751461-46751461 on Build GRCh38
rs887203219 Chr.11: 46751438-46751438 on Build GRCh38
rs1011838827 Chr.11: 46751161-46751161 on Build GRCh38
rs775404271 Chr.11: 46751298-46751298 on Build GRCh38
rs757655640 Chr.11: 46751394-46751394 on Build GRCh38
rs373055109 Chr.11: 46751211-46751211 on Build GRCh38
rs531144157 Chr.11: 46751074-46751074 on Build GRCh38
rs779293427 Chr.11: 46751134-46751134 on Build GRCh38
rs111277483 Chr.11: 46751220-46751220 on Build GRCh38
rs544629928 Chr.11: 46751084-46751084 on Build GRCh38
rs536983016 Chr.11: 46751073-46751073 on Build GRCh38
rs912787960 Chr.11: 46751276-46751276 on Build GRCh38
rs770280349 Chr.11: 46751122-46751122 on Build GRCh38
rs773806575 Chr.11: 46751270-46751270 on Build GRCh38
rs267602896 Chr.11: 46751261-46751261 on Build GRCh38
rs752670102 Chr.11: 46751149-46751149 on Build GRCh38
rs369323220 Chr.11: 46751232-46751232 on Build GRCh38
rs144266860 Chr.11: 46751173-46751173 on Build GRCh38
rs755600134 Chr.11: 46751103-46751106 on Build GRCh38
rs755149415 Chr.11: 46751223-46751223 on Build GRCh38
rs766168653 Chr.11: 46751316-46751316 on Build GRCh38
rs774708343 Chr.11: 46751492-46751492 on Build GRCh38
rs371086109 Chr.11: 46751324-46751324 on Build GRCh38
rs759348878 Chr.11: 46751062-46751062 on Build GRCh38
rs764020515 Chr.11: 46751186-46751186 on Build GRCh38
rs750829349 Chr.11: 46751249-46751249 on Build GRCh38
rs891124851 Chr.11: 46751253-46751253 on Build GRCh38
rs756595178 Chr.11: 46751268-46751268 on Build GRCh38
rs368963583 Chr.11: 46751094-46751094 on Build GRCh38
rs899689327 Chr.11: 46751109-46751109 on Build GRCh38
rs748494485 Chr.11: 46751080-46751080 on Build GRCh38
rs756658081 Chr.11: 46751367-46751367 on Build GRCh38
rs1030757148 Chr.11: 46751247-46751247 on Build GRCh38
rs767770984 Chr.11: 46751343-46751343 on Build GRCh38
rs549527156 Chr.11: 46751238-46751238 on Build GRCh38
rs778511792 Chr.11: 46751399-46751399 on Build GRCh38
rs780232262 Chr.11: 46751206-46751206 on Build GRCh38
rs753944221 Chr.11: 46751369-46751369 on Build GRCh38

View Full Product Details