Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47594011-47594510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF4
Entrez Gene ID
114900
Gene Name
C1q and tumor necrosis factor related protein 4
Gene Aliases
CTRP4, ZACRP4
UniGene
Hs.662633
Gene Chromosome Location
Chr.11: 47589664-47596336 on Build GRCh38
Transcripts
NM_031909.2 XM_017017165.1 XM_017017166.1

SNP Detail

SNP ID Chromosome Location
rs989813935 Chr.11: 47594455-47594455 on Build GRCh38
rs937413314 Chr.11: 47594123-47594123 on Build GRCh38
rs947922372 Chr.11: 47594476-47594476 on Build GRCh38
rs1007704587 Chr.11: 47594189-47594189 on Build GRCh38
rs1009731679 Chr.11: 47594446-47594446 on Build GRCh38
rs970939653 Chr.11: 47594385-47594399 on Build GRCh38
rs914706099 Chr.11: 47594125-47594125 on Build GRCh38
rs906402144 Chr.11: 47594418-47594418 on Build GRCh38
rs899528112 Chr.11: 47594419-47594419 on Build GRCh38
rs975493130 Chr.11: 47594408-47594408 on Build GRCh38
rs35624992 Chr.11: 47594246-47594246 on Build GRCh38
rs1036096293 Chr.11: 47594236-47594236 on Build GRCh38
rs149860042 Chr.11: 47594177-47594177 on Build GRCh38
rs975501611 Chr.11: 47594201-47594214 on Build GRCh38
rs145527423 Chr.11: 47594250-47594250 on Build GRCh38
rs529173066 Chr.11: 47594313-47594313 on Build GRCh38
rs1016045798 Chr.11: 47594190-47594190 on Build GRCh38
rs1021581161 Chr.11: 47594454-47594454 on Build GRCh38
rs977561922 Chr.11: 47594265-47594265 on Build GRCh38
rs997105509 Chr.11: 47594178-47594178 on Build GRCh38
rs898971803 Chr.11: 47594197-47594197 on Build GRCh38
rs1003190225 Chr.11: 47594353-47594353 on Build GRCh38
rs943063796 Chr.11: 47594356-47594356 on Build GRCh38
rs964132114 Chr.11: 47594196-47594202 on Build GRCh38
rs539072285 Chr.11: 47594467-47594467 on Build GRCh38
rs563351087 Chr.11: 47594055-47594055 on Build GRCh38
rs1017537906 Chr.11: 47594194-47594194 on Build GRCh38
rs932318089 Chr.11: 47594428-47594428 on Build GRCh38
rs947584013 Chr.11: 47594159-47594159 on Build GRCh38
rs1033629728 Chr.11: 47594475-47594475 on Build GRCh38
rs1023231200 Chr.11: 47594132-47594132 on Build GRCh38
rs1055921717 Chr.11: 47594124-47594124 on Build GRCh38
rs924815863 Chr.11: 47594284-47594284 on Build GRCh38
rs887342451 Chr.11: 47594365-47594365 on Build GRCh38
rs550617367 Chr.11: 47594346-47594346 on Build GRCh38
rs902342023 Chr.11: 47594036-47594036 on Build GRCh38
rs906218746 Chr.11: 47594140-47594140 on Build GRCh38
rs993350771 Chr.11: 47594201-47594201 on Build GRCh38
rs984327039 Chr.11: 47594186-47594186 on Build GRCh38
rs1006228488 Chr.11: 47594367-47594367 on Build GRCh38
rs906197749 Chr.11: 47594339-47594339 on Build GRCh38
rs559319451 Chr.11: 47594422-47594422 on Build GRCh38
rs1037834760 Chr.11: 47594373-47594373 on Build GRCh38
rs1051859604 Chr.11: 47594180-47594180 on Build GRCh38
rs891374261 Chr.11: 47594444-47594444 on Build GRCh38
rs958756056 Chr.11: 47594243-47594243 on Build GRCh38
rs528116278 Chr.11: 47594216-47594216 on Build GRCh38
rs898919960 Chr.11: 47594396-47594396 on Build GRCh38
rs112029759 Chr.11: 47594248-47594248 on Build GRCh38
rs556193048 Chr.11: 47594414-47594414 on Build GRCh38
rs935235853 Chr.11: 47594430-47594430 on Build GRCh38
rs933572405 Chr.11: 47594304-47594304 on Build GRCh38
rs1041922656 Chr.11: 47594160-47594160 on Build GRCh38
rs10838733 Chr.11: 47594093-47594093 on Build GRCh38
rs970638106 Chr.11: 47594230-47594230 on Build GRCh38
rs369101689 Chr.11: 47594355-47594355 on Build GRCh38
rs915042766 Chr.11: 47594464-47594464 on Build GRCh38
rs560913067 Chr.11: 47594181-47594181 on Build GRCh38
rs550945498 Chr.11: 47594231-47594231 on Build GRCh38
rs951340098 Chr.11: 47594074-47594074 on Build GRCh38
rs981320031 Chr.11: 47594414-47594428 on Build GRCh38
rs541296128 Chr.11: 47594400-47594400 on Build GRCh38
rs1026198819 Chr.11: 47594207-47594207 on Build GRCh38
rs947774657 Chr.11: 47594306-47594306 on Build GRCh38
rs567774093 Chr.11: 47594351-47594351 on Build GRCh38
rs1051328849 Chr.11: 47594434-47594434 on Build GRCh38
rs746681721 Chr.11: 47594405-47594405 on Build GRCh38
rs991968891 Chr.11: 47594244-47594244 on Build GRCh38
rs928700063 Chr.11: 47594094-47594094 on Build GRCh38

View Full Product Details