Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61338978-61339477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TKFC
Entrez Gene ID
26007
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK, NET45
UniGene
Hs.6278
Gene Chromosome Location
Chr.11: 61333182-61353426 on Build GRCh38
Transcripts
NM_015533.3 XM_011544912.2 XM_017017517.1 XM_017017518.1 XM_017017519.1 XM_017017520.1 XM_017017521.1 XM_017017522.1 XM_017017523.1 XM_017017524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4034590 c.343C>T p.R115W Chr.11: 61339292-61339292 on Build GRCh38
6069710 c.294G>A p.Q98Q Chr.11: 61339166-61339166 on Build GRCh38
6639973 c.418G>A p.D140N Chr.11: 61339367-61339367 on Build GRCh38
3953549 c.226A>G p.I76V Chr.11: 61339098-61339098 on Build GRCh38
5341638 c.476T>A p.L159H Chr.11: 61339425-61339425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779842201 Chr.11: 61339075-61339075 on Build GRCh38
rs370439014 Chr.11: 61339268-61339268 on Build GRCh38
rs185836791 Chr.11: 61339404-61339404 on Build GRCh38
rs180855288 Chr.11: 61339320-61339320 on Build GRCh38
rs755856348 Chr.11: 61339197-61339197 on Build GRCh38
rs960647340 Chr.11: 61339424-61339424 on Build GRCh38
rs773310649 Chr.11: 61339247-61339247 on Build GRCh38
rs746348610 Chr.11: 61339104-61339104 on Build GRCh38
rs1043424139 Chr.11: 61339080-61339080 on Build GRCh38
rs748915118 Chr.11: 61339261-61339261 on Build GRCh38
rs567418824 Chr.11: 61339123-61339123 on Build GRCh38
rs530349834 Chr.11: 61339259-61339259 on Build GRCh38
rs141905816 Chr.11: 61339253-61339253 on Build GRCh38
rs557756151 Chr.11: 61339045-61339045 on Build GRCh38
rs750037556 Chr.11: 61339326-61339326 on Build GRCh38
rs763190395 Chr.11: 61339249-61339249 on Build GRCh38
rs761137715 Chr.11: 61339052-61339052 on Build GRCh38
rs771230345 Chr.11: 61339362-61339362 on Build GRCh38
rs368669120 Chr.11: 61339040-61339040 on Build GRCh38
rs371388511 Chr.11: 61339030-61339030 on Build GRCh38
rs202102256 Chr.11: 61339056-61339056 on Build GRCh38
rs946402010 Chr.11: 61339143-61339143 on Build GRCh38
rs778035941 Chr.11: 61339348-61339348 on Build GRCh38
rs944852661 Chr.11: 61339364-61339364 on Build GRCh38
rs752031119 Chr.11: 61339308-61339309 on Build GRCh38
rs376636000 Chr.11: 61339322-61339322 on Build GRCh38
rs373807994 Chr.11: 61339455-61339455 on Build GRCh38
rs143016521 Chr.11: 61339365-61339365 on Build GRCh38
rs932193642 Chr.11: 61339333-61339333 on Build GRCh38
rs112242971 Chr.11: 61339170-61339170 on Build GRCh38
rs371411471 Chr.11: 61339292-61339292 on Build GRCh38
rs552874545 Chr.11: 61339359-61339359 on Build GRCh38
rs140281606 Chr.11: 61339408-61339408 on Build GRCh38
rs780114599 Chr.11: 61339158-61339158 on Build GRCh38
rs373623918 Chr.11: 61339303-61339303 on Build GRCh38
rs748620750 Chr.11: 61339343-61339343 on Build GRCh38
rs770309389 Chr.11: 61339347-61339347 on Build GRCh38
rs757985334 Chr.11: 61339341-61339341 on Build GRCh38
rs564696910 Chr.11: 61339302-61339302 on Build GRCh38
rs779478150 Chr.11: 61339407-61339407 on Build GRCh38
rs144455681 Chr.11: 61339100-61339100 on Build GRCh38
rs199784392 Chr.11: 61339024-61339024 on Build GRCh38
rs773572870 Chr.11: 61339289-61339289 on Build GRCh38
rs369626074 Chr.11: 61339240-61339240 on Build GRCh38
rs756957642 Chr.11: 61339339-61339339 on Build GRCh38
rs772166296 Chr.11: 61339367-61339367 on Build GRCh38
rs375300749 Chr.11: 61339062-61339062 on Build GRCh38
rs376310837 Chr.11: 61339260-61339260 on Build GRCh38
rs370756103 Chr.11: 61339021-61339021 on Build GRCh38
rs139771453 Chr.11: 61339157-61339157 on Build GRCh38
rs781010563 Chr.11: 61339175-61339175 on Build GRCh38
rs760571222 Chr.11: 61339383-61339383 on Build GRCh38
rs201326183 Chr.11: 61339205-61339205 on Build GRCh38
rs2512809 Chr.11: 61339053-61339053 on Build GRCh38
rs201441428 Chr.11: 61339283-61339283 on Build GRCh38
rs538530023 Chr.11: 61339445-61339445 on Build GRCh38
rs752576247 Chr.11: 61339257-61339257 on Build GRCh38
rs775617467 Chr.11: 61339373-61339373 on Build GRCh38
rs767521245 Chr.11: 61339304-61339304 on Build GRCh38
rs980672009 Chr.11: 61339246-61339246 on Build GRCh38
rs970248692 Chr.11: 61339206-61339206 on Build GRCh38
rs768559166 Chr.11: 61339454-61339454 on Build GRCh38
rs905212953 Chr.11: 61339414-61339414 on Build GRCh38
rs573480382 Chr.11: 61339415-61339415 on Build GRCh38
rs532277913 Chr.11: 61339327-61339327 on Build GRCh38
rs372347031 Chr.11: 61339043-61339043 on Build GRCh38
rs1052145620 Chr.11: 61339102-61339102 on Build GRCh38
rs746801038 Chr.11: 61339022-61339022 on Build GRCh38
rs777289885 Chr.11: 61339278-61339278 on Build GRCh38
rs778328525 Chr.11: 61339417-61339417 on Build GRCh38
rs762359509 Chr.11: 61339103-61339103 on Build GRCh38
rs771883051 Chr.11: 61339018-61339018 on Build GRCh38
rs761223611 Chr.11: 61339124-61339124 on Build GRCh38
rs111677410 Chr.11: 61339386-61339386 on Build GRCh38
rs201876999 Chr.11: 61339250-61339250 on Build GRCh38
rs767431212 Chr.11: 61339256-61339256 on Build GRCh38
rs763510414 Chr.11: 61339084-61339086 on Build GRCh38
rs779671347 Chr.11: 61339342-61339342 on Build GRCh38
rs762167938 Chr.11: 61339132-61339132 on Build GRCh38
rs368722603 Chr.11: 61339101-61339101 on Build GRCh38
rs747570318 Chr.11: 61339211-61339211 on Build GRCh38
rs775503299 Chr.11: 61339447-61339447 on Build GRCh38
rs772254579 Chr.11: 61339434-61339434 on Build GRCh38
rs765371179 Chr.11: 61339139-61339139 on Build GRCh38
rs769342778 Chr.11: 61339276-61339276 on Build GRCh38
rs754234400 Chr.11: 61339129-61339129 on Build GRCh38
rs369912473 Chr.11: 61339204-61339204 on Build GRCh38
rs151177803 Chr.11: 61339366-61339366 on Build GRCh38
rs764662952 Chr.11: 61339120-61339120 on Build GRCh38
rs749692826 Chr.11: 61339418-61339418 on Build GRCh38
rs753639543 Chr.11: 61339324-61339324 on Build GRCh38
rs144902033 Chr.11: 61339293-61339293 on Build GRCh38
rs746138534 Chr.11: 61339430-61339430 on Build GRCh38
rs756912409 Chr.11: 61339266-61339266 on Build GRCh38
rs374441635 Chr.11: 61339038-61339038 on Build GRCh38
rs981594083 Chr.11: 61339210-61339210 on Build GRCh38
rs746890969 Chr.11: 61339078-61339078 on Build GRCh38
rs765055346 Chr.11: 61339401-61339401 on Build GRCh38
rs377465224 Chr.11: 61339203-61339203 on Build GRCh38
rs113335007 Chr.11: 61339154-61339154 on Build GRCh38
rs780922305 Chr.11: 61339087-61339087 on Build GRCh38
rs756929289 Chr.11: 61339325-61339325 on Build GRCh38
rs74878556 Chr.11: 61339072-61339072 on Build GRCh38
rs112901356 Chr.11: 61339238-61339238 on Build GRCh38
rs760798315 Chr.11: 61339449-61339449 on Build GRCh38
rs756033895 Chr.11: 61339314-61339314 on Build GRCh38
rs747904236 Chr.11: 61339191-61339191 on Build GRCh38
rs201898222 Chr.11: 61339239-61339239 on Build GRCh38
rs371048615 Chr.11: 61339403-61339403 on Build GRCh38
rs199709993 Chr.11: 61339398-61339398 on Build GRCh38
rs747818339 Chr.11: 61339091-61339091 on Build GRCh38
rs1029195117 Chr.11: 61339015-61339015 on Build GRCh38
rs75924231 Chr.11: 61339169-61339169 on Build GRCh38
rs768439545 Chr.11: 61339084-61339084 on Build GRCh38
rs565293005 Chr.11: 61339208-61339208 on Build GRCh38
rs201453927 Chr.11: 61339060-61339060 on Build GRCh38
rs149026556 Chr.11: 61339313-61339313 on Build GRCh38
rs768353683 Chr.11: 61339023-61339023 on Build GRCh38
rs761711882 Chr.11: 61339400-61339400 on Build GRCh38
rs758426408 Chr.11: 61339074-61339074 on Build GRCh38

View Full Product Details