Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61795260-61795761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEN1
Entrez Gene ID
2237
Gene Name
flap structure-specific endonuclease 1
Gene Aliases
FEN-1, MF1, RAD2
UniGene
Hs.409065
Gene Chromosome Location
Chr.11: 61792637-61797244 on Build GRCh38
Transcripts
NM_004111.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4880792 c.288G>A p.E96E Chr.11: 61795649-61795649 on Build GRCh38
7305922 c.298C>T p.R100C Chr.11: 61795659-61795659 on Build GRCh38
4684308 c.165T>C p.N55N Chr.11: 61795526-61795526 on Build GRCh38
7109021 c.332T>C p.L111P Chr.11: 61795693-61795693 on Build GRCh38
4169890 c.392T>G p.V131G Chr.11: 61795753-61795753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201579214 Chr.11: 61795571-61795571 on Build GRCh38
rs776543195 Chr.11: 61795509-61795509 on Build GRCh38
rs750703675 Chr.11: 61795672-61795672 on Build GRCh38
rs766466870 Chr.11: 61795311-61795311 on Build GRCh38
rs752884600 Chr.11: 61795443-61795443 on Build GRCh38
rs773119449 Chr.11: 61795382-61795382 on Build GRCh38
rs3218838 Chr.11: 61795304-61795304 on Build GRCh38
rs764173967 Chr.11: 61795652-61795652 on Build GRCh38
rs534561563 Chr.11: 61795359-61795359 on Build GRCh38
rs766377130 Chr.11: 61795409-61795409 on Build GRCh38
rs766728314 Chr.11: 61795674-61795674 on Build GRCh38
rs393487 Chr.11: 61795333-61795333 on Build GRCh38
rs752871732 Chr.11: 61795336-61795336 on Build GRCh38
rs779121706 Chr.11: 61795315-61795315 on Build GRCh38
rs771890165 Chr.11: 61795373-61795373 on Build GRCh38
rs538220260 Chr.11: 61795303-61795303 on Build GRCh38
rs568602534 Chr.11: 61795457-61795457 on Build GRCh38
rs747417941 Chr.11: 61795347-61795347 on Build GRCh38
rs561933742 Chr.11: 61795604-61795604 on Build GRCh38
rs764353660 Chr.11: 61795446-61795446 on Build GRCh38
rs1029309545 Chr.11: 61795493-61795493 on Build GRCh38
rs375531645 Chr.11: 61795314-61795314 on Build GRCh38
rs757985226 Chr.11: 61795580-61795580 on Build GRCh38
rs746376478 Chr.11: 61795484-61795484 on Build GRCh38
rs769573710 Chr.11: 61795620-61795620 on Build GRCh38
rs760566909 Chr.11: 61795390-61795390 on Build GRCh38
rs774473787 Chr.11: 61795654-61795654 on Build GRCh38
rs754406395 Chr.11: 61795680-61795680 on Build GRCh38
rs756703191 Chr.11: 61795500-61795500 on Build GRCh38
rs773997085 Chr.11: 61795362-61795362 on Build GRCh38
rs974120293 Chr.11: 61795309-61795309 on Build GRCh38
rs764263966 Chr.11: 61795560-61795560 on Build GRCh38
rs189588818 Chr.11: 61795508-61795508 on Build GRCh38
rs756841522 Chr.11: 61795705-61795705 on Build GRCh38
rs145287449 Chr.11: 61795579-61795579 on Build GRCh38
rs373769891 Chr.11: 61795312-61795312 on Build GRCh38
rs777413094 Chr.11: 61795684-61795684 on Build GRCh38
rs140458864 Chr.11: 61795501-61795501 on Build GRCh38
rs904014727 Chr.11: 61795419-61795419 on Build GRCh38
rs761130481 Chr.11: 61795673-61795673 on Build GRCh38
rs374732129 Chr.11: 61795562-61795562 on Build GRCh38
rs761359771 Chr.11: 61795368-61795368 on Build GRCh38
rs370694477 Chr.11: 61795346-61795346 on Build GRCh38
rs371846488 Chr.11: 61795646-61795646 on Build GRCh38
rs147881814 Chr.11: 61795445-61795445 on Build GRCh38
rs753408832 Chr.11: 61795442-61795444 on Build GRCh38
rs759383889 Chr.11: 61795520-61795520 on Build GRCh38
rs565918212 Chr.11: 61795349-61795349 on Build GRCh38
rs115282527 Chr.11: 61795592-61795592 on Build GRCh38
rs765213906 Chr.11: 61795438-61795438 on Build GRCh38
rs780924065 Chr.11: 61795706-61795706 on Build GRCh38
rs767759900 Chr.11: 61795570-61795570 on Build GRCh38
rs756555908 Chr.11: 61795576-61795576 on Build GRCh38
rs149164506 Chr.11: 61795682-61795682 on Build GRCh38
rs767828296 Chr.11: 61795671-61795671 on Build GRCh38
rs759600121 Chr.11: 61795435-61795435 on Build GRCh38
rs775415268 Chr.11: 61795536-61795536 on Build GRCh38
rs150399201 Chr.11: 61795544-61795544 on Build GRCh38
rs768058398 Chr.11: 61795360-61795360 on Build GRCh38
rs775440777 Chr.11: 61795628-61795628 on Build GRCh38
rs781676618 Chr.11: 61795474-61795474 on Build GRCh38
rs771800022 Chr.11: 61795503-61795503 on Build GRCh38
rs745612425 Chr.11: 61795718-61795718 on Build GRCh38
rs773029413 Chr.11: 61795504-61795504 on Build GRCh38
rs374555458 Chr.11: 61795719-61795719 on Build GRCh38
rs765263498 Chr.11: 61795521-61795521 on Build GRCh38
rs776255928 Chr.11: 61795613-61795613 on Build GRCh38
rs762018999 Chr.11: 61795669-61795669 on Build GRCh38
rs746730684 Chr.11: 61795507-61795507 on Build GRCh38
rs768648090 Chr.11: 61795730-61795730 on Build GRCh38
rs377525711 Chr.11: 61795325-61795325 on Build GRCh38
rs945654836 Chr.11: 61795701-61795701 on Build GRCh38
rs758674866 Chr.11: 61795337-61795337 on Build GRCh38
rs899549369 Chr.11: 61795465-61795465 on Build GRCh38
rs371381466 Chr.11: 61795698-61795698 on Build GRCh38
rs373582072 Chr.11: 61795554-61795554 on Build GRCh38
rs918211900 Chr.11: 61795295-61795295 on Build GRCh38
rs781574757 Chr.11: 61795351-61795351 on Build GRCh38
rs770679611 Chr.11: 61795605-61795605 on Build GRCh38
rs1032766776 Chr.11: 61795458-61795458 on Build GRCh38
rs753721269 Chr.11: 61795416-61795416 on Build GRCh38
rs145709748 Chr.11: 61795578-61795578 on Build GRCh38
rs751874785 Chr.11: 61795447-61795447 on Build GRCh38
rs202006260 Chr.11: 61795569-61795569 on Build GRCh38

View Full Product Details