Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61873425-61873918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5164072 c.1287-2A>G p.? Chr.11: 61873867-61873867 on Build GRCh38
1604876 c.1320C>A p.D440E Chr.11: 61873832-61873832 on Build GRCh38
122948 c.1321G>A p.A441T Chr.11: 61873831-61873831 on Build GRCh38
4463378 c.1289C>T p.S430F Chr.11: 61873863-61873863 on Build GRCh38
3670825 c.1309A>G p.I437V Chr.11: 61873843-61873843 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766604844 Chr.11: 61873853-61873853 on Build GRCh38
rs536946811 Chr.11: 61873713-61873713 on Build GRCh38
rs772163746 Chr.11: 61873832-61873832 on Build GRCh38
rs545935070 Chr.11: 61873753-61873753 on Build GRCh38
rs768083181 Chr.11: 61873576-61873576 on Build GRCh38
rs531276544 Chr.11: 61873878-61873878 on Build GRCh38
rs908786486 Chr.11: 61873737-61873737 on Build GRCh38
rs1028314982 Chr.11: 61873768-61873768 on Build GRCh38
rs1024080459 Chr.11: 61873668-61873668 on Build GRCh38
rs930271678 Chr.11: 61873619-61873619 on Build GRCh38
rs894197675 Chr.11: 61873627-61873627 on Build GRCh38
rs573254325 Chr.11: 61873764-61873764 on Build GRCh38
rs759692570 Chr.11: 61873875-61873875 on Build GRCh38
rs776905980 Chr.11: 61873865-61873865 on Build GRCh38
rs774027864 Chr.11: 61873827-61873827 on Build GRCh38
rs373904243 Chr.11: 61873889-61873889 on Build GRCh38
rs897753940 Chr.11: 61873571-61873571 on Build GRCh38
rs185716597 Chr.11: 61873447-61873447 on Build GRCh38
rs1048733530 Chr.11: 61873623-61873623 on Build GRCh38
rs779205600 Chr.11: 61873788-61873788 on Build GRCh38
rs766892198 Chr.11: 61873882-61873883 on Build GRCh38
rs562037669 Chr.11: 61873837-61873837 on Build GRCh38
rs370423175 Chr.11: 61873883-61873883 on Build GRCh38
rs541889537 Chr.11: 61873799-61873799 on Build GRCh38
rs763210090 Chr.11: 61873688-61873688 on Build GRCh38
rs755337616 Chr.11: 61873786-61873786 on Build GRCh38
rs754160982 Chr.11: 61873785-61873785 on Build GRCh38
rs748963808 Chr.11: 61873814-61873814 on Build GRCh38
rs907360380 Chr.11: 61873725-61873725 on Build GRCh38
rs576636252 Chr.11: 61873752-61873752 on Build GRCh38
rs1000704007 Chr.11: 61873501-61873501 on Build GRCh38
rs952850909 Chr.11: 61873663-61873663 on Build GRCh38
rs368397981 Chr.11: 61873894-61873894 on Build GRCh38
rs376103076 Chr.11: 61873755-61873755 on Build GRCh38
rs755673208 Chr.11: 61873803-61873803 on Build GRCh38
rs555706986 Chr.11: 61873502-61873502 on Build GRCh38
rs77882990 Chr.11: 61873728-61873728 on Build GRCh38
rs6591665 Chr.11: 61873739-61873739 on Build GRCh38
rs1009964416 Chr.11: 61873526-61873526 on Build GRCh38
rs544058801 Chr.11: 61873630-61873631 on Build GRCh38
rs760814318 Chr.11: 61873848-61873848 on Build GRCh38
rs745409845 Chr.11: 61873801-61873801 on Build GRCh38
rs754236351 Chr.11: 61873886-61873887 on Build GRCh38
rs373538919 Chr.11: 61873810-61873810 on Build GRCh38
rs748766664 Chr.11: 61873448-61873448 on Build GRCh38
rs986933856 Chr.11: 61873498-61873498 on Build GRCh38
rs765572103 Chr.11: 61873877-61873877 on Build GRCh38
rs984341728 Chr.11: 61873685-61873685 on Build GRCh38
rs201037232 Chr.11: 61873614-61873614 on Build GRCh38
rs970713828 Chr.11: 61873557-61873557 on Build GRCh38
rs956687955 Chr.11: 61873522-61873522 on Build GRCh38
rs753191197 Chr.11: 61873796-61873796 on Build GRCh38
rs796929223 Chr.11: 61873742-61873742 on Build GRCh38
rs369996758 Chr.11: 61873798-61873798 on Build GRCh38
rs1025328301 Chr.11: 61873882-61873882 on Build GRCh38
rs932736791 Chr.11: 61873496-61873496 on Build GRCh38
rs748047476 Chr.11: 61873828-61873828 on Build GRCh38
rs763877655 Chr.11: 61873831-61873831 on Build GRCh38
rs568042878 Chr.11: 61873577-61873577 on Build GRCh38
rs763063722 Chr.11: 61873760-61873760 on Build GRCh38
rs888901827 Chr.11: 61873756-61873756 on Build GRCh38
rs758853166 Chr.11: 61873881-61873881 on Build GRCh38
rs1009456535 Chr.11: 61873870-61873870 on Build GRCh38
rs768481436 Chr.11: 61873825-61873825 on Build GRCh38
rs1025002769 Chr.11: 61873553-61873553 on Build GRCh38

View Full Product Details