Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61877420-61877913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133824 c.873C>T p.C291C Chr.11: 61877523-61877523 on Build GRCh38
2037983 c.817C>T p.P273S Chr.11: 61877579-61877579 on Build GRCh38
5482510 c.819G>A p.P273P Chr.11: 61877577-61877577 on Build GRCh38
4198673 c.817C>T p.P273S Chr.11: 61877579-61877579 on Build GRCh38
244344 c.878A>G p.Q293R Chr.11: 61877518-61877518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001413011 Chr.11: 61877675-61877675 on Build GRCh38
rs566336473 Chr.11: 61877724-61877724 on Build GRCh38
rs552598884 Chr.11: 61877699-61877699 on Build GRCh38
rs369417448 Chr.11: 61877622-61877622 on Build GRCh38
rs200549776 Chr.11: 61877586-61877586 on Build GRCh38
rs752560493 Chr.11: 61877578-61877578 on Build GRCh38
rs770028144 Chr.11: 61877593-61877593 on Build GRCh38
rs372745221 Chr.11: 61877628-61877628 on Build GRCh38
rs369043120 Chr.11: 61877538-61877538 on Build GRCh38
rs976537639 Chr.11: 61877754-61877754 on Build GRCh38
rs932176820 Chr.11: 61877569-61877569 on Build GRCh38
rs922073743 Chr.11: 61877768-61877768 on Build GRCh38
rs543881782 Chr.11: 61877521-61877521 on Build GRCh38
rs746038992 Chr.11: 61877591-61877591 on Build GRCh38
rs193273849 Chr.11: 61877452-61877452 on Build GRCh38
rs776704456 Chr.11: 61877484-61877484 on Build GRCh38
rs891474968 Chr.11: 61877609-61877609 on Build GRCh38
rs763403507 Chr.11: 61877493-61877493 on Build GRCh38
rs11285797 Chr.11: 61877711-61877711 on Build GRCh38
rs756857909 Chr.11: 61877468-61877468 on Build GRCh38
rs907852301 Chr.11: 61877694-61877694 on Build GRCh38
rs867553634 Chr.11: 61877606-61877606 on Build GRCh38
rs532500099 Chr.11: 61877689-61877689 on Build GRCh38
rs1040499803 Chr.11: 61877526-61877526 on Build GRCh38
rs376097613 Chr.11: 61877594-61877594 on Build GRCh38
rs930635319 Chr.11: 61877482-61877482 on Build GRCh38
rs966036117 Chr.11: 61877741-61877741 on Build GRCh38
rs17156535 Chr.11: 61877864-61877864 on Build GRCh38
rs747025936 Chr.11: 61877581-61877581 on Build GRCh38
rs754678317 Chr.11: 61877568-61877568 on Build GRCh38
rs80230932 Chr.11: 61877867-61877867 on Build GRCh38
rs780956894 Chr.11: 61877786-61877786 on Build GRCh38
rs375316312 Chr.11: 61877511-61877511 on Build GRCh38
rs750128536 Chr.11: 61877794-61877794 on Build GRCh38
rs777949772 Chr.11: 61877477-61877477 on Build GRCh38
rs749626221 Chr.11: 61877604-61877604 on Build GRCh38
rs773705286 Chr.11: 61877624-61877624 on Build GRCh38
rs1024631186 Chr.11: 61877672-61877672 on Build GRCh38
rs959565252 Chr.11: 61877626-61877626 on Build GRCh38
rs567396015 Chr.11: 61877829-61877829 on Build GRCh38
rs767249475 Chr.11: 61877745-61877745 on Build GRCh38
rs549812906 Chr.11: 61877456-61877456 on Build GRCh38
rs548786780 Chr.11: 61877770-61877770 on Build GRCh38
rs146826857 Chr.11: 61877577-61877577 on Build GRCh38
rs561060179 Chr.11: 61877451-61877451 on Build GRCh38
rs935556220 Chr.11: 61877859-61877859 on Build GRCh38
rs764566538 Chr.11: 61877502-61877502 on Build GRCh38
rs886469969 Chr.11: 61877458-61877458 on Build GRCh38
rs907877474 Chr.11: 61877831-61877831 on Build GRCh38
rs757226141 Chr.11: 61877585-61877585 on Build GRCh38
rs536348319 Chr.11: 61877848-61877848 on Build GRCh38
rs759818192 Chr.11: 61877486-61877486 on Build GRCh38
rs534922976 Chr.11: 61877729-61877729 on Build GRCh38
rs951969550 Chr.11: 61877690-61877690 on Build GRCh38
rs922176163 Chr.11: 61877769-61877778 on Build GRCh38
rs760095252 Chr.11: 61877512-61877512 on Build GRCh38
rs758316528 Chr.11: 61877580-61877580 on Build GRCh38
rs983377696 Chr.11: 61877691-61877691 on Build GRCh38
rs944741492 Chr.11: 61877522-61877522 on Build GRCh38
rs1024062620 Chr.11: 61877632-61877632 on Build GRCh38
rs768174540 Chr.11: 61877535-61877535 on Build GRCh38
rs372803947 Chr.11: 61877459-61877459 on Build GRCh38
rs753370663 Chr.11: 61877557-61877557 on Build GRCh38
rs377336842 Chr.11: 61877646-61877646 on Build GRCh38
rs772165928 Chr.11: 61877471-61877471 on Build GRCh38
rs747036768 Chr.11: 61877478-61877478 on Build GRCh38
rs771082147 Chr.11: 61877479-61877479 on Build GRCh38
rs925514546 Chr.11: 61877830-61877830 on Build GRCh38
rs377121948 Chr.11: 61877610-61877610 on Build GRCh38
rs1055353060 Chr.11: 61877892-61877892 on Build GRCh38
rs900627976 Chr.11: 61877539-61877539 on Build GRCh38
rs970263725 Chr.11: 61877649-61877649 on Build GRCh38
rs563527043 Chr.11: 61877487-61877487 on Build GRCh38

View Full Product Details