Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61878358-61878886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2037986 c.670G>A p.A224T Chr.11: 61878589-61878589 on Build GRCh38
6657931 c.741C>T p.S247S Chr.11: 61878518-61878518 on Build GRCh38
3791683 c.563C>G p.S188C Chr.11: 61878807-61878807 on Build GRCh38
6133822 c.553G>T p.G185C Chr.11: 61878817-61878817 on Build GRCh38
5999822 c.602A>C p.K201T Chr.11: 61878768-61878768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763143301 Chr.11: 61878652-61878652 on Build GRCh38
rs757686611 Chr.11: 61878670-61878670 on Build GRCh38
rs191385221 Chr.11: 61878857-61878857 on Build GRCh38
rs778921217 Chr.11: 61878749-61878749 on Build GRCh38
rs988196396 Chr.11: 61878496-61878496 on Build GRCh38
rs756606931 Chr.11: 61878676-61878676 on Build GRCh38
rs377115399 Chr.11: 61878625-61878625 on Build GRCh38
rs984478849 Chr.11: 61878617-61878617 on Build GRCh38
rs773018713 Chr.11: 61878515-61878515 on Build GRCh38
rs754456378 Chr.11: 61878558-61878558 on Build GRCh38
rs758436055 Chr.11: 61878730-61878734 on Build GRCh38
rs562321390 Chr.11: 61878551-61878551 on Build GRCh38
rs768010610 Chr.11: 61878674-61878674 on Build GRCh38
rs940486155 Chr.11: 61878380-61878380 on Build GRCh38
rs746423300 Chr.11: 61878557-61878557 on Build GRCh38
rs759908821 Chr.11: 61878477-61878483 on Build GRCh38
rs751568238 Chr.11: 61878852-61878853 on Build GRCh38
rs770654064 Chr.11: 61878718-61878718 on Build GRCh38
rs745770235 Chr.11: 61878730-61878730 on Build GRCh38
rs758580469 Chr.11: 61878542-61878542 on Build GRCh38
rs937394914 Chr.11: 61878393-61878393 on Build GRCh38
rs140891212 Chr.11: 61878590-61878590 on Build GRCh38
rs373839824 Chr.11: 61878476-61878476 on Build GRCh38
rs747793266 Chr.11: 61878577-61878577 on Build GRCh38
rs781764082 Chr.11: 61878552-61878552 on Build GRCh38
rs762134990 Chr.11: 61878754-61878754 on Build GRCh38
rs1032388492 Chr.11: 61878473-61878473 on Build GRCh38
rs746772623 Chr.11: 61878607-61878607 on Build GRCh38
rs766762981 Chr.11: 61878779-61878779 on Build GRCh38
rs780946502 Chr.11: 61878833-61878833 on Build GRCh38
rs532210945 Chr.11: 61878737-61878737 on Build GRCh38
rs174453 Chr.11: 61878495-61878495 on Build GRCh38
rs771731285 Chr.11: 61878589-61878589 on Build GRCh38
rs115282539 Chr.11: 61878764-61878764 on Build GRCh38
rs765566852 Chr.11: 61878666-61878677 on Build GRCh38
rs766279636 Chr.11: 61878518-61878518 on Build GRCh38
rs769598053 Chr.11: 61878731-61878731 on Build GRCh38
rs752912955 Chr.11: 61878700-61878709 on Build GRCh38
rs746011256 Chr.11: 61878813-61878813 on Build GRCh38
rs531148440 Chr.11: 61878626-61878626 on Build GRCh38
rs567982589 Chr.11: 61878853-61878853 on Build GRCh38
rs141077762 Chr.11: 61878432-61878432 on Build GRCh38
rs144099007 Chr.11: 61878517-61878517 on Build GRCh38
rs765320619 Chr.11: 61878646-61878646 on Build GRCh38
rs577254441 Chr.11: 61878450-61878450 on Build GRCh38
rs111324241 Chr.11: 61878404-61878404 on Build GRCh38
rs747586335 Chr.11: 61878706-61878706 on Build GRCh38
rs892483420 Chr.11: 61878710-61878710 on Build GRCh38
rs116061818 Chr.11: 61878778-61878778 on Build GRCh38
rs150972989 Chr.11: 61878532-61878532 on Build GRCh38
rs542548263 Chr.11: 61878541-61878541 on Build GRCh38
rs977524977 Chr.11: 61878709-61878710 on Build GRCh38
rs369029437 Chr.11: 61878653-61878653 on Build GRCh38
rs774661108 Chr.11: 61878752-61878752 on Build GRCh38
rs771589591 Chr.11: 61878477-61878477 on Build GRCh38
rs991523054 Chr.11: 61878695-61878695 on Build GRCh38
rs767888295 Chr.11: 61878763-61878763 on Build GRCh38
rs368573265 Chr.11: 61878478-61878478 on Build GRCh38
rs35479241 Chr.11: 61878794-61878794 on Build GRCh38
rs901691583 Chr.11: 61878410-61878410 on Build GRCh38
rs776294841 Chr.11: 61878719-61878719 on Build GRCh38
rs753811059 Chr.11: 61878519-61878519 on Build GRCh38
rs774043881 Chr.11: 61878488-61878488 on Build GRCh38
rs759404488 Chr.11: 61878643-61878643 on Build GRCh38
rs889071757 Chr.11: 61878693-61878693 on Build GRCh38
rs778287206 Chr.11: 61878560-61878560 on Build GRCh38
rs777946568 Chr.11: 61878733-61878734 on Build GRCh38
rs200649493 Chr.11: 61878716-61878716 on Build GRCh38
rs377600477 Chr.11: 61878708-61878708 on Build GRCh38
rs751900081 Chr.11: 61878662-61878662 on Build GRCh38
rs201469463 Chr.11: 61878501-61878501 on Build GRCh38
rs34511441 Chr.11: 61878611-61878611 on Build GRCh38
rs372141360 Chr.11: 61878471-61878471 on Build GRCh38
rs368190921 Chr.11: 61878464-61878464 on Build GRCh38
rs750112001 Chr.11: 61878436-61878436 on Build GRCh38
rs174452 Chr.11: 61878420-61878420 on Build GRCh38
rs1053748678 Chr.11: 61878474-61878474 on Build GRCh38
rs377474838 Chr.11: 61878675-61878675 on Build GRCh38
rs775454512 Chr.11: 61878651-61878651 on Build GRCh38
rs138062761 Chr.11: 61878818-61878818 on Build GRCh38
rs756925798 Chr.11: 61878832-61878832 on Build GRCh38
rs940393056 Chr.11: 61878629-61878629 on Build GRCh38
rs114764370 Chr.11: 61878800-61878800 on Build GRCh38
rs755812101 Chr.11: 61878461-61878461 on Build GRCh38
rs373812045 Chr.11: 61878707-61878707 on Build GRCh38
rs759655325 Chr.11: 61878522-61878522 on Build GRCh38
rs920591252 Chr.11: 61878381-61878381 on Build GRCh38
rs764505559 Chr.11: 61878545-61878545 on Build GRCh38
rs776616849 Chr.11: 61878634-61878634 on Build GRCh38
rs1009565389 Chr.11: 61878714-61878714 on Build GRCh38
rs748803533 Chr.11: 61878484-61878484 on Build GRCh38
rs777278320 Chr.11: 61878810-61878810 on Build GRCh38
rs901638066 Chr.11: 61878672-61878672 on Build GRCh38
rs746721136 Chr.11: 61878715-61878715 on Build GRCh38
rs779163607 Chr.11: 61878463-61878463 on Build GRCh38

View Full Product Details