Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61878627-61879130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
UniGene
Hs.21765
Gene Chromosome Location
Chr.11: 61873523-61892224 on Build GRCh38
Transcripts
NM_021727.4 XM_011545023.2 XM_017017723.1 XM_017017724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7231855 c.625-8C>A p.? Chr.11: 61878642-61878642 on Build GRCh38
3791683 c.563C>G p.S188C Chr.11: 61878807-61878807 on Build GRCh38
6133822 c.553G>T p.G185C Chr.11: 61878817-61878817 on Build GRCh38
7231856 c.625-8C>A p.? Chr.11: 61878642-61878642 on Build GRCh38
5999822 c.602A>C p.K201T Chr.11: 61878768-61878768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762082082 Chr.11: 61878886-61878886 on Build GRCh38
rs773525966 Chr.11: 61878893-61878893 on Build GRCh38
rs763143301 Chr.11: 61878652-61878652 on Build GRCh38
rs924697310 Chr.11: 61879061-61879061 on Build GRCh38
rs757686611 Chr.11: 61878670-61878670 on Build GRCh38
rs774599965 Chr.11: 61878879-61878879 on Build GRCh38
rs191385221 Chr.11: 61878857-61878857 on Build GRCh38
rs778921217 Chr.11: 61878749-61878749 on Build GRCh38
rs756606931 Chr.11: 61878676-61878676 on Build GRCh38
rs933168270 Chr.11: 61878888-61878888 on Build GRCh38
rs977495951 Chr.11: 61878985-61878985 on Build GRCh38
rs758436055 Chr.11: 61878730-61878734 on Build GRCh38
rs768010610 Chr.11: 61878674-61878674 on Build GRCh38
rs751568238 Chr.11: 61878852-61878853 on Build GRCh38
rs770654064 Chr.11: 61878718-61878718 on Build GRCh38
rs745770235 Chr.11: 61878730-61878730 on Build GRCh38
rs564694386 Chr.11: 61879097-61879097 on Build GRCh38
rs781226633 Chr.11: 61879028-61879028 on Build GRCh38
rs200953117 Chr.11: 61878875-61878875 on Build GRCh38
rs1024291103 Chr.11: 61878947-61878947 on Build GRCh38
rs762134990 Chr.11: 61878754-61878754 on Build GRCh38
rs4578389 Chr.11: 61879014-61879014 on Build GRCh38
rs768786562 Chr.11: 61878876-61878876 on Build GRCh38
rs780946502 Chr.11: 61878833-61878833 on Build GRCh38
rs766762981 Chr.11: 61878779-61878779 on Build GRCh38
rs532210945 Chr.11: 61878737-61878737 on Build GRCh38
rs75930575 Chr.11: 61878945-61878945 on Build GRCh38
rs115282539 Chr.11: 61878764-61878764 on Build GRCh38
rs765566852 Chr.11: 61878666-61878677 on Build GRCh38
rs769598053 Chr.11: 61878731-61878731 on Build GRCh38
rs752912955 Chr.11: 61878700-61878709 on Build GRCh38
rs746011256 Chr.11: 61878813-61878813 on Build GRCh38
rs746069330 Chr.11: 61879081-61879081 on Build GRCh38
rs567982589 Chr.11: 61878853-61878853 on Build GRCh38
rs747586335 Chr.11: 61878706-61878706 on Build GRCh38
rs892483420 Chr.11: 61878710-61878710 on Build GRCh38
rs116061818 Chr.11: 61878778-61878778 on Build GRCh38
rs200028748 Chr.11: 61878887-61878887 on Build GRCh38
rs977524977 Chr.11: 61878709-61878710 on Build GRCh38
rs1046472512 Chr.11: 61879017-61879017 on Build GRCh38
rs573275300 Chr.11: 61878942-61878942 on Build GRCh38
rs369029437 Chr.11: 61878653-61878653 on Build GRCh38
rs774661108 Chr.11: 61878752-61878752 on Build GRCh38
rs897966267 Chr.11: 61878901-61878901 on Build GRCh38
rs767888295 Chr.11: 61878763-61878763 on Build GRCh38
rs991523054 Chr.11: 61878695-61878695 on Build GRCh38
rs35479241 Chr.11: 61878794-61878794 on Build GRCh38
rs906632504 Chr.11: 61879020-61879020 on Build GRCh38
rs776294841 Chr.11: 61878719-61878719 on Build GRCh38
rs889071757 Chr.11: 61878693-61878693 on Build GRCh38
rs777946568 Chr.11: 61878733-61878734 on Build GRCh38
rs971774107 Chr.11: 61878873-61878873 on Build GRCh38
rs971424325 Chr.11: 61878957-61878957 on Build GRCh38
rs200649493 Chr.11: 61878716-61878716 on Build GRCh38
rs377600477 Chr.11: 61878708-61878708 on Build GRCh38
rs751900081 Chr.11: 61878662-61878662 on Build GRCh38
rs369897424 Chr.11: 61878894-61878894 on Build GRCh38
rs574208475 Chr.11: 61879087-61879087 on Build GRCh38
rs377474838 Chr.11: 61878675-61878675 on Build GRCh38
rs138062761 Chr.11: 61878818-61878818 on Build GRCh38
rs775454512 Chr.11: 61878651-61878651 on Build GRCh38
rs756925798 Chr.11: 61878832-61878832 on Build GRCh38
rs183602912 Chr.11: 61879086-61879086 on Build GRCh38
rs756994 Chr.11: 61878941-61878941 on Build GRCh38
rs114764370 Chr.11: 61878800-61878800 on Build GRCh38
rs373812045 Chr.11: 61878707-61878707 on Build GRCh38
rs1034235511 Chr.11: 61879036-61879036 on Build GRCh38
rs1009565389 Chr.11: 61878714-61878714 on Build GRCh38
rs901638066 Chr.11: 61878672-61878672 on Build GRCh38
rs777278320 Chr.11: 61878810-61878810 on Build GRCh38
rs144712997 Chr.11: 61878943-61878943 on Build GRCh38
rs746721136 Chr.11: 61878715-61878715 on Build GRCh38
rs910458703 Chr.11: 61878937-61878937 on Build GRCh38

View Full Product Details