Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62356719-62357217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASRGL1
Entrez Gene ID
80150
Gene Name
asparaginase like 1
Gene Aliases
ALP, ALP1, CRASH
UniGene
Hs.535326
Gene Chromosome Location
Chr.11: 62337302-62401399 on Build GRCh38
Transcripts
NM_001083926.1 NM_025080.3 XM_005274305.3 XM_005274306.2 XM_011545265.2 XM_011545266.2 XM_017018354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968384 c.389G>T p.G130V Chr.11: 62357042-62357042 on Build GRCh38
7175552 c.341A>T p.H114L Chr.11: 62356994-62356994 on Build GRCh38
3935545 c.474G>T p.Q158H Chr.11: 62357127-62357127 on Build GRCh38
5726917 c.67G>A p.E23K Chr.11: 62357104-62357104 on Build GRCh38
7131486 c.472C>A p.Q158K Chr.11: 62357125-62357125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754768361 Chr.11: 62356969-62356969 on Build GRCh38
rs187022418 Chr.11: 62356959-62356959 on Build GRCh38
rs1052719230 Chr.11: 62356825-62356828 on Build GRCh38
rs995242834 Chr.11: 62357141-62357141 on Build GRCh38
rs995071615 Chr.11: 62356955-62356955 on Build GRCh38
rs536882543 Chr.11: 62357098-62357098 on Build GRCh38
rs765843116 Chr.11: 62357052-62357052 on Build GRCh38
rs549115796 Chr.11: 62357160-62357160 on Build GRCh38
rs758277116 Chr.11: 62357016-62357016 on Build GRCh38
rs778634465 Chr.11: 62356991-62356991 on Build GRCh38
rs763504270 Chr.11: 62356944-62356944 on Build GRCh38
rs367883842 Chr.11: 62357137-62357137 on Build GRCh38
rs1028600191 Chr.11: 62356855-62356855 on Build GRCh38
rs934405766 Chr.11: 62357032-62357032 on Build GRCh38
rs755645241 Chr.11: 62356962-62356962 on Build GRCh38
rs564551123 Chr.11: 62357008-62357008 on Build GRCh38
rs775950913 Chr.11: 62357126-62357126 on Build GRCh38
rs115767892 Chr.11: 62356809-62356809 on Build GRCh38
rs775529761 Chr.11: 62357103-62357104 on Build GRCh38
rs145178428 Chr.11: 62357120-62357120 on Build GRCh38
rs749655327 Chr.11: 62357115-62357115 on Build GRCh38
rs547493771 Chr.11: 62357035-62357035 on Build GRCh38
rs201316929 Chr.11: 62357133-62357133 on Build GRCh38
rs752425276 Chr.11: 62357082-62357082 on Build GRCh38
rs890053028 Chr.11: 62356862-62356862 on Build GRCh38
rs775939197 Chr.11: 62357034-62357034 on Build GRCh38
rs370679289 Chr.11: 62357021-62357021 on Build GRCh38
rs746117317 Chr.11: 62357030-62357030 on Build GRCh38
rs775919051 Chr.11: 62357114-62357114 on Build GRCh38
rs778023105 Chr.11: 62356936-62356936 on Build GRCh38
rs1044388894 Chr.11: 62357136-62357136 on Build GRCh38
rs924437109 Chr.11: 62356823-62356823 on Build GRCh38
rs991246644 Chr.11: 62357007-62357007 on Build GRCh38
rs770073190 Chr.11: 62357108-62357108 on Build GRCh38
rs764691817 Chr.11: 62357039-62357039 on Build GRCh38
rs747161185 Chr.11: 62357131-62357132 on Build GRCh38
rs777695027 Chr.11: 62357017-62357017 on Build GRCh38
rs1057185791 Chr.11: 62356846-62356846 on Build GRCh38
rs573800955 Chr.11: 62356798-62356798 on Build GRCh38
rs758258713 Chr.11: 62357083-62357083 on Build GRCh38
rs972845145 Chr.11: 62356956-62356956 on Build GRCh38
rs779741497 Chr.11: 62356966-62356966 on Build GRCh38
rs759757314 Chr.11: 62356957-62356957 on Build GRCh38
rs915711989 Chr.11: 62356796-62356796 on Build GRCh38
rs141452891 Chr.11: 62357060-62357060 on Build GRCh38
rs763837582 Chr.11: 62357191-62357191 on Build GRCh38
rs777206182 Chr.11: 62356949-62356950 on Build GRCh38
rs781092984 Chr.11: 62357103-62357103 on Build GRCh38
rs75102382 Chr.11: 62357028-62357028 on Build GRCh38
rs113827204 Chr.11: 62357148-62357148 on Build GRCh38
rs562842363 Chr.11: 62356818-62356818 on Build GRCh38
rs1001341003 Chr.11: 62357085-62357085 on Build GRCh38
rs754580834 Chr.11: 62357064-62357064 on Build GRCh38
rs947298951 Chr.11: 62356800-62356800 on Build GRCh38
rs758168902 Chr.11: 62357165-62357165 on Build GRCh38
rs764710203 Chr.11: 62356957-62356957 on Build GRCh38
rs930547195 Chr.11: 62356794-62356794 on Build GRCh38
rs776518240 Chr.11: 62357143-62357146 on Build GRCh38
rs746969296 Chr.11: 62357093-62357093 on Build GRCh38
rs895897836 Chr.11: 62356888-62356888 on Build GRCh38
rs924521837 Chr.11: 62357118-62357118 on Build GRCh38
rs777676546 Chr.11: 62357084-62357084 on Build GRCh38
rs747683291 Chr.11: 62356913-62356913 on Build GRCh38
rs975060981 Chr.11: 62356763-62356763 on Build GRCh38
rs978664591 Chr.11: 62357101-62357101 on Build GRCh38
rs200094029 Chr.11: 62357041-62357041 on Build GRCh38
rs763414105 Chr.11: 62357038-62357038 on Build GRCh38
rs764868898 Chr.11: 62357146-62357146 on Build GRCh38
rs769189461 Chr.11: 62357116-62357116 on Build GRCh38
rs1049000101 Chr.11: 62356795-62356795 on Build GRCh38
rs760208448 Chr.11: 62357042-62357042 on Build GRCh38
rs763126761 Chr.11: 62357113-62357114 on Build GRCh38
rs972696140 Chr.11: 62356757-62356757 on Build GRCh38
rs758601428 Chr.11: 62356906-62356906 on Build GRCh38
rs770070586 Chr.11: 62356978-62356978 on Build GRCh38
rs879366106 Chr.11: 62356961-62356961 on Build GRCh38
rs770171289 Chr.11: 62357031-62357031 on Build GRCh38
rs950951680 Chr.11: 62357025-62357025 on Build GRCh38
rs892839057 Chr.11: 62356840-62356840 on Build GRCh38
rs369931742 Chr.11: 62356958-62356958 on Build GRCh38
rs200861693 Chr.11: 62357164-62357164 on Build GRCh38
rs753478917 Chr.11: 62356967-62356967 on Build GRCh38
rs934746441 Chr.11: 62356842-62356842 on Build GRCh38
rs770975541 Chr.11: 62357022-62357022 on Build GRCh38
rs762183576 Chr.11: 62357121-62357121 on Build GRCh38
rs758009513 Chr.11: 62356979-62356979 on Build GRCh38
rs1008367687 Chr.11: 62356887-62356887 on Build GRCh38
rs764887146 Chr.11: 62357081-62357081 on Build GRCh38
rs1019402662 Chr.11: 62357029-62357029 on Build GRCh38
rs753224835 Chr.11: 62356941-62356941 on Build GRCh38
rs1044357485 Chr.11: 62356922-62356922 on Build GRCh38
rs116195544 Chr.11: 62357125-62357125 on Build GRCh38
rs899431806 Chr.11: 62356926-62356926 on Build GRCh38
rs531205399 Chr.11: 62356856-62356856 on Build GRCh38

View Full Product Details