Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64243882-64244381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP2
Entrez Gene ID
2286
Gene Name
FK506 binding protein 2
Gene Aliases
FKBP-13, PPIase
UniGene
Hs.227729
Gene Chromosome Location
Chr.11: 64240941-64244135 on Build GRCh38
Transcripts
NM_001135208.1 NM_004470.3 NM_057092.2 XM_005273848.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861152 c.392T>G p.V131G Chr.11: 64243992-64243992 on Build GRCh38
4822180 c.416G>T p.R139L Chr.11: 64244016-64244016 on Build GRCh38
1298360 c.394G>C p.E132Q Chr.11: 64243994-64243994 on Build GRCh38
930042 c.388G>A p.E130K Chr.11: 64243988-64243988 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931914916 Chr.11: 64243906-64243906 on Build GRCh38
rs201304525 Chr.11: 64244151-64244152 on Build GRCh38
rs896249907 Chr.11: 64244016-64244016 on Build GRCh38
rs893393745 Chr.11: 64244111-64244114 on Build GRCh38
rs999192543 Chr.11: 64243993-64243993 on Build GRCh38
rs185914660 Chr.11: 64244067-64244067 on Build GRCh38
rs201962119 Chr.11: 64244150-64244150 on Build GRCh38
rs951855629 Chr.11: 64244024-64244025 on Build GRCh38
rs3177399 Chr.11: 64244130-64244130 on Build GRCh38
rs1054613672 Chr.11: 64244098-64244099 on Build GRCh38
rs552389491 Chr.11: 64243987-64243987 on Build GRCh38
rs2298631 Chr.11: 64244084-64244084 on Build GRCh38
rs775400361 Chr.11: 64243929-64243929 on Build GRCh38
rs922039189 Chr.11: 64244361-64244361 on Build GRCh38
rs762932526 Chr.11: 64243957-64243957 on Build GRCh38
rs891832307 Chr.11: 64244313-64244313 on Build GRCh38
rs774324964 Chr.11: 64244078-64244078 on Build GRCh38
rs369839073 Chr.11: 64244301-64244301 on Build GRCh38
rs772369485 Chr.11: 64244013-64244013 on Build GRCh38
rs751285598 Chr.11: 64243965-64243965 on Build GRCh38
rs750656365 Chr.11: 64243975-64243975 on Build GRCh38
rs654573 Chr.11: 64243969-64243969 on Build GRCh38
rs779595679 Chr.11: 64244008-64244008 on Build GRCh38
rs1003325768 Chr.11: 64244186-64244186 on Build GRCh38
rs570140207 Chr.11: 64244219-64244219 on Build GRCh38
rs201615451 Chr.11: 64244049-64244049 on Build GRCh38
rs931911129 Chr.11: 64244250-64244250 on Build GRCh38
rs966557889 Chr.11: 64244207-64244207 on Build GRCh38
rs568326806 Chr.11: 64244092-64244092 on Build GRCh38
rs138590813 Chr.11: 64243970-64243970 on Build GRCh38
rs35989122 Chr.11: 64244143-64244144 on Build GRCh38
rs905990008 Chr.11: 64244338-64244338 on Build GRCh38
rs776169540 Chr.11: 64243914-64243914 on Build GRCh38
rs902259908 Chr.11: 64244161-64244161 on Build GRCh38
rs1010256888 Chr.11: 64244324-64244324 on Build GRCh38
rs655896 Chr.11: 64244270-64244270 on Build GRCh38
rs896218834 Chr.11: 64244316-64244316 on Build GRCh38
rs181364026 Chr.11: 64243962-64243962 on Build GRCh38
rs1010539296 Chr.11: 64244134-64244134 on Build GRCh38
rs1030738749 Chr.11: 64244306-64244306 on Build GRCh38
rs748473913 Chr.11: 64244012-64244012 on Build GRCh38
rs762452775 Chr.11: 64244064-64244064 on Build GRCh38
rs774280499 Chr.11: 64244047-64244047 on Build GRCh38
rs932219270 Chr.11: 64244093-64244094 on Build GRCh38
rs1001714431 Chr.11: 64244345-64244345 on Build GRCh38
rs376378612 Chr.11: 64244015-64244015 on Build GRCh38
rs577453814 Chr.11: 64244184-64244184 on Build GRCh38
rs755478218 Chr.11: 64243988-64243988 on Build GRCh38
rs1013789284 Chr.11: 64244318-64244318 on Build GRCh38
rs368428015 Chr.11: 64244082-64244083 on Build GRCh38
rs756218134 Chr.11: 64243982-64243982 on Build GRCh38
rs764693999 Chr.11: 64243925-64243925 on Build GRCh38
rs1014410885 Chr.11: 64244043-64244043 on Build GRCh38
rs879864176 Chr.11: 64243964-64243964 on Build GRCh38
rs549858423 Chr.11: 64244213-64244213 on Build GRCh38
rs952009838 Chr.11: 64244100-64244100 on Build GRCh38
rs907902135 Chr.11: 64244289-64244289 on Build GRCh38
rs558756623 Chr.11: 64244290-64244290 on Build GRCh38
rs559583407 Chr.11: 64244062-64244062 on Build GRCh38
rs966142194 Chr.11: 64244294-64244294 on Build GRCh38
rs983436103 Chr.11: 64244129-64244129 on Build GRCh38
rs534856586 Chr.11: 64244305-64244305 on Build GRCh38
rs759062024 Chr.11: 64243917-64243917 on Build GRCh38
rs769329674 Chr.11: 64244057-64244057 on Build GRCh38
rs925208147 Chr.11: 64244282-64244282 on Build GRCh38
rs763137985 Chr.11: 64244242-64244242 on Build GRCh38
rs1045892237 Chr.11: 64244319-64244319 on Build GRCh38
rs3031454 Chr.11: 64244150-64244151 on Build GRCh38
rs780056779 Chr.11: 64243983-64243983 on Build GRCh38
rs975922838 Chr.11: 64244214-64244214 on Build GRCh38
rs768489822 Chr.11: 64243986-64243987 on Build GRCh38

View Full Product Details