Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53274633-53275015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPL1
Entrez Gene ID
9700
Gene Name
extra spindle pole bodies like 1, separase
Gene Aliases
ESP1, SEPA
UniGene
Hs.153479
Gene Chromosome Location
Chr.12: 53268257-53293643 on Build GRCh38
Transcripts
NM_012291.4 XM_006719705.3 XM_011539024.2 XM_011539025.2 XM_017020253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6393364 c.1594G>A p.A532T Chr.12: 53274904-53274904 on Build GRCh38
4043030 c.1635A>G p.P545P Chr.12: 53274945-53274945 on Build GRCh38
5841362 c.1598C>T p.A533V Chr.12: 53274908-53274908 on Build GRCh38
4681699 c.1552G>A p.G518S Chr.12: 53274862-53274862 on Build GRCh38
292310 c.1692A>G p.L564L Chr.12: 53275002-53275002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937570466 Chr.12: 53274738-53274738 on Build GRCh38
rs200381228 Chr.12: 53274887-53274887 on Build GRCh38
rs965586352 Chr.12: 53274749-53274749 on Build GRCh38
rs899694656 Chr.12: 53274944-53274944 on Build GRCh38
rs976819995 Chr.12: 53274752-53274753 on Build GRCh38
rs914838845 Chr.12: 53274789-53274789 on Build GRCh38
rs928857705 Chr.12: 53274733-53274733 on Build GRCh38
rs750244042 Chr.12: 53274715-53274715 on Build GRCh38
rs374148983 Chr.12: 53274792-53274792 on Build GRCh38
rs777469941 Chr.12: 53274772-53274772 on Build GRCh38
rs546107819 Chr.12: 53274986-53274986 on Build GRCh38
rs774976343 Chr.12: 53274964-53274964 on Build GRCh38
rs773367755 Chr.12: 53274916-53274916 on Build GRCh38
rs201612380 Chr.12: 53274814-53274814 on Build GRCh38
rs754709315 Chr.12: 53274753-53274753 on Build GRCh38
rs776505790 Chr.12: 53274954-53274954 on Build GRCh38
rs958496407 Chr.12: 53274722-53274722 on Build GRCh38
rs1055068505 Chr.12: 53274927-53274927 on Build GRCh38
rs918698393 Chr.12: 53274773-53274773 on Build GRCh38
rs949278125 Chr.12: 53274934-53274934 on Build GRCh38
rs750786661 Chr.12: 53274774-53274774 on Build GRCh38
rs753239624 Chr.12: 53274723-53274723 on Build GRCh38
rs984154992 Chr.12: 53274878-53274878 on Build GRCh38
rs762507993 Chr.12: 53274984-53274984 on Build GRCh38
rs765907251 Chr.12: 53274662-53274662 on Build GRCh38
rs765195130 Chr.12: 53274962-53274962 on Build GRCh38
rs575455248 Chr.12: 53274942-53274942 on Build GRCh38
rs371221962 Chr.12: 53274922-53274922 on Build GRCh38
rs902313163 Chr.12: 53274938-53274938 on Build GRCh38
rs760968100 Chr.12: 53274920-53274920 on Build GRCh38
rs377436215 Chr.12: 53274897-53274897 on Build GRCh38
rs1046115697 Chr.12: 53274935-53274935 on Build GRCh38
rs894944652 Chr.12: 53274930-53274930 on Build GRCh38
rs553137940 Chr.12: 53274730-53274730 on Build GRCh38
rs943844358 Chr.12: 53274931-53274931 on Build GRCh38
rs984774415 Chr.12: 53274692-53274692 on Build GRCh38
rs1014924001 Chr.12: 53274695-53274695 on Build GRCh38
rs563406109 Chr.12: 53274905-53274905 on Build GRCh38
rs749721242 Chr.12: 53274838-53274838 on Build GRCh38
rs759443544 Chr.12: 53274960-53274960 on Build GRCh38

View Full Product Details