Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52776344-52776905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPNAT1
Entrez Gene ID
64841
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Gene Aliases
GNA1, GNPNAT, Gpnat1
UniGene
Hs.478025
Gene Chromosome Location
Chr.14: 52775193-52791668 on Build GRCh38
Transcripts
NM_198066.3 XM_005268012.2 XM_006720238.3

SNP Detail

SNP ID Chromosome Location
rs981317339 Chr.14: 52776701-52776701 on Build GRCh38
rs1044308730 Chr.14: 52776584-52776585 on Build GRCh38
rs1027177769 Chr.14: 52776662-52776662 on Build GRCh38
rs558451336 Chr.14: 52776747-52776747 on Build GRCh38
rs890861342 Chr.14: 52776606-52776606 on Build GRCh38
rs552115062 Chr.14: 52776636-52776636 on Build GRCh38
rs532100084 Chr.14: 52776619-52776619 on Build GRCh38
rs536878237 Chr.14: 52776862-52776862 on Build GRCh38
rs1015673496 Chr.14: 52776615-52776615 on Build GRCh38
rs1053878886 Chr.14: 52776556-52776556 on Build GRCh38
rs1012676815 Chr.14: 52776582-52776582 on Build GRCh38
rs974842218 Chr.14: 52776748-52776748 on Build GRCh38
rs964349666 Chr.14: 52776419-52776419 on Build GRCh38
rs936771584 Chr.14: 52776533-52776533 on Build GRCh38
rs1008402238 Chr.14: 52776607-52776607 on Build GRCh38
rs377066177 Chr.14: 52776505-52776505 on Build GRCh38
rs117291444 Chr.14: 52776560-52776560 on Build GRCh38
rs568769277 Chr.14: 52776649-52776649 on Build GRCh38
rs368620691 Chr.14: 52776667-52776667 on Build GRCh38
rs147604994 Chr.14: 52776700-52776700 on Build GRCh38
rs918018367 Chr.14: 52776681-52776681 on Build GRCh38
rs1045940988 Chr.14: 52776476-52776476 on Build GRCh38
rs187390805 Chr.14: 52776668-52776668 on Build GRCh38
rs538442711 Chr.14: 52776655-52776655 on Build GRCh38
rs182327943 Chr.14: 52776434-52776434 on Build GRCh38
rs961621384 Chr.14: 52776648-52776648 on Build GRCh38
rs1005887471 Chr.14: 52776703-52776703 on Build GRCh38
rs949949689 Chr.14: 52776424-52776424 on Build GRCh38
rs762964067 Chr.14: 52776583-52776583 on Build GRCh38
rs908848517 Chr.14: 52776417-52776417 on Build GRCh38
rs1054135903 Chr.14: 52776520-52776520 on Build GRCh38
rs762175177 Chr.14: 52776833-52776833 on Build GRCh38
rs535403514 Chr.14: 52776834-52776834 on Build GRCh38
rs923528346 Chr.14: 52776802-52776802 on Build GRCh38
rs567847685 Chr.14: 52776788-52776788 on Build GRCh38
rs771521019 Chr.14: 52776437-52776437 on Build GRCh38
rs773940760 Chr.14: 52776713-52776713 on Build GRCh38
rs555932644 Chr.14: 52776481-52776481 on Build GRCh38
rs149932355 Chr.14: 52776529-52776529 on Build GRCh38
rs934468442 Chr.14: 52776716-52776716 on Build GRCh38
rs567959134 Chr.14: 52776728-52776729 on Build GRCh38
rs932368022 Chr.14: 52776846-52776846 on Build GRCh38
rs1049534191 Chr.14: 52776577-52776577 on Build GRCh38
rs924526221 Chr.14: 52776486-52776486 on Build GRCh38
rs561135905 Chr.14: 52776410-52776410 on Build GRCh38
rs776790125 Chr.14: 52776793-52776793 on Build GRCh38
rs535556742 Chr.14: 52776387-52776387 on Build GRCh38
rs575821075 Chr.14: 52776501-52776501 on Build GRCh38
rs895106234 Chr.14: 52776575-52776575 on Build GRCh38

View Full Product Details