Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 596547-597125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLUL1
Entrez Gene ID
27098
Gene Name
clusterin like 1
Gene Aliases
RA337M
UniGene
Hs.732341
Gene Chromosome Location
Chr.18: 596998-650298 on Build GRCh38
Transcripts
NM_001289036.1 NM_014410.4 XM_006722322.2 XM_011525648.2 XM_011525649.1 XM_011525652.1

SNP Detail

SNP ID Chromosome Location
rs190950707 Chr.18: 596953-596953 on Build GRCh38
rs771176572 Chr.18: 596663-596663 on Build GRCh38
rs891035347 Chr.18: 596762-596762 on Build GRCh38
rs981376905 Chr.18: 596673-596673 on Build GRCh38
rs934928769 Chr.18: 596752-596752 on Build GRCh38
rs867551073 Chr.18: 596824-596824 on Build GRCh38
rs908551486 Chr.18: 596719-596719 on Build GRCh38
rs559753027 Chr.18: 596819-596820 on Build GRCh38
rs184918064 Chr.18: 596945-596945 on Build GRCh38
rs926784307 Chr.18: 597069-597069 on Build GRCh38
rs1027600910 Chr.18: 596693-596693 on Build GRCh38
rs969819546 Chr.18: 596933-596933 on Build GRCh38
rs950510926 Chr.18: 596701-596701 on Build GRCh38
rs567826204 Chr.18: 596851-596851 on Build GRCh38
rs562129149 Chr.18: 597070-597070 on Build GRCh38
rs1014351840 Chr.18: 596637-596637 on Build GRCh38
rs398120087 Chr.18: 596586-596587 on Build GRCh38
rs62090065 Chr.18: 596941-596941 on Build GRCh38
rs927378879 Chr.18: 597062-597062 on Build GRCh38
rs557367333 Chr.18: 596995-596995 on Build GRCh38
rs932761057 Chr.18: 597006-597006 on Build GRCh38
rs540012461 Chr.18: 597010-597010 on Build GRCh38
rs114732404 Chr.18: 596734-596734 on Build GRCh38
rs540514505 Chr.18: 596735-596735 on Build GRCh38
rs561119356 Chr.18: 596733-596733 on Build GRCh38
rs112478972 Chr.18: 596710-596710 on Build GRCh38
rs980961942 Chr.18: 596955-596956 on Build GRCh38
rs1056434211 Chr.18: 596704-596704 on Build GRCh38
rs930729309 Chr.18: 597073-597073 on Build GRCh38
rs988907385 Chr.18: 596757-596757 on Build GRCh38
rs891487597 Chr.18: 596799-596799 on Build GRCh38
rs908522213 Chr.18: 597000-597000 on Build GRCh38
rs974558059 Chr.18: 596730-596730 on Build GRCh38
rs562166155 Chr.18: 596800-596800 on Build GRCh38
rs1049109063 Chr.18: 596949-596949 on Build GRCh38
rs1015778069 Chr.18: 597092-597092 on Build GRCh38
rs1034269939 Chr.18: 596964-596964 on Build GRCh38
rs1049057868 Chr.18: 597076-597076 on Build GRCh38
rs967788210 Chr.18: 597009-597009 on Build GRCh38
rs960948122 Chr.18: 596988-596988 on Build GRCh38
rs190953570 Chr.18: 596819-596819 on Build GRCh38
rs187477516 Chr.18: 597003-597003 on Build GRCh38
rs917429869 Chr.18: 596761-596761 on Build GRCh38
rs73942572 Chr.18: 596992-596992 on Build GRCh38
rs774552721 Chr.18: 597032-597032 on Build GRCh38
rs558332367 Chr.18: 597017-597017 on Build GRCh38
rs886442822 Chr.18: 597080-597080 on Build GRCh38
rs930652703 Chr.18: 596787-596787 on Build GRCh38
rs915391790 Chr.18: 596593-596594 on Build GRCh38
rs966151722 Chr.18: 596714-596714 on Build GRCh38
rs563673038 Chr.18: 596972-596973 on Build GRCh38
rs575764358 Chr.18: 596613-596613 on Build GRCh38
rs980577767 Chr.18: 597047-597047 on Build GRCh38
rs1033827888 Chr.18: 597046-597046 on Build GRCh38
rs532283901 Chr.18: 596764-596764 on Build GRCh38
rs1028765329 Chr.18: 597005-597005 on Build GRCh38
rs536934612 Chr.18: 596803-596803 on Build GRCh38
rs544115237 Chr.18: 597039-597039 on Build GRCh38
rs891745803 Chr.18: 597019-597019 on Build GRCh38
rs868500809 Chr.18: 597099-597099 on Build GRCh38
rs554808617 Chr.18: 597001-597001 on Build GRCh38
rs536727095 Chr.18: 596997-596997 on Build GRCh38
rs577076477 Chr.18: 596870-596870 on Build GRCh38
rs950174029 Chr.18: 596768-596768 on Build GRCh38
rs905900360 Chr.18: 596684-596684 on Build GRCh38
rs34324884 Chr.18: 596574-596575 on Build GRCh38
rs886326878 Chr.18: 596968-596968 on Build GRCh38
rs556868152 Chr.18: 596827-596827 on Build GRCh38
rs1045209890 Chr.18: 597050-597050 on Build GRCh38
rs183137131 Chr.18: 596956-596956 on Build GRCh38
rs11661285 Chr.18: 596749-596749 on Build GRCh38
rs1053040026 Chr.18: 597013-597013 on Build GRCh38
rs62090064 Chr.18: 596890-596890 on Build GRCh38
rs1018792699 Chr.18: 596712-596712 on Build GRCh38
rs961173328 Chr.18: 597052-597052 on Build GRCh38
rs866518142 Chr.18: 596839-596839 on Build GRCh38
rs576697854 Chr.18: 597033-597033 on Build GRCh38
rs569306868 Chr.18: 596996-596996 on Build GRCh38
rs542779393 Chr.18: 596648-596648 on Build GRCh38
rs865977073 Chr.18: 596905-596905 on Build GRCh38
rs1004694872 Chr.18: 596707-596707 on Build GRCh38
rs551745994 Chr.18: 596946-596948 on Build GRCh38
rs943947577 Chr.18: 597011-597011 on Build GRCh38
rs950276793 Chr.18: 597040-597040 on Build GRCh38
rs1021371925 Chr.18: 596840-596840 on Build GRCh38
rs550113164 Chr.18: 596792-596792 on Build GRCh38
rs533220686 Chr.18: 596844-596846 on Build GRCh38
rs765358545 Chr.18: 596839-596842 on Build GRCh38
rs905844262 Chr.18: 596783-596783 on Build GRCh38
rs71174258 Chr.18: 596961-596962 on Build GRCh38
rs1044517854 Chr.18: 596777-596777 on Build GRCh38
rs397759694 Chr.18: 596585-596586 on Build GRCh38
rs530344109 Chr.18: 597095-597095 on Build GRCh38
rs994747216 Chr.18: 596687-596687 on Build GRCh38
rs956249785 Chr.18: 597024-597024 on Build GRCh38
rs1019191981 Chr.18: 596999-596999 on Build GRCh38
rs1786126 Chr.18: 596742-596742 on Build GRCh38
rs926186192 Chr.18: 596744-596744 on Build GRCh38
rs989903686 Chr.18: 596590-596594 on Build GRCh38

View Full Product Details