Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 32406412-32406964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC6
Entrez Gene ID
57448
Gene Name
baculoviral IAP repeat containing 6
Gene Aliases
APOLLON, BRUCE
UniGene
Hs.150107
Gene Chromosome Location
Chr.2: 32356907-32618899 on Build GRCh38
Transcripts
NM_016252.3 XM_005264449.4 XM_005264450.4 XM_005264451.4 XM_005264452.4 XM_005264453.4 XM_005264454.4 XM_005264455.4 XM_006712054.3 XM_006712055.3 XM_006712056.3 XM_011533003.2 XM_011533005.2 XM_017004556.1 XM_017004557.1 XM_017004558.1 XM_017004559.1 XM_017004560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7161967 c.1439C>T p.S480L Chr.2: 32406519-32406519 on Build GRCh38
3839422 c.1455T>C p.H485H Chr.2: 32406535-32406535 on Build GRCh38
477326 c.1445G>A p.S482N Chr.2: 32406525-32406525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558439332 Chr.2: 32406846-32406846 on Build GRCh38
rs951296036 Chr.2: 32406783-32406783 on Build GRCh38
rs762497140 Chr.2: 32406601-32406602 on Build GRCh38
rs375215789 Chr.2: 32406796-32406796 on Build GRCh38
rs996121542 Chr.2: 32406476-32406480 on Build GRCh38
rs544811865 Chr.2: 32406825-32406825 on Build GRCh38
rs984907652 Chr.2: 32406688-32406688 on Build GRCh38
rs550400928 Chr.2: 32406525-32406525 on Build GRCh38
rs570208725 Chr.2: 32406610-32406610 on Build GRCh38
rs748019908 Chr.2: 32406551-32406551 on Build GRCh38
rs940339855 Chr.2: 32406450-32406450 on Build GRCh38
rs778847496 Chr.2: 32406502-32406502 on Build GRCh38
rs553042920 Chr.2: 32406629-32406629 on Build GRCh38
rs772650272 Chr.2: 32406484-32406485 on Build GRCh38
rs746171154 Chr.2: 32406504-32406504 on Build GRCh38
rs556075597 Chr.2: 32406770-32406770 on Build GRCh38
rs991976200 Chr.2: 32406786-32406786 on Build GRCh38
rs369537563 Chr.2: 32406908-32406908 on Build GRCh38
rs778813764 Chr.2: 32406815-32406815 on Build GRCh38
rs991105102 Chr.2: 32406621-32406621 on Build GRCh38
rs183661796 Chr.2: 32406469-32406469 on Build GRCh38
rs980921092 Chr.2: 32406833-32406833 on Build GRCh38
rs199750993 Chr.2: 32406455-32406455 on Build GRCh38
rs370543186 Chr.2: 32406603-32406603 on Build GRCh38
rs769411274 Chr.2: 32406575-32406575 on Build GRCh38
rs758558417 Chr.2: 32406505-32406505 on Build GRCh38
rs192965199 Chr.2: 32406850-32406850 on Build GRCh38
rs781325587 Chr.2: 32406535-32406535 on Build GRCh38
rs535455567 Chr.2: 32406722-32406722 on Build GRCh38
rs754314498 Chr.2: 32406464-32406464 on Build GRCh38
rs1025532070 Chr.2: 32406709-32406709 on Build GRCh38
rs749092036 Chr.2: 32406619-32406619 on Build GRCh38
rs764634456 Chr.2: 32406839-32406839 on Build GRCh38
rs375147219 Chr.2: 32406620-32406620 on Build GRCh38
rs770592689 Chr.2: 32406594-32406594 on Build GRCh38
rs936918536 Chr.2: 32406583-32406583 on Build GRCh38
rs780370959 Chr.2: 32406521-32406521 on Build GRCh38
rs746828459 Chr.2: 32406522-32406522 on Build GRCh38
rs529301698 Chr.2: 32406454-32406454 on Build GRCh38
rs566310579 Chr.2: 32406679-32406679 on Build GRCh38
rs188475925 Chr.2: 32406524-32406524 on Build GRCh38
rs371982466 Chr.2: 32406932-32406932 on Build GRCh38
rs936652766 Chr.2: 32406902-32406902 on Build GRCh38
rs768076915 Chr.2: 32406694-32406694 on Build GRCh38
rs868454889 Chr.2: 32406523-32406523 on Build GRCh38
rs746489749 Chr.2: 32406602-32406602 on Build GRCh38
rs560245579 Chr.2: 32406445-32406445 on Build GRCh38
rs962444715 Chr.2: 32406695-32406695 on Build GRCh38
rs922954150 Chr.2: 32406813-32406814 on Build GRCh38
rs948127960 Chr.2: 32406813-32406813 on Build GRCh38
rs903887385 Chr.2: 32406561-32406561 on Build GRCh38
rs767973888 Chr.2: 32406475-32406476 on Build GRCh38
rs974147320 Chr.2: 32406696-32406696 on Build GRCh38
rs773953479 Chr.2: 32406595-32406595 on Build GRCh38
rs539114598 Chr.2: 32406627-32406627 on Build GRCh38
rs947245034 Chr.2: 32406851-32406851 on Build GRCh38
rs74971431 Chr.2: 32406936-32406936 on Build GRCh38
rs1018349834 Chr.2: 32406673-32406673 on Build GRCh38
rs762466761 Chr.2: 32406593-32406593 on Build GRCh38
rs761068778 Chr.2: 32406597-32406597 on Build GRCh38
rs907612508 Chr.2: 32406696-32406697 on Build GRCh38
rs940454932 Chr.2: 32406774-32406774 on Build GRCh38
rs772601270 Chr.2: 32406589-32406589 on Build GRCh38

View Full Product Details