Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131532449-131532995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC74A
Entrez Gene ID
90557
Gene Name
coiled-coil domain containing 74A
Gene Aliases
-
UniGene
Hs.351461
Gene Chromosome Location
Chr.2: 131522621-131533666 on Build GRCh38
Transcripts
NM_001258304.1 NM_001258305.1 NM_001258306.1 NM_138770.2 XM_005263835.3 XM_005263836.4 XM_005263837.1 XM_006712828.3 XM_017005234.1 XM_017005235.1 XM_017005236.1 XM_017005237.1 XM_017005238.1 XM_017005239.1 XM_017005240.1 XM_017005241.1 XM_017005242.1 XM_017005243.1 XM_017005244.1 XM_017005245.1 XM_017005246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599035 c.881G>A p.R294Q Chr.2: 131532868-131532868 on Build GRCh38
354214 c.919C>A p.Q307K Chr.2: 131532906-131532906 on Build GRCh38
3115486 c.730A>T p.N244Y Chr.2: 131532635-131532635 on Build GRCh38
6153904 c.760G>T p.A254S Chr.2: 131532665-131532665 on Build GRCh38
4439372 c.708T>C p.N236N Chr.2: 131532613-131532613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200432356 Chr.2: 131532592-131532592 on Build GRCh38
rs774532043 Chr.2: 131532799-131532799 on Build GRCh38
rs761887443 Chr.2: 131532825-131532825 on Build GRCh38
rs369824578 Chr.2: 131532666-131532666 on Build GRCh38
rs368729304 Chr.2: 131532951-131532951 on Build GRCh38
rs761217763 Chr.2: 131532895-131532895 on Build GRCh38
rs752719318 Chr.2: 131532746-131532746 on Build GRCh38
rs202052126 Chr.2: 131532842-131532842 on Build GRCh38
rs766808431 Chr.2: 131532651-131532651 on Build GRCh38
rs775296893 Chr.2: 131532675-131532675 on Build GRCh38
rs573381369 Chr.2: 131532913-131532913 on Build GRCh38
rs529281590 Chr.2: 131532621-131532621 on Build GRCh38
rs747674570 Chr.2: 131532728-131532728 on Build GRCh38
rs372671510 Chr.2: 131532839-131532839 on Build GRCh38
rs777473723 Chr.2: 131532788-131532788 on Build GRCh38
rs545840828 Chr.2: 131532880-131532880 on Build GRCh38
rs368395160 Chr.2: 131532925-131532925 on Build GRCh38
rs758889615 Chr.2: 131532649-131532649 on Build GRCh38
rs781391784 Chr.2: 131532665-131532665 on Build GRCh38
rs763227336 Chr.2: 131532973-131532973 on Build GRCh38
rs201037266 Chr.2: 131532921-131532921 on Build GRCh38
rs371209501 Chr.2: 131532938-131532938 on Build GRCh38
rs773770666 Chr.2: 131532618-131532618 on Build GRCh38
rs769502307 Chr.2: 131532730-131532730 on Build GRCh38
rs780137326 Chr.2: 131532546-131532546 on Build GRCh38
rs141323157 Chr.2: 131532731-131532731 on Build GRCh38
rs201119299 Chr.2: 131532916-131532916 on Build GRCh38
rs534659950 Chr.2: 131532744-131532744 on Build GRCh38
rs756067437 Chr.2: 131532914-131532914 on Build GRCh38
rs751588999 Chr.2: 131532654-131532654 on Build GRCh38
rs569398337 Chr.2: 131532667-131532667 on Build GRCh38
rs1051842395 Chr.2: 131532841-131532841 on Build GRCh38
rs752658649 Chr.2: 131532844-131532844 on Build GRCh38
rs759009765 Chr.2: 131532545-131532545 on Build GRCh38
rs774819631 Chr.2: 131532743-131532743 on Build GRCh38
rs754902768 Chr.2: 131532778-131532778 on Build GRCh38
rs763737771 Chr.2: 131532626-131532626 on Build GRCh38
rs145104286 Chr.2: 131532917-131532917 on Build GRCh38
rs967839412 Chr.2: 131532697-131532697 on Build GRCh38
rs772504670 Chr.2: 131532801-131532801 on Build GRCh38
rs151240749 Chr.2: 131532680-131532680 on Build GRCh38
rs535098651 Chr.2: 131532804-131532804 on Build GRCh38
rs979309190 Chr.2: 131532491-131532491 on Build GRCh38
rs766831382 Chr.2: 131532683-131532683 on Build GRCh38
rs878991675 Chr.2: 131532656-131532656 on Build GRCh38
rs551127805 Chr.2: 131532641-131532641 on Build GRCh38
rs375423073 Chr.2: 131532943-131532943 on Build GRCh38
rs2599956 Chr.2: 131532669-131532669 on Build GRCh38
rs760139303 Chr.2: 131532685-131532685 on Build GRCh38
rs777987967 Chr.2: 131532704-131532704 on Build GRCh38
rs1006693904 Chr.2: 131532652-131532652 on Build GRCh38
rs138310081 Chr.2: 131532705-131532705 on Build GRCh38
rs750845123 Chr.2: 131532646-131532646 on Build GRCh38
rs763700969 Chr.2: 131532824-131532825 on Build GRCh38
rs377101795 Chr.2: 131532846-131532846 on Build GRCh38
rs779872188 Chr.2: 131532799-131532799 on Build GRCh38
rs748861919 Chr.2: 131532790-131532790 on Build GRCh38
rs149420878 Chr.2: 131532745-131532745 on Build GRCh38
rs57007130 Chr.2: 131532826-131532826 on Build GRCh38
rs1036402821 Chr.2: 131532795-131532795 on Build GRCh38
rs769414492 Chr.2: 131532782-131532782 on Build GRCh38
rs773653436 Chr.2: 131532681-131532681 on Build GRCh38
rs369236637 Chr.2: 131532557-131532557 on Build GRCh38
rs370686999 Chr.2: 131532850-131532850 on Build GRCh38
rs373531878 Chr.2: 131532727-131532727 on Build GRCh38
rs765663951 Chr.2: 131532642-131532642 on Build GRCh38
rs758090840 Chr.2: 131532930-131532930 on Build GRCh38
rs147150547 Chr.2: 131532867-131532867 on Build GRCh38
rs747038015 Chr.2: 131532676-131532676 on Build GRCh38
rs765299506 Chr.2: 131532892-131532892 on Build GRCh38
rs779670405 Chr.2: 131532769-131532769 on Build GRCh38
rs552095823 Chr.2: 131532695-131532695 on Build GRCh38
rs556509409 Chr.2: 131532824-131532824 on Build GRCh38
rs746258819 Chr.2: 131532800-131532800 on Build GRCh38
rs771991885 Chr.2: 131532671-131532671 on Build GRCh38
rs779960354 Chr.2: 131532718-131532718 on Build GRCh38
rs563030884 Chr.2: 131532480-131532480 on Build GRCh38
rs761248407 Chr.2: 131532954-131532954 on Build GRCh38
rs776421303 Chr.2: 131532631-131532631 on Build GRCh38
rs773465363 Chr.2: 131532972-131532972 on Build GRCh38
rs745789414 Chr.2: 131532609-131532610 on Build GRCh38
rs2599958 Chr.2: 131532613-131532613 on Build GRCh38
rs764229302 Chr.2: 131532750-131532750 on Build GRCh38
rs760751680 Chr.2: 131532747-131532747 on Build GRCh38
rs375622068 Chr.2: 131532858-131532858 on Build GRCh38
rs528788274 Chr.2: 131532531-131532531 on Build GRCh38
rs142708014 Chr.2: 131532678-131532678 on Build GRCh38
rs753996868 Chr.2: 131532752-131532752 on Build GRCh38
rs948069985 Chr.2: 131532548-131532548 on Build GRCh38
rs771179554 Chr.2: 131532737-131532737 on Build GRCh38
rs748854676 Chr.2: 131532849-131532849 on Build GRCh38
rs769169560 Chr.2: 131532869-131532869 on Build GRCh38
rs562111497 Chr.2: 131532594-131532594 on Build GRCh38
rs780344064 Chr.2: 131532936-131532936 on Build GRCh38
rs767555815 Chr.2: 131532691-131532691 on Build GRCh38
rs768305635 Chr.2: 131532841-131532842 on Build GRCh38
rs745589507 Chr.2: 131532861-131532861 on Build GRCh38
rs763315597 Chr.2: 131532904-131532904 on Build GRCh38
rs772061983 Chr.2: 131532675-131532676 on Build GRCh38
rs34466000 Chr.2: 131532974-131532974 on Build GRCh38
rs546099762 Chr.2: 131532498-131532498 on Build GRCh38
rs748434320 Chr.2: 131532570-131532570 on Build GRCh38
rs13660 Chr.2: 131532891-131532891 on Build GRCh38
rs770648703 Chr.2: 131532792-131532792 on Build GRCh38
rs564653470 Chr.2: 131532509-131532509 on Build GRCh38
rs373004533 Chr.2: 131532562-131532562 on Build GRCh38
rs757536484 Chr.2: 131532767-131532767 on Build GRCh38
rs144849829 Chr.2: 131532770-131532770 on Build GRCh38
rs750942406 Chr.2: 131532544-131532544 on Build GRCh38
rs769894697 Chr.2: 131532966-131532966 on Build GRCh38
rs947925383 Chr.2: 131532760-131532760 on Build GRCh38
rs76069883 Chr.2: 131532868-131532868 on Build GRCh38
rs751994968 Chr.2: 131532684-131532684 on Build GRCh38
rs761232318 Chr.2: 131532815-131532815 on Build GRCh38
rs574958611 Chr.2: 131532493-131532493 on Build GRCh38
rs770701028 Chr.2: 131532582-131532582 on Build GRCh38
rs746887280 Chr.2: 131532723-131532723 on Build GRCh38
rs376130814 Chr.2: 131532779-131532779 on Build GRCh38
rs752805377 Chr.2: 131532693-131532693 on Build GRCh38
rs202206116 Chr.2: 131532915-131532915 on Build GRCh38
rs755007478 Chr.2: 131532663-131532663 on Build GRCh38
rs776036970 Chr.2: 131532930-131532930 on Build GRCh38
rs772417364 Chr.2: 131532866-131532866 on Build GRCh38
rs767388757 Chr.2: 131532905-131532905 on Build GRCh38

View Full Product Details