Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232130350-232130751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L2
Entrez Gene ID
129563
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Gene Aliases
FAM6A, PRLMNS, hDIS3L2
UniGene
Hs.732236
Gene Chromosome Location
Chr.2: 231961583-232343968 on Build GRCh38
Transcripts
NM_001257281.1 NM_001257282.1 NM_152383.4 NR_046476.1 NR_046477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391035 c.665G>C p.R222T Chr.2: 232130682-232130682 on Build GRCh38
4391037 c.665G>C p.R222T Chr.2: 232130682-232130682 on Build GRCh38
319892 c.602-2A>G p.? Chr.2: 232130617-232130617 on Build GRCh38
7088222 c.616G>A p.D206N Chr.2: 232130633-232130633 on Build GRCh38
5834404 c.724delC p.P242fs*>8 Chr.2: 232130741-232130741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778439768 Chr.2: 232130599-232130602 on Build GRCh38
rs761214979 Chr.2: 232130624-232130624 on Build GRCh38
rs376643772 Chr.2: 232130603-232130603 on Build GRCh38
rs747478295 Chr.2: 232130572-232130572 on Build GRCh38
rs764220626 Chr.2: 232130681-232130681 on Build GRCh38
rs114918841 Chr.2: 232130450-232130450 on Build GRCh38
rs748745642 Chr.2: 232130704-232130705 on Build GRCh38
rs554088562 Chr.2: 232130569-232130569 on Build GRCh38
rs188984456 Chr.2: 232130509-232130509 on Build GRCh38
rs1006843527 Chr.2: 232130636-232130636 on Build GRCh38
rs199740245 Chr.2: 232130704-232130704 on Build GRCh38
rs756569370 Chr.2: 232130692-232130692 on Build GRCh38
rs201020526 Chr.2: 232130679-232130679 on Build GRCh38
rs751721861 Chr.2: 232130683-232130683 on Build GRCh38
rs192246283 Chr.2: 232130590-232130590 on Build GRCh38
rs773100809 Chr.2: 232130641-232130641 on Build GRCh38
rs752191280 Chr.2: 232130730-232130730 on Build GRCh38
rs976355699 Chr.2: 232130391-232130391 on Build GRCh38
rs752873692 Chr.2: 232130678-232130678 on Build GRCh38
rs747395729 Chr.2: 232130707-232130707 on Build GRCh38
rs748922151 Chr.2: 232130599-232130599 on Build GRCh38
rs781775529 Chr.2: 232130685-232130685 on Build GRCh38
rs968594468 Chr.2: 232130647-232130647 on Build GRCh38
rs73003124 Chr.2: 232130702-232130702 on Build GRCh38
rs1057445 Chr.2: 232130723-232130723 on Build GRCh38
rs962262191 Chr.2: 232130377-232130377 on Build GRCh38
rs775315695 Chr.2: 232130598-232130598 on Build GRCh38
rs373201666 Chr.2: 232130589-232130589 on Build GRCh38
rs962232288 Chr.2: 232130722-232130722 on Build GRCh38
rs201453807 Chr.2: 232130691-232130691 on Build GRCh38
rs538368984 Chr.2: 232130488-232130488 on Build GRCh38
rs150738622 Chr.2: 232130533-232130533 on Build GRCh38
rs747734015 Chr.2: 232130712-232130712 on Build GRCh38
rs985223747 Chr.2: 232130480-232130480 on Build GRCh38
rs757853425 Chr.2: 232130575-232130575 on Build GRCh38
rs922279136 Chr.2: 232130406-232130406 on Build GRCh38
rs760490430 Chr.2: 232130676-232130676 on Build GRCh38
rs758425762 Chr.2: 232130568-232130568 on Build GRCh38
rs773977505 Chr.2: 232130610-232130610 on Build GRCh38
rs571039032 Chr.2: 232130481-232130481 on Build GRCh38
rs568507225 Chr.2: 232130513-232130513 on Build GRCh38
rs1022276697 Chr.2: 232130640-232130640 on Build GRCh38
rs767490286 Chr.2: 232130627-232130627 on Build GRCh38
rs372259747 Chr.2: 232130578-232130578 on Build GRCh38
rs780410754 Chr.2: 232130700-232130700 on Build GRCh38
rs184764939 Chr.2: 232130729-232130729 on Build GRCh38
rs1055806361 Chr.2: 232130659-232130659 on Build GRCh38

View Full Product Details