Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232379747-232380296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPP
Entrez Gene ID
250
Gene Name
alkaline phosphatase, placental
Gene Aliases
ALP, PALP, PLAP, PLAP-1
UniGene
Hs.284255
Gene Chromosome Location
Chr.2: 232378534-232382889 on Build GRCh38
Transcripts
NM_001632.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795173 c.747G>A p.R249R Chr.2: 232380275-232380275 on Build GRCh38
6563334 c.567C>A p.N189K Chr.2: 232379846-232379846 on Build GRCh38
5696576 c.544T>G p.Y182D Chr.2: 232379823-232379823 on Build GRCh38
7427945 c.525C>T p.H175H Chr.2: 232379804-232379804 on Build GRCh38
4958853 c.535G>A p.A179T Chr.2: 232379814-232379814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048989 Chr.2: 232380227-232380227 on Build GRCh38
rs780847752 Chr.2: 232379797-232379797 on Build GRCh38
rs750469562 Chr.2: 232380201-232380201 on Build GRCh38
rs751673059 Chr.2: 232379925-232379925 on Build GRCh38
rs200005340 Chr.2: 232380240-232380240 on Build GRCh38
rs747794460 Chr.2: 232380146-232380146 on Build GRCh38
rs1130345 Chr.2: 232379936-232379936 on Build GRCh38
rs756648591 Chr.2: 232380137-232380137 on Build GRCh38
rs572913027 Chr.2: 232379986-232379986 on Build GRCh38
rs1037784140 Chr.2: 232380058-232380058 on Build GRCh38
rs1048988 Chr.2: 232380220-232380220 on Build GRCh38
rs765165696 Chr.2: 232380159-232380159 on Build GRCh38
rs145336626 Chr.2: 232379909-232379909 on Build GRCh38
rs746821321 Chr.2: 232379794-232379794 on Build GRCh38
rs559539627 Chr.2: 232379810-232379810 on Build GRCh38
rs759755247 Chr.2: 232379855-232379855 on Build GRCh38
rs746646720 Chr.2: 232380222-232380222 on Build GRCh38
rs748248964 Chr.2: 232379839-232379839 on Build GRCh38
rs779193102 Chr.2: 232380215-232380215 on Build GRCh38
rs529989675 Chr.2: 232380075-232380075 on Build GRCh38
rs756593411 Chr.2: 232379931-232379931 on Build GRCh38
rs768878566 Chr.2: 232379958-232379959 on Build GRCh38
rs542651128 Chr.2: 232379941-232379941 on Build GRCh38
rs552753986 Chr.2: 232379948-232379948 on Build GRCh38
rs569414542 Chr.2: 232380163-232380163 on Build GRCh38
rs1030258968 Chr.2: 232380119-232380119 on Build GRCh38
rs756125131 Chr.2: 232380203-232380203 on Build GRCh38
rs559120855 Chr.2: 232379903-232379903 on Build GRCh38
rs749567604 Chr.2: 232379798-232379798 on Build GRCh38
rs768375929 Chr.2: 232379875-232379875 on Build GRCh38
rs148987850 Chr.2: 232379862-232379862 on Build GRCh38
rs755257870 Chr.2: 232379833-232379833 on Build GRCh38
rs763584788 Chr.2: 232379826-232379826 on Build GRCh38
rs375542442 Chr.2: 232379902-232379902 on Build GRCh38
rs755391442 Chr.2: 232380140-232380140 on Build GRCh38
rs148017705 Chr.2: 232379865-232379865 on Build GRCh38
rs775182821 Chr.2: 232379861-232379861 on Build GRCh38
rs757845998 Chr.2: 232379825-232379825 on Build GRCh38
rs150558405 Chr.2: 232379807-232379807 on Build GRCh38
rs375831788 Chr.2: 232379983-232379983 on Build GRCh38
rs772707977 Chr.2: 232379806-232379806 on Build GRCh38
rs763638115 Chr.2: 232379863-232379863 on Build GRCh38
rs753892405 Chr.2: 232380275-232380275 on Build GRCh38
rs754581296 Chr.2: 232379775-232379775 on Build GRCh38
rs750382140 Chr.2: 232379908-232379908 on Build GRCh38
rs949634822 Chr.2: 232379990-232379997 on Build GRCh38
rs1048984 Chr.2: 232379795-232379795 on Build GRCh38
rs993463812 Chr.2: 232380074-232380074 on Build GRCh38
rs74337556 Chr.2: 232379906-232379906 on Build GRCh38
rs767501476 Chr.2: 232379907-232379907 on Build GRCh38
rs375078005 Chr.2: 232380197-232380197 on Build GRCh38
rs376708291 Chr.2: 232380157-232380157 on Build GRCh38
rs544091893 Chr.2: 232380059-232380059 on Build GRCh38
rs200913342 Chr.2: 232380161-232380161 on Build GRCh38
rs371349871 Chr.2: 232379881-232379881 on Build GRCh38
rs766368940 Chr.2: 232379952-232379952 on Build GRCh38
rs746375137 Chr.2: 232379900-232379900 on Build GRCh38
rs771732098 Chr.2: 232380149-232380149 on Build GRCh38
rs766430979 Chr.2: 232380204-232380204 on Build GRCh38
rs780421951 Chr.2: 232379933-232379933 on Build GRCh38
rs781328476 Chr.2: 232379926-232379926 on Build GRCh38
rs573678723 Chr.2: 232379940-232379940 on Build GRCh38
rs200893684 Chr.2: 232380134-232380134 on Build GRCh38
rs758663154 Chr.2: 232379878-232379878 on Build GRCh38
rs765112836 Chr.2: 232379964-232379964 on Build GRCh38
rs147969003 Chr.2: 232380205-232380205 on Build GRCh38
rs780818988 Chr.2: 232380226-232380226 on Build GRCh38
rs199856553 Chr.2: 232380153-232380153 on Build GRCh38
rs182497388 Chr.2: 232379889-232379889 on Build GRCh38
rs142656870 Chr.2: 232379841-232379841 on Build GRCh38
rs768298169 Chr.2: 232379939-232379939 on Build GRCh38
rs149986064 Chr.2: 232379834-232379834 on Build GRCh38
rs377737669 Chr.2: 232380239-232380239 on Build GRCh38
rs767545091 Chr.2: 232379982-232379982 on Build GRCh38
rs1045660401 Chr.2: 232379990-232379990 on Build GRCh38
rs369915782 Chr.2: 232379928-232379928 on Build GRCh38
rs778047262 Chr.2: 232379880-232379880 on Build GRCh38
rs758716290 Chr.2: 232379912-232379912 on Build GRCh38
rs374484845 Chr.2: 232380213-232380213 on Build GRCh38
rs764759340 Chr.2: 232379817-232379817 on Build GRCh38
rs753584591 Chr.2: 232379770-232379770 on Build GRCh38
rs2853374 Chr.2: 232379876-232379876 on Build GRCh38
rs55977653 Chr.2: 232380133-232380133 on Build GRCh38
rs774509069 Chr.2: 232379805-232379805 on Build GRCh38
rs757472787 Chr.2: 232379979-232379979 on Build GRCh38
rs769724986 Chr.2: 232380229-232380229 on Build GRCh38
rs781496701 Chr.2: 232379891-232379891 on Build GRCh38
rs773537802 Chr.2: 232380257-232380257 on Build GRCh38
rs905385913 Chr.2: 232379992-232379992 on Build GRCh38
rs565070796 Chr.2: 232380021-232380021 on Build GRCh38
rs770931424 Chr.2: 232379805-232379813 on Build GRCh38
rs12620827 Chr.2: 232379993-232379993 on Build GRCh38
rs764841819 Chr.2: 232379778-232379778 on Build GRCh38
rs778995657 Chr.2: 232379934-232379940 on Build GRCh38
rs372567488 Chr.2: 232379967-232379967 on Build GRCh38
rs779329094 Chr.2: 232380144-232380144 on Build GRCh38
rs373387017 Chr.2: 232379944-232379944 on Build GRCh38
rs138519491 Chr.2: 232380258-232380258 on Build GRCh38
rs773878040 Chr.2: 232379905-232379905 on Build GRCh38
rs1043802257 Chr.2: 232379843-232379843 on Build GRCh38
rs368860764 Chr.2: 232379961-232379961 on Build GRCh38
rs2853375 Chr.2: 232380151-232380151 on Build GRCh38
rs761966803 Chr.2: 232380176-232380176 on Build GRCh38
rs747043312 Chr.2: 232379942-232379943 on Build GRCh38
rs755444774 Chr.2: 232380211-232380211 on Build GRCh38
rs778075670 Chr.2: 232379917-232379917 on Build GRCh38
rs758365037 Chr.2: 232379793-232379793 on Build GRCh38
rs369220345 Chr.2: 232380138-232380138 on Build GRCh38
rs376800608 Chr.2: 232379946-232379946 on Build GRCh38
rs776572558 Chr.2: 232379957-232379957 on Build GRCh38
rs147926516 Chr.2: 232379854-232379854 on Build GRCh38
rs149065046 Chr.2: 232380224-232380224 on Build GRCh38
rs141504317 Chr.2: 232380054-232380054 on Build GRCh38
rs753056192 Chr.2: 232380219-232380219 on Build GRCh38
rs368010490 Chr.2: 232379864-232379864 on Build GRCh38
rs1048985 Chr.2: 232379804-232379804 on Build GRCh38
rs781093591 Chr.2: 232379827-232379827 on Build GRCh38
rs138033708 Chr.2: 232379904-232379904 on Build GRCh38
rs12620832 Chr.2: 232380067-232380067 on Build GRCh38
rs75604235 Chr.2: 232379858-232379858 on Build GRCh38
rs145551892 Chr.2: 232380274-232380274 on Build GRCh38
rs78051268 Chr.2: 232379820-232379820 on Build GRCh38
rs775183986 Chr.2: 232380235-232380235 on Build GRCh38
rs371726976 Chr.2: 232379809-232379809 on Build GRCh38
rs367631776 Chr.2: 232379783-232379783 on Build GRCh38
rs530857701 Chr.2: 232379842-232379842 on Build GRCh38
rs150864150 Chr.2: 232379816-232379816 on Build GRCh38

View Full Product Details