Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36739333-36739892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK3
Entrez Gene ID
85443
Gene Name
doublecortin like kinase 3
Gene Aliases
CLR, DCAMKL3, DCDC3C, DCK3
UniGene
Hs.631907
Gene Chromosome Location
Chr.3: 36712422-36764206 on Build GRCh38
Transcripts
NM_033403.1 XM_011534167.2 XM_011534168.2 XM_011534169.1

SNP Detail

SNP ID Chromosome Location
rs190703666 Chr.3: 36739600-36739600 on Build GRCh38
rs940545295 Chr.3: 36739431-36739431 on Build GRCh38
rs926702610 Chr.3: 36739650-36739651 on Build GRCh38
rs1026822907 Chr.3: 36739610-36739610 on Build GRCh38
rs907700749 Chr.3: 36739423-36739423 on Build GRCh38
rs931749833 Chr.3: 36739588-36739588 on Build GRCh38
rs943782061 Chr.3: 36739780-36739780 on Build GRCh38
rs1054001813 Chr.3: 36739416-36739416 on Build GRCh38
rs369992117 Chr.3: 36739693-36739693 on Build GRCh38
rs923108368 Chr.3: 36739744-36739744 on Build GRCh38
rs549831663 Chr.3: 36739372-36739372 on Build GRCh38
rs952864292 Chr.3: 36739681-36739681 on Build GRCh38
rs749471930 Chr.3: 36739660-36739660 on Build GRCh38
rs1050007893 Chr.3: 36739596-36739596 on Build GRCh38
rs113776412 Chr.3: 36739743-36739743 on Build GRCh38
rs1041421287 Chr.3: 36739534-36739534 on Build GRCh38
rs985148736 Chr.3: 36739419-36739419 on Build GRCh38
rs936916803 Chr.3: 36739403-36739403 on Build GRCh38
rs553726527 Chr.3: 36739732-36739732 on Build GRCh38
rs1055351229 Chr.3: 36739484-36739495 on Build GRCh38
rs1039414036 Chr.3: 36739785-36739785 on Build GRCh38
rs60103506 Chr.3: 36739737-36739737 on Build GRCh38
rs984708419 Chr.3: 36739685-36739685 on Build GRCh38
rs112323060 Chr.3: 36739864-36739864 on Build GRCh38
rs750833500 Chr.3: 36739587-36739587 on Build GRCh38
rs565828072 Chr.3: 36739606-36739606 on Build GRCh38
rs892809898 Chr.3: 36739599-36739599 on Build GRCh38
rs768861429 Chr.3: 36739675-36739675 on Build GRCh38
rs143104946 Chr.3: 36739629-36739629 on Build GRCh38
rs907813652 Chr.3: 36739707-36739707 on Build GRCh38
rs561973072 Chr.3: 36739448-36739448 on Build GRCh38
rs374782048 Chr.3: 36739617-36739617 on Build GRCh38
rs765542279 Chr.3: 36739535-36739535 on Build GRCh38
rs538387238 Chr.3: 36739637-36739637 on Build GRCh38
rs1004153006 Chr.3: 36739622-36739622 on Build GRCh38
rs778930964 Chr.3: 36739691-36739691 on Build GRCh38
rs560474006 Chr.3: 36739729-36739729 on Build GRCh38
rs757771006 Chr.3: 36739426-36739426 on Build GRCh38
rs1013013838 Chr.3: 36739648-36739648 on Build GRCh38
rs781641880 Chr.3: 36739493-36739493 on Build GRCh38
rs903332405 Chr.3: 36739611-36739611 on Build GRCh38
rs535502156 Chr.3: 36739713-36739713 on Build GRCh38
rs539518816 Chr.3: 36739615-36739615 on Build GRCh38
rs182546267 Chr.3: 36739650-36739650 on Build GRCh38
rs890851407 Chr.3: 36739539-36739539 on Build GRCh38
rs528892490 Chr.3: 36739551-36739551 on Build GRCh38
rs1020995582 Chr.3: 36739605-36739605 on Build GRCh38
rs545321438 Chr.3: 36739819-36739819 on Build GRCh38
rs991465583 Chr.3: 36739367-36739367 on Build GRCh38
rs930544100 Chr.3: 36739381-36739381 on Build GRCh38
rs115028505 Chr.3: 36739638-36739638 on Build GRCh38

View Full Product Details