Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 58494957-58495532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPT
Entrez Gene ID
202309
Gene Name
GRB2-binding adaptor protein, transmembrane
Gene Aliases
C5orf29
UniGene
Hs.547697
Gene Chromosome Location
Chr.5: 58491435-58496358 on Build GRCh38
Transcripts
NM_001304428.1 NM_001304429.1 NM_001304431.1 NM_152687.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5680865 c.434C>T p.P145L Chr.5: 58494970-58494970 on Build GRCh38
4780583 c.429A>G p.E143E Chr.5: 58494965-58494965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999372110 Chr.5: 58495285-58495285 on Build GRCh38
rs779426661 Chr.5: 58495009-58495009 on Build GRCh38
rs866119679 Chr.5: 58495214-58495214 on Build GRCh38
rs947978222 Chr.5: 58495410-58495413 on Build GRCh38
rs535285452 Chr.5: 58495210-58495210 on Build GRCh38
rs761636636 Chr.5: 58495032-58495032 on Build GRCh38
rs192759468 Chr.5: 58495027-58495027 on Build GRCh38
rs1031046512 Chr.5: 58495436-58495436 on Build GRCh38
rs906546883 Chr.5: 58495425-58495425 on Build GRCh38
rs187738631 Chr.5: 58495359-58495359 on Build GRCh38
rs984431889 Chr.5: 58495399-58495399 on Build GRCh38
rs902391062 Chr.5: 58495260-58495260 on Build GRCh38
rs769405081 Chr.5: 58494995-58494995 on Build GRCh38
rs556010725 Chr.5: 58495124-58495124 on Build GRCh38
rs757004472 Chr.5: 58494987-58494987 on Build GRCh38
rs747682256 Chr.5: 58495367-58495367 on Build GRCh38
rs567864165 Chr.5: 58495160-58495160 on Build GRCh38
rs879469033 Chr.5: 58495134-58495134 on Build GRCh38
rs767821062 Chr.5: 58495152-58495152 on Build GRCh38
rs943675775 Chr.5: 58495328-58495328 on Build GRCh38
rs1048345094 Chr.5: 58495100-58495100 on Build GRCh38
rs937498614 Chr.5: 58495497-58495497 on Build GRCh38
rs761116671 Chr.5: 58495177-58495177 on Build GRCh38
rs755371498 Chr.5: 58494982-58494982 on Build GRCh38
rs908090602 Chr.5: 58495087-58495087 on Build GRCh38
rs1036802311 Chr.5: 58495329-58495329 on Build GRCh38
rs965900732 Chr.5: 58495305-58495305 on Build GRCh38
rs575526071 Chr.5: 58495320-58495320 on Build GRCh38
rs908121362 Chr.5: 58495423-58495423 on Build GRCh38
rs771650766 Chr.5: 58495404-58495404 on Build GRCh38
rs1031015289 Chr.5: 58495220-58495220 on Build GRCh38
rs1389308 Chr.5: 58495484-58495484 on Build GRCh38
rs368087484 Chr.5: 58495023-58495023 on Build GRCh38
rs745497011 Chr.5: 58494990-58494990 on Build GRCh38
rs796564195 Chr.5: 58495165-58495165 on Build GRCh38
rs79700589 Chr.5: 58495293-58495293 on Build GRCh38
rs149917833 Chr.5: 58495323-58495323 on Build GRCh38
rs999908283 Chr.5: 58495428-58495428 on Build GRCh38
rs1011505094 Chr.5: 58495186-58495186 on Build GRCh38
rs760799273 Chr.5: 58495043-58495043 on Build GRCh38
rs775876682 Chr.5: 58495159-58495159 on Build GRCh38
rs529344797 Chr.5: 58495323-58495323 on Build GRCh38
rs779430118 Chr.5: 58494984-58494984 on Build GRCh38
rs115736777 Chr.5: 58495284-58495284 on Build GRCh38
rs542489672 Chr.5: 58495441-58495441 on Build GRCh38
rs35850273 Chr.5: 58495183-58495184 on Build GRCh38
rs931142252 Chr.5: 58495345-58495345 on Build GRCh38
rs577535018 Chr.5: 58495143-58495143 on Build GRCh38
rs766481762 Chr.5: 58495047-58495047 on Build GRCh38
rs75921367 Chr.5: 58495040-58495040 on Build GRCh38
rs1029100680 Chr.5: 58495489-58495489 on Build GRCh38
rs751295160 Chr.5: 58495507-58495507 on Build GRCh38
rs895103022 Chr.5: 58495388-58495388 on Build GRCh38
rs544664757 Chr.5: 58495327-58495327 on Build GRCh38
rs966150130 Chr.5: 58494991-58494991 on Build GRCh38
rs951705718 Chr.5: 58495344-58495344 on Build GRCh38
rs753799684 Chr.5: 58495057-58495057 on Build GRCh38
rs764628393 Chr.5: 58495258-58495258 on Build GRCh38
rs113958678 Chr.5: 58494985-58494985 on Build GRCh38
rs58414332 Chr.5: 58495036-58495036 on Build GRCh38
rs558448273 Chr.5: 58495247-58495247 on Build GRCh38
rs192370435 Chr.5: 58495482-58495482 on Build GRCh38
rs796800599 Chr.5: 58495493-58495493 on Build GRCh38
rs182836147 Chr.5: 58495337-58495337 on Build GRCh38
rs940886560 Chr.5: 58495141-58495141 on Build GRCh38
rs922193256 Chr.5: 58495273-58495273 on Build GRCh38
rs770540656 Chr.5: 58495060-58495060 on Build GRCh38
rs985068930 Chr.5: 58495077-58495077 on Build GRCh38
rs749255701 Chr.5: 58495015-58495015 on Build GRCh38

View Full Product Details