Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117286975-117287486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE1
Entrez Gene ID
23621
Gene Name
beta-secretase 1
Gene Aliases
ASP2, BACE, HSPC104
UniGene
Hs.504003
Gene Chromosome Location
Chr.11: 117285686-117316256 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3

SNP Detail

SNP ID Chromosome Location
rs1053018643 Chr.11: 117287346-117287346 on Build GRCh38
rs187577640 Chr.11: 117287401-117287401 on Build GRCh38
rs1003575926 Chr.11: 117287276-117287276 on Build GRCh38
rs534893415 Chr.11: 117287204-117287204 on Build GRCh38
rs912568386 Chr.11: 117287296-117287296 on Build GRCh38
rs747147618 Chr.11: 117287123-117287123 on Build GRCh38
rs182042107 Chr.11: 117287389-117287389 on Build GRCh38
rs554273984 Chr.11: 117287441-117287441 on Build GRCh38
rs991674373 Chr.11: 117287353-117287353 on Build GRCh38
rs1001055346 Chr.11: 117287338-117287338 on Build GRCh38
rs375256678 Chr.11: 117287275-117287275 on Build GRCh38
rs573620079 Chr.11: 117287010-117287010 on Build GRCh38
rs904100160 Chr.11: 117287329-117287329 on Build GRCh38
rs12288248 Chr.11: 117287305-117287305 on Build GRCh38
rs929903403 Chr.11: 117287166-117287166 on Build GRCh38
rs1040013322 Chr.11: 117287324-117287324 on Build GRCh38
rs138603306 Chr.11: 117287185-117287185 on Build GRCh38
rs28989510 Chr.11: 117287169-117287169 on Build GRCh38
rs1032799103 Chr.11: 117287061-117287061 on Build GRCh38
rs549692327 Chr.11: 117287051-117287051 on Build GRCh38
rs752139307 Chr.11: 117287265-117287265 on Build GRCh38
rs1038057070 Chr.11: 117287083-117287083 on Build GRCh38
rs1011503229 Chr.11: 117287225-117287225 on Build GRCh38
rs916308089 Chr.11: 117287372-117287372 on Build GRCh38
rs45497100 Chr.11: 117287452-117287452 on Build GRCh38
rs890988808 Chr.11: 117287177-117287177 on Build GRCh38
rs1020270115 Chr.11: 117287227-117287227 on Build GRCh38
rs962552231 Chr.11: 117287207-117287207 on Build GRCh38
rs552151980 Chr.11: 117287264-117287264 on Build GRCh38
rs979993114 Chr.11: 117287196-117287196 on Build GRCh38
rs371136858 Chr.11: 117287351-117287351 on Build GRCh38
rs764076131 Chr.11: 117287458-117287459 on Build GRCh38
rs918495534 Chr.11: 117287097-117287097 on Build GRCh38
rs878928164 Chr.11: 117287060-117287060 on Build GRCh38
rs973717109 Chr.11: 117287442-117287442 on Build GRCh38
rs916339360 Chr.11: 117287000-117287000 on Build GRCh38
rs974310915 Chr.11: 117287233-117287233 on Build GRCh38
rs528722309 Chr.11: 117287443-117287444 on Build GRCh38
rs555630776 Chr.11: 117287432-117287434 on Build GRCh38
rs369798383 Chr.11: 117287314-117287314 on Build GRCh38
rs893241319 Chr.11: 117287212-117287212 on Build GRCh38

View Full Product Details