Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118406845-118407350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5L
Entrez Gene ID
10632
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit G
Gene Aliases
ATP5JG
UniGene
Hs.486360
Gene Chromosome Location
Chr.11: 118401389-118409847 on Build GRCh38
Transcripts
NM_006476.4 NR_033759.1 NR_046369.1 NR_046370.1

Gene Symbol
LOC100131626
Entrez Gene ID
100131626
Gene Name
uncharacterized LOC100131626
Gene Aliases
-
UniGene
Hs.721614
Gene Chromosome Location
Chr.11: 118381220-118418815 on Build GRCh38
Transcripts
NM_006476.4 NR_033759.1 NR_046369.1 NR_046370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923566 c.131C>T p.A44V Chr.11: 118407015-118407015 on Build GRCh38
5645965 c.144A>T p.R48S Chr.11: 118407028-118407028 on Build GRCh38
3443935 c.126C>T p.T42T Chr.11: 118407010-118407010 on Build GRCh38
259571 c.68C>T p.S23L Chr.11: 118406952-118406952 on Build GRCh38
5143826 c.160delA p.I56fs*1 Chr.11: 118407044-118407044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11549141 Chr.11: 118407033-118407033 on Build GRCh38
rs781810238 Chr.11: 118407075-118407075 on Build GRCh38
rs782588894 Chr.11: 118406995-118406995 on Build GRCh38
rs571756379 Chr.11: 118406961-118406961 on Build GRCh38
rs782399738 Chr.11: 118406950-118406950 on Build GRCh38
rs917077619 Chr.11: 118407286-118407286 on Build GRCh38
rs1802487 Chr.11: 118407043-118407043 on Build GRCh38
rs949949302 Chr.11: 118407299-118407299 on Build GRCh38
rs782802669 Chr.11: 118406988-118406988 on Build GRCh38
rs148780758 Chr.11: 118407238-118407238 on Build GRCh38
rs782767043 Chr.11: 118406983-118406983 on Build GRCh38
rs1015754103 Chr.11: 118406965-118406965 on Build GRCh38
rs782323803 Chr.11: 118407060-118407060 on Build GRCh38
rs181465227 Chr.11: 118407027-118407027 on Build GRCh38
rs782143908 Chr.11: 118407081-118407081 on Build GRCh38
rs782167849 Chr.11: 118407069-118407069 on Build GRCh38
rs934396860 Chr.11: 118407217-118407217 on Build GRCh38
rs551867454 Chr.11: 118407064-118407064 on Build GRCh38
rs781920279 Chr.11: 118407144-118407144 on Build GRCh38
rs988595224 Chr.11: 118407284-118407284 on Build GRCh38
rs368307639 Chr.11: 118406953-118406953 on Build GRCh38
rs1802489 Chr.11: 118407067-118407067 on Build GRCh38
rs533476820 Chr.11: 118406984-118406984 on Build GRCh38
rs782179113 Chr.11: 118407057-118407057 on Build GRCh38
rs782644885 Chr.11: 118406907-118406907 on Build GRCh38
rs782144573 Chr.11: 118406968-118406968 on Build GRCh38
rs1026949162 Chr.11: 118406884-118406884 on Build GRCh38
rs202019744 Chr.11: 118407129-118407129 on Build GRCh38
rs782698562 Chr.11: 118407256-118407256 on Build GRCh38
rs566222326 Chr.11: 118406960-118406960 on Build GRCh38
rs782199810 Chr.11: 118407148-118407148 on Build GRCh38
rs782641428 Chr.11: 118407016-118407016 on Build GRCh38
rs781818934 Chr.11: 118407005-118407005 on Build GRCh38
rs145345193 Chr.11: 118407121-118407121 on Build GRCh38
rs782484652 Chr.11: 118406901-118406901 on Build GRCh38
rs782088006 Chr.11: 118407079-118407079 on Build GRCh38
rs782416089 Chr.11: 118406971-118406971 on Build GRCh38
rs782025040 Chr.11: 118407068-118407068 on Build GRCh38
rs781889135 Chr.11: 118406908-118406909 on Build GRCh38
rs782211129 Chr.11: 118407120-118407120 on Build GRCh38
rs534029093 Chr.11: 118406989-118406989 on Build GRCh38
rs375395914 Chr.11: 118407030-118407030 on Build GRCh38
rs782538725 Chr.11: 118406931-118406931 on Build GRCh38
rs782612653 Chr.11: 118407058-118407058 on Build GRCh38
rs782529931 Chr.11: 118407104-118407104 on Build GRCh38
rs905796462 Chr.11: 118406870-118406870 on Build GRCh38
rs141559547 Chr.11: 118407091-118407091 on Build GRCh38
rs1044247091 Chr.11: 118407314-118407314 on Build GRCh38
rs372897017 Chr.11: 118406957-118406957 on Build GRCh38
rs376128440 Chr.11: 118407138-118407138 on Build GRCh38
rs782417321 Chr.11: 118406913-118406914 on Build GRCh38
rs372834761 Chr.11: 118406924-118406924 on Build GRCh38
rs148370674 Chr.11: 118407055-118407055 on Build GRCh38
rs782582356 Chr.11: 118406960-118406960 on Build GRCh38
rs527436462 Chr.11: 118407210-118407210 on Build GRCh38
rs782700007 Chr.11: 118407070-118407070 on Build GRCh38
rs782432403 Chr.11: 118407108-118407108 on Build GRCh38
rs782003268 Chr.11: 118406964-118406964 on Build GRCh38
rs782581744 Chr.11: 118407116-118407116 on Build GRCh38
rs558056726 Chr.11: 118406902-118406902 on Build GRCh38
rs556449658 Chr.11: 118407105-118407105 on Build GRCh38
rs782202197 Chr.11: 118407092-118407093 on Build GRCh38
rs782058231 Chr.11: 118406977-118406977 on Build GRCh38
rs533139 Chr.11: 118406921-118406921 on Build GRCh38
rs781973112 Chr.11: 118407061-118407061 on Build GRCh38
rs782321752 Chr.11: 118407128-118407128 on Build GRCh38
rs78889201 Chr.11: 118406883-118406883 on Build GRCh38
rs782683609 Chr.11: 118406934-118406934 on Build GRCh38
rs781897545 Chr.11: 118407106-118407106 on Build GRCh38
rs781915602 Chr.11: 118406973-118406973 on Build GRCh38
rs564929019 Chr.11: 118407161-118407161 on Build GRCh38
rs782180381 Chr.11: 118406951-118406951 on Build GRCh38
rs782476833 Chr.11: 118407011-118407011 on Build GRCh38
rs782136985 Chr.11: 118406985-118406985 on Build GRCh38
rs17851277 Chr.11: 118407054-118407054 on Build GRCh38
rs782745180 Chr.11: 118407085-118407085 on Build GRCh38
rs200138806 Chr.11: 118406992-118406992 on Build GRCh38

View Full Product Details