Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118581159-118581664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4018368 c.225C>T p.N75N Chr.11: 118581467-118581467 on Build GRCh38
4018367 c.21G>A p.A7A Chr.11: 118581263-118581263 on Build GRCh38
4913381 c.252G>A p.R84R Chr.11: 118581494-118581494 on Build GRCh38
923617 c.251G>T p.R84M Chr.11: 118581493-118581493 on Build GRCh38
6602825 c.83G>A p.R28Q Chr.11: 118581325-118581325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782236845 Chr.11: 118581438-118581438 on Build GRCh38
rs782792709 Chr.11: 118581362-118581362 on Build GRCh38
rs142869705 Chr.11: 118581263-118581263 on Build GRCh38
rs11216911 Chr.11: 118581186-118581186 on Build GRCh38
rs781824770 Chr.11: 118581383-118581383 on Build GRCh38
rs533031438 Chr.11: 118581199-118581199 on Build GRCh38
rs781790703 Chr.11: 118581549-118581549 on Build GRCh38
rs782499577 Chr.11: 118581377-118581377 on Build GRCh38
rs374764727 Chr.11: 118581238-118581238 on Build GRCh38
rs782550649 Chr.11: 118581243-118581243 on Build GRCh38
rs781868275 Chr.11: 118581226-118581226 on Build GRCh38
rs558571200 Chr.11: 118581568-118581568 on Build GRCh38
rs373088344 Chr.11: 118581423-118581423 on Build GRCh38
rs369897117 Chr.11: 118581413-118581413 on Build GRCh38
rs782569438 Chr.11: 118581410-118581410 on Build GRCh38
rs782724596 Chr.11: 118581349-118581349 on Build GRCh38
rs367554879 Chr.11: 118581319-118581319 on Build GRCh38
rs530734033 Chr.11: 118581405-118581405 on Build GRCh38
rs371768462 Chr.11: 118581339-118581339 on Build GRCh38
rs372671683 Chr.11: 118581212-118581212 on Build GRCh38
rs577020755 Chr.11: 118581600-118581600 on Build GRCh38
rs782304859 Chr.11: 118581302-118581302 on Build GRCh38
rs11546473 Chr.11: 118581346-118581346 on Build GRCh38
rs782668654 Chr.11: 118581424-118581424 on Build GRCh38
rs886040858 Chr.11: 118581399-118581400 on Build GRCh38
rs782199352 Chr.11: 118581415-118581415 on Build GRCh38
rs782522980 Chr.11: 118581262-118581262 on Build GRCh38
rs782127122 Chr.11: 118581326-118581326 on Build GRCh38
rs376378749 Chr.11: 118581519-118581519 on Build GRCh38
rs781929294 Chr.11: 118581357-118581357 on Build GRCh38
rs782795777 Chr.11: 118581378-118581378 on Build GRCh38
rs782677904 Chr.11: 118581251-118581251 on Build GRCh38
rs782757383 Chr.11: 118581220-118581220 on Build GRCh38
rs969316162 Chr.11: 118581629-118581629 on Build GRCh38
rs782739403 Chr.11: 118581528-118581528 on Build GRCh38
rs956550187 Chr.11: 118581612-118581612 on Build GRCh38
rs782804215 Chr.11: 118581553-118581553 on Build GRCh38
rs146475733 Chr.11: 118581317-118581317 on Build GRCh38
rs1024746532 Chr.11: 118581628-118581628 on Build GRCh38
rs11216912 Chr.11: 118581619-118581619 on Build GRCh38
rs782075272 Chr.11: 118581361-118581361 on Build GRCh38
rs782495814 Chr.11: 118581228-118581228 on Build GRCh38
rs781938118 Chr.11: 118581305-118581305 on Build GRCh38
rs147424028 Chr.11: 118581596-118581596 on Build GRCh38
rs782002072 Chr.11: 118581525-118581525 on Build GRCh38
rs138863361 Chr.11: 118581258-118581258 on Build GRCh38
rs782231699 Chr.11: 118581288-118581288 on Build GRCh38
rs781907942 Chr.11: 118581366-118581366 on Build GRCh38
rs782377528 Chr.11: 118581498-118581498 on Build GRCh38
rs546954815 Chr.11: 118581635-118581635 on Build GRCh38
rs1030726234 Chr.11: 118581607-118581607 on Build GRCh38
rs782271411 Chr.11: 118581308-118581308 on Build GRCh38
rs549252303 Chr.11: 118581581-118581581 on Build GRCh38
rs782145607 Chr.11: 118581216-118581216 on Build GRCh38
rs782170049 Chr.11: 118581524-118581524 on Build GRCh38
rs781941527 Chr.11: 118581505-118581505 on Build GRCh38
rs886040859 Chr.11: 118581502-118581502 on Build GRCh38
rs782189649 Chr.11: 118581507-118581507 on Build GRCh38
rs782022381 Chr.11: 118581521-118581521 on Build GRCh38

View Full Product Details