Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118583270-118583819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5927624 c.364C>T p.R122W Chr.11: 118583275-118583275 on Build GRCh38
5435605 c.403G>A p.E135K Chr.11: 118583314-118583314 on Build GRCh38
7076136 c.418G>A p.E140K Chr.11: 118583329-118583329 on Build GRCh38
6290215 c.458G>A p.R153H Chr.11: 118583819-118583819 on Build GRCh38
6865419 c.382G>C p.A128P Chr.11: 118583293-118583293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945348317 Chr.11: 118583556-118583556 on Build GRCh38
rs781864494 Chr.11: 118583346-118583346 on Build GRCh38
rs143812760 Chr.11: 118583633-118583633 on Build GRCh38
rs781987168 Chr.11: 118583408-118583408 on Build GRCh38
rs549411122 Chr.11: 118583689-118583689 on Build GRCh38
rs782225824 Chr.11: 118583370-118583370 on Build GRCh38
rs71469182 Chr.11: 118583607-118583608 on Build GRCh38
rs190453820 Chr.11: 118583465-118583465 on Build GRCh38
rs782420179 Chr.11: 118583393-118583393 on Build GRCh38
rs782037970 Chr.11: 118583784-118583784 on Build GRCh38
rs782785959 Chr.11: 118583792-118583792 on Build GRCh38
rs201640805 Chr.11: 118583770-118583770 on Build GRCh38
rs782039572 Chr.11: 118583767-118583767 on Build GRCh38
rs78574894 Chr.11: 118583455-118583455 on Build GRCh38
rs782455164 Chr.11: 118583617-118583617 on Build GRCh38
rs17842763 Chr.11: 118583462-118583462 on Build GRCh38
rs184379177 Chr.11: 118583433-118583433 on Build GRCh38
rs556971767 Chr.11: 118583349-118583349 on Build GRCh38
rs35189263 Chr.11: 118583597-118583598 on Build GRCh38
rs915257919 Chr.11: 118583468-118583468 on Build GRCh38
rs782383131 Chr.11: 118583769-118583770 on Build GRCh38
rs371290581 Chr.11: 118583327-118583327 on Build GRCh38
rs565156126 Chr.11: 118583586-118583586 on Build GRCh38
rs782689458 Chr.11: 118583343-118583343 on Build GRCh38
rs371110213 Chr.11: 118583793-118583793 on Build GRCh38
rs562540656 Chr.11: 118583734-118583734 on Build GRCh38
rs781811111 Chr.11: 118583324-118583324 on Build GRCh38
rs113120334 Chr.11: 118583563-118583563 on Build GRCh38
rs370697952 Chr.11: 118583672-118583672 on Build GRCh38
rs111590377 Chr.11: 118583770-118583770 on Build GRCh38
rs182216691 Chr.11: 118583683-118583683 on Build GRCh38
rs566377900 Chr.11: 118583583-118583583 on Build GRCh38
rs74319736 Chr.11: 118583456-118583456 on Build GRCh38
rs939445492 Chr.11: 118583570-118583570 on Build GRCh38
rs368207677 Chr.11: 118583781-118583781 on Build GRCh38
rs370719275 Chr.11: 118583379-118583379 on Build GRCh38
rs906657637 Chr.11: 118583569-118583569 on Build GRCh38
rs985722978 Chr.11: 118583444-118583444 on Build GRCh38
rs972495380 Chr.11: 118583394-118583394 on Build GRCh38
rs782341289 Chr.11: 118583758-118583758 on Build GRCh38
rs77830210 Chr.11: 118583457-118583457 on Build GRCh38
rs782307557 Chr.11: 118583375-118583375 on Build GRCh38
rs201154675 Chr.11: 118583766-118583766 on Build GRCh38
rs535975072 Chr.11: 118583445-118583445 on Build GRCh38
rs114802679 Chr.11: 118583430-118583430 on Build GRCh38
rs782122598 Chr.11: 118583790-118583790 on Build GRCh38
rs374333744 Chr.11: 118583350-118583350 on Build GRCh38
rs1055741827 Chr.11: 118583621-118583621 on Build GRCh38
rs1017169460 Chr.11: 118583703-118583703 on Build GRCh38
rs782259569 Chr.11: 118583410-118583410 on Build GRCh38
rs911056945 Chr.11: 118583454-118583454 on Build GRCh38
rs782535805 Chr.11: 118583334-118583334 on Build GRCh38

View Full Product Details