Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118583403-118583938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1245300 c.478C>T p.R160C Chr.11: 118583839-118583839 on Build GRCh38
3359034 c.545G>T p.G182V Chr.11: 118583906-118583906 on Build GRCh38
6552719 c.517G>T p.A173S Chr.11: 118583878-118583878 on Build GRCh38
6454459 c.469G>A p.A157T Chr.11: 118583830-118583830 on Build GRCh38
4018372 c.553G>A p.G185R Chr.11: 118583914-118583914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945348317 Chr.11: 118583556-118583556 on Build GRCh38
rs1018525321 Chr.11: 118583869-118583869 on Build GRCh38
rs782518843 Chr.11: 118583860-118583860 on Build GRCh38
rs782225407 Chr.11: 118583877-118583877 on Build GRCh38
rs143812760 Chr.11: 118583633-118583633 on Build GRCh38
rs782307738 Chr.11: 118583887-118583887 on Build GRCh38
rs782491121 Chr.11: 118583828-118583828 on Build GRCh38
rs549411122 Chr.11: 118583689-118583689 on Build GRCh38
rs71469182 Chr.11: 118583607-118583608 on Build GRCh38
rs190453820 Chr.11: 118583465-118583465 on Build GRCh38
rs782238075 Chr.11: 118583841-118583841 on Build GRCh38
rs782037970 Chr.11: 118583784-118583784 on Build GRCh38
rs201640805 Chr.11: 118583770-118583770 on Build GRCh38
rs782785959 Chr.11: 118583792-118583792 on Build GRCh38
rs782039572 Chr.11: 118583767-118583767 on Build GRCh38
rs78574894 Chr.11: 118583455-118583455 on Build GRCh38
rs782455164 Chr.11: 118583617-118583617 on Build GRCh38
rs17842763 Chr.11: 118583462-118583462 on Build GRCh38
rs184379177 Chr.11: 118583433-118583433 on Build GRCh38
rs782646865 Chr.11: 118583870-118583870 on Build GRCh38
rs35189263 Chr.11: 118583597-118583598 on Build GRCh38
rs915257919 Chr.11: 118583468-118583468 on Build GRCh38
rs782383131 Chr.11: 118583769-118583770 on Build GRCh38
rs565156126 Chr.11: 118583586-118583586 on Build GRCh38
rs371110213 Chr.11: 118583793-118583793 on Build GRCh38
rs562540656 Chr.11: 118583734-118583734 on Build GRCh38
rs113120334 Chr.11: 118583563-118583563 on Build GRCh38
rs370697952 Chr.11: 118583672-118583672 on Build GRCh38
rs111590377 Chr.11: 118583770-118583770 on Build GRCh38
rs182216691 Chr.11: 118583683-118583683 on Build GRCh38
rs374528659 Chr.11: 118583833-118583833 on Build GRCh38
rs151181680 Chr.11: 118583913-118583913 on Build GRCh38
rs782527886 Chr.11: 118583839-118583839 on Build GRCh38
rs782702068 Chr.11: 118583832-118583832 on Build GRCh38
rs566377900 Chr.11: 118583583-118583583 on Build GRCh38
rs74319736 Chr.11: 118583456-118583456 on Build GRCh38
rs939445492 Chr.11: 118583570-118583570 on Build GRCh38
rs782373336 Chr.11: 118583914-118583914 on Build GRCh38
rs368207677 Chr.11: 118583781-118583781 on Build GRCh38
rs906657637 Chr.11: 118583569-118583569 on Build GRCh38
rs985722978 Chr.11: 118583444-118583444 on Build GRCh38
rs782690322 Chr.11: 118583840-118583840 on Build GRCh38
rs782341289 Chr.11: 118583758-118583758 on Build GRCh38
rs77830210 Chr.11: 118583457-118583457 on Build GRCh38
rs201154675 Chr.11: 118583766-118583766 on Build GRCh38
rs535975072 Chr.11: 118583445-118583445 on Build GRCh38
rs782122598 Chr.11: 118583790-118583790 on Build GRCh38
rs114802679 Chr.11: 118583430-118583430 on Build GRCh38
rs1055741827 Chr.11: 118583621-118583621 on Build GRCh38
rs782596288 Chr.11: 118583876-118583876 on Build GRCh38
rs1017169460 Chr.11: 118583703-118583703 on Build GRCh38
rs911056945 Chr.11: 118583454-118583454 on Build GRCh38
rs142087997 Chr.11: 118583899-118583899 on Build GRCh38

View Full Product Details