Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118583914-118584451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6580626 c.635T>C p.V212A Chr.11: 118583996-118583996 on Build GRCh38
923618 c.606G>T p.E202D Chr.11: 118583967-118583967 on Build GRCh38
2151213 c.588_589delAG p.A197fs*9 Chr.11: 118583949-118583950 on Build GRCh38
4018372 c.553G>A p.G185R Chr.11: 118583914-118583914 on Build GRCh38
1704641 c.587C>T p.T196I Chr.11: 118583948-118583948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782346904 Chr.11: 118584033-118584033 on Build GRCh38
rs782068782 Chr.11: 118583956-118583956 on Build GRCh38
rs782193561 Chr.11: 118584026-118584026 on Build GRCh38
rs782088012 Chr.11: 118583949-118583949 on Build GRCh38
rs371280810 Chr.11: 118584024-118584024 on Build GRCh38
rs536258319 Chr.11: 118584176-118584176 on Build GRCh38
rs185950602 Chr.11: 118583955-118583955 on Build GRCh38
rs782806877 Chr.11: 118583967-118583967 on Build GRCh38
rs568827977 Chr.11: 118584133-118584133 on Build GRCh38
rs550533494 Chr.11: 118584130-118584130 on Build GRCh38
rs782651258 Chr.11: 118584027-118584027 on Build GRCh38
rs140083980 Chr.11: 118583992-118583992 on Build GRCh38
rs978099657 Chr.11: 118583966-118583966 on Build GRCh38
rs571050423 Chr.11: 118584074-118584074 on Build GRCh38
rs538533150 Chr.11: 118584121-118584121 on Build GRCh38
rs782165621 Chr.11: 118584065-118584065 on Build GRCh38
rs967306925 Chr.11: 118584367-118584367 on Build GRCh38
rs781813411 Chr.11: 118583997-118583997 on Build GRCh38
rs572870221 Chr.11: 118584398-118584398 on Build GRCh38
rs782612480 Chr.11: 118584040-118584040 on Build GRCh38
rs994412032 Chr.11: 118584166-118584166 on Build GRCh38
rs752018696 Chr.11: 118584178-118584181 on Build GRCh38
rs895316758 Chr.11: 118584067-118584067 on Build GRCh38
rs782751278 Chr.11: 118583950-118583950 on Build GRCh38
rs782019966 Chr.11: 118584061-118584061 on Build GRCh38
rs782484136 Chr.11: 118583998-118583998 on Build GRCh38
rs1038528323 Chr.11: 118584147-118584147 on Build GRCh38
rs782207880 Chr.11: 118584340-118584340 on Build GRCh38
rs897438985 Chr.11: 118584151-118584151 on Build GRCh38
rs1029371754 Chr.11: 118584202-118584202 on Build GRCh38
rs913176572 Chr.11: 118583951-118583951 on Build GRCh38
rs374082927 Chr.11: 118584025-118584025 on Build GRCh38
rs886040860 Chr.11: 118583994-118583994 on Build GRCh38
rs782557537 Chr.11: 118584299-118584299 on Build GRCh38
rs890756596 Chr.11: 118584229-118584229 on Build GRCh38
rs782768022 Chr.11: 118583993-118583993 on Build GRCh38
rs554623853 Chr.11: 118584349-118584349 on Build GRCh38
rs990047053 Chr.11: 118583936-118583936 on Build GRCh38
rs367830871 Chr.11: 118584018-118584018 on Build GRCh38
rs1020067916 Chr.11: 118584362-118584362 on Build GRCh38
rs1018002953 Chr.11: 118584272-118584272 on Build GRCh38
rs989972015 Chr.11: 118584354-118584354 on Build GRCh38
rs957333115 Chr.11: 118584339-118584339 on Build GRCh38
rs368134894 Chr.11: 118584043-118584043 on Build GRCh38
rs1007057137 Chr.11: 118584256-118584256 on Build GRCh38
rs782815533 Chr.11: 118584053-118584054 on Build GRCh38
rs782489814 Chr.11: 118583974-118583974 on Build GRCh38

View Full Product Details