Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120420638-120421155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF12
Entrez Gene ID
23365
Gene Name
Rho guanine nucleotide exchange factor 12
Gene Aliases
LARG, PRO2792
UniGene
Hs.24598
Gene Chromosome Location
Chr.11: 120336276-120489936 on Build GRCh38
Transcripts
NM_001198665.1 NM_001301084.1 NM_015313.2 XM_006718805.3 XM_011542720.2 XM_017017420.1 XM_017017421.1 XM_017017422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786996 c.217G>A p.V73I Chr.11: 120420770-120420770 on Build GRCh38
6131584 c.202C>G p.L68V Chr.11: 120420755-120420755 on Build GRCh38
4433764 c.280G>A p.V94I Chr.11: 120420833-120420833 on Build GRCh38
5753614 c.212G>A p.R71H Chr.11: 120420765-120420765 on Build GRCh38
2107289 c.254C>G p.T85R Chr.11: 120420807-120420807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777826563 Chr.11: 120420901-120420901 on Build GRCh38
rs752994249 Chr.11: 120420883-120420883 on Build GRCh38
rs1018081992 Chr.11: 120421098-120421098 on Build GRCh38
rs779474166 Chr.11: 120420722-120420724 on Build GRCh38
rs762407733 Chr.11: 120420837-120420837 on Build GRCh38
rs985985416 Chr.11: 120421036-120421036 on Build GRCh38
rs945642060 Chr.11: 120420682-120420682 on Build GRCh38
rs34754222 Chr.11: 120420770-120420770 on Build GRCh38
rs757925250 Chr.11: 120420707-120420707 on Build GRCh38
rs913929145 Chr.11: 120421116-120421116 on Build GRCh38
rs576840146 Chr.11: 120421043-120421046 on Build GRCh38
rs756408416 Chr.11: 120420900-120420900 on Build GRCh38
rs746812292 Chr.11: 120420792-120420792 on Build GRCh38
rs774254420 Chr.11: 120420986-120420986 on Build GRCh38
rs1054529775 Chr.11: 120421032-120421032 on Build GRCh38
rs764771631 Chr.11: 120420833-120420833 on Build GRCh38
rs755304232 Chr.11: 120420878-120420878 on Build GRCh38
rs1012746624 Chr.11: 120420662-120420662 on Build GRCh38
rs7118147 Chr.11: 120420754-120420754 on Build GRCh38
rs907068745 Chr.11: 120421107-120421107 on Build GRCh38
rs961439124 Chr.11: 120420913-120420913 on Build GRCh38
rs374153562 Chr.11: 120420765-120420765 on Build GRCh38
rs1002658595 Chr.11: 120421119-120421119 on Build GRCh38
rs759356874 Chr.11: 120421099-120421099 on Build GRCh38
rs932915934 Chr.11: 120420935-120420935 on Build GRCh38
rs530323645 Chr.11: 120420994-120420994 on Build GRCh38
rs772648089 Chr.11: 120420808-120420808 on Build GRCh38
rs770846281 Chr.11: 120420977-120420977 on Build GRCh38
rs904060230 Chr.11: 120420817-120420817 on Build GRCh38
rs776284902 Chr.11: 120420814-120420814 on Build GRCh38
rs564188614 Chr.11: 120421006-120421006 on Build GRCh38
rs775530301 Chr.11: 120420789-120420789 on Build GRCh38
rs1013849327 Chr.11: 120420916-120420916 on Build GRCh38
rs370997007 Chr.11: 120420832-120420832 on Build GRCh38
rs774186113 Chr.11: 120421094-120421094 on Build GRCh38
rs756290111 Chr.11: 120420742-120420742 on Build GRCh38
rs752007785 Chr.11: 120420872-120420872 on Build GRCh38
rs766891281 Chr.11: 120420846-120420846 on Build GRCh38
rs150994739 Chr.11: 120421001-120421001 on Build GRCh38
rs976070651 Chr.11: 120421115-120421115 on Build GRCh38
rs199795842 Chr.11: 120420769-120420769 on Build GRCh38
rs767996623 Chr.11: 120420880-120420880 on Build GRCh38
rs867120391 Chr.11: 120420925-120420925 on Build GRCh38
rs902925565 Chr.11: 120421024-120421025 on Build GRCh38
rs993398386 Chr.11: 120420976-120420976 on Build GRCh38
rs934394925 Chr.11: 120421025-120421025 on Build GRCh38
rs985681497 Chr.11: 120420945-120420945 on Build GRCh38
rs951937800 Chr.11: 120421013-120421013 on Build GRCh38
rs762879701 Chr.11: 120420920-120420920 on Build GRCh38
rs897734017 Chr.11: 120420969-120420969 on Build GRCh38
rs1040189219 Chr.11: 120421020-120421020 on Build GRCh38
rs748339645 Chr.11: 120420737-120420737 on Build GRCh38
rs535630194 Chr.11: 120420926-120420926 on Build GRCh38

View Full Product Details