Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120421731-120422111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF12
Entrez Gene ID
23365
Gene Name
Rho guanine nucleotide exchange factor 12
Gene Aliases
LARG, PRO2792
UniGene
Hs.24598
Gene Chromosome Location
Chr.11: 120336276-120489936 on Build GRCh38
Transcripts
NM_001198665.1 NM_001301084.1 NM_015313.2 XM_006718805.3 XM_011542720.2 XM_017017420.1 XM_017017421.1 XM_017017422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147682 c.320G>A p.G107E Chr.11: 120421824-120421824 on Build GRCh38
266030 c.342C>T p.I114I Chr.11: 120421846-120421846 on Build GRCh38
923937 c.338G>A p.R113Q Chr.11: 120421842-120421842 on Build GRCh38
3868588 c.308C>T p.A103V Chr.11: 120421812-120421812 on Build GRCh38
6603959 c.337C>T p.R113* Chr.11: 120421841-120421841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754683208 Chr.11: 120421889-120421890 on Build GRCh38
rs115048809 Chr.11: 120422016-120422016 on Build GRCh38
rs760151289 Chr.11: 120421841-120421841 on Build GRCh38
rs532109985 Chr.11: 120422025-120422025 on Build GRCh38
rs569826050 Chr.11: 120422080-120422080 on Build GRCh38
rs369791110 Chr.11: 120421817-120421817 on Build GRCh38
rs768463478 Chr.11: 120421767-120421767 on Build GRCh38
rs908964885 Chr.11: 120422078-120422078 on Build GRCh38
rs532006 Chr.11: 120421861-120421861 on Build GRCh38
rs950520027 Chr.11: 120422034-120422034 on Build GRCh38
rs965341766 Chr.11: 120421829-120421829 on Build GRCh38
rs190745148 Chr.11: 120421965-120421965 on Build GRCh38
rs902580447 Chr.11: 120421920-120421920 on Build GRCh38
rs760909849 Chr.11: 120421860-120421860 on Build GRCh38
rs780686575 Chr.11: 120421890-120421890 on Build GRCh38
rs140386854 Chr.11: 120421787-120421787 on Build GRCh38
rs113223215 Chr.11: 120421833-120421833 on Build GRCh38
rs1012210043 Chr.11: 120421983-120421983 on Build GRCh38
rs542860737 Chr.11: 120421897-120421897 on Build GRCh38
rs1049189172 Chr.11: 120421796-120421796 on Build GRCh38
rs367644465 Chr.11: 120421761-120421761 on Build GRCh38
rs76693355 Chr.11: 120421768-120421768 on Build GRCh38
rs375592171 Chr.11: 120421760-120421760 on Build GRCh38
rs753146119 Chr.11: 120421858-120421858 on Build GRCh38
rs1037396162 Chr.11: 120422050-120422050 on Build GRCh38
rs1042256249 Chr.11: 120421911-120421911 on Build GRCh38
rs970967839 Chr.11: 120422015-120422015 on Build GRCh38
rs779550539 Chr.11: 120421794-120421794 on Build GRCh38
rs901762064 Chr.11: 120421881-120421881 on Build GRCh38
rs182245211 Chr.11: 120421989-120421989 on Build GRCh38
rs769515415 Chr.11: 120421775-120421775 on Build GRCh38
rs1000981156 Chr.11: 120421958-120421958 on Build GRCh38
rs750447298 Chr.11: 120421872-120421872 on Build GRCh38
rs757545938 Chr.11: 120421864-120421864 on Build GRCh38
rs545210599 Chr.11: 120421771-120421771 on Build GRCh38
rs376995719 Chr.11: 120421880-120421880 on Build GRCh38
rs747010341 Chr.11: 120421764-120421764 on Build GRCh38
rs769413993 Chr.11: 120421902-120421902 on Build GRCh38
rs551115117 Chr.11: 120421987-120421987 on Build GRCh38
rs748018809 Chr.11: 120421904-120421905 on Build GRCh38
rs189078263 Chr.11: 120421964-120421964 on Build GRCh38
rs984593840 Chr.11: 120422035-120422035 on Build GRCh38
rs751066733 Chr.11: 120421771-120421772 on Build GRCh38
rs933115194 Chr.11: 120421895-120421895 on Build GRCh38
rs767889107 Chr.11: 120421842-120421842 on Build GRCh38
rs929100615 Chr.11: 120421788-120421788 on Build GRCh38
rs754003664 Chr.11: 120421755-120421755 on Build GRCh38
rs754044668 Chr.11: 120421862-120421862 on Build GRCh38
rs1025159591 Chr.11: 120421984-120421984 on Build GRCh38
rs780947842 Chr.11: 120421890-120421890 on Build GRCh38

View Full Product Details