Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123058546-123059049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1 NR_001454.2

Gene Symbol
SNORD14D
Entrez Gene ID
85390
Gene Name
small nucleolar RNA, C/D box 14D
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 123058909-123058995 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1 NR_001454.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297758 c.1327T>C p.Y443H Chr.11: 123058827-123058827 on Build GRCh38
3808623 c.1383C>T p.L461L Chr.11: 123058771-123058771 on Build GRCh38
686780 c.1404C>T p.P468P Chr.11: 123058750-123058750 on Build GRCh38
466481 c.1457T>C p.L486P Chr.11: 123058697-123058697 on Build GRCh38
1297757 c.1445C>T p.A482V Chr.11: 123058709-123058709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879653768 Chr.11: 123058894-123058894 on Build GRCh38
rs373976955 Chr.11: 123058605-123058605 on Build GRCh38
rs541493124 Chr.11: 123058953-123058957 on Build GRCh38
rs372007616 Chr.11: 123059020-123059020 on Build GRCh38
rs377257990 Chr.11: 123058786-123058786 on Build GRCh38
rs375810107 Chr.11: 123058861-123058861 on Build GRCh38
rs996347750 Chr.11: 123058898-123058898 on Build GRCh38
rs766069802 Chr.11: 123058643-123058643 on Build GRCh38
rs150260166 Chr.11: 123059002-123059002 on Build GRCh38
rs767610060 Chr.11: 123059024-123059024 on Build GRCh38
rs916360248 Chr.11: 123058994-123058994 on Build GRCh38
rs764034057 Chr.11: 123058756-123058756 on Build GRCh38
rs11551598 Chr.11: 123058779-123058779 on Build GRCh38
rs1461496 Chr.11: 123058916-123058916 on Build GRCh38
rs375700185 Chr.11: 123058862-123058862 on Build GRCh38
rs765230787 Chr.11: 123058619-123058619 on Build GRCh38
rs780280602 Chr.11: 123058847-123058847 on Build GRCh38
rs77171215 Chr.11: 123058837-123058837 on Build GRCh38
rs566577895 Chr.11: 123058874-123058874 on Build GRCh38
rs774627510 Chr.11: 123059000-123059000 on Build GRCh38
rs778359512 Chr.11: 123058844-123058844 on Build GRCh38
rs774366672 Chr.11: 123058872-123058872 on Build GRCh38
rs528685013 Chr.11: 123058613-123058613 on Build GRCh38
rs546409549 Chr.11: 123058620-123058620 on Build GRCh38
rs771376397 Chr.11: 123058696-123058696 on Build GRCh38
rs571307052 Chr.11: 123058690-123058690 on Build GRCh38
rs754334609 Chr.11: 123058869-123058904 on Build GRCh38
rs779303299 Chr.11: 123058839-123058839 on Build GRCh38
rs1029611491 Chr.11: 123058904-123058904 on Build GRCh38
rs147529697 Chr.11: 123058669-123058669 on Build GRCh38
rs553205643 Chr.11: 123058893-123058893 on Build GRCh38
rs543249357 Chr.11: 123058586-123058586 on Build GRCh38
rs759731819 Chr.11: 123058610-123058610 on Build GRCh38
rs977745166 Chr.11: 123058951-123058951 on Build GRCh38
rs76450893 Chr.11: 123058990-123058990 on Build GRCh38
rs138544510 Chr.11: 123058675-123058675 on Build GRCh38
rs561503650 Chr.11: 123058591-123058591 on Build GRCh38
rs531181092 Chr.11: 123058584-123058584 on Build GRCh38
rs770171764 Chr.11: 123058925-123058925 on Build GRCh38
rs771430988 Chr.11: 123058969-123058969 on Build GRCh38
rs752441020 Chr.11: 123058882-123058882 on Build GRCh38
rs747179523 Chr.11: 123058855-123058855 on Build GRCh38
rs763006619 Chr.11: 123058869-123058869 on Build GRCh38
rs771159184 Chr.11: 123058592-123058592 on Build GRCh38
rs751387334 Chr.11: 123058789-123058789 on Build GRCh38
rs752359934 Chr.11: 123058833-123058833 on Build GRCh38
rs925015154 Chr.11: 123058975-123058975 on Build GRCh38
rs753270496 Chr.11: 123059015-123059015 on Build GRCh38
rs759347844 Chr.11: 123058792-123058792 on Build GRCh38
rs773513797 Chr.11: 123058930-123058930 on Build GRCh38
rs758237764 Chr.11: 123058633-123058633 on Build GRCh38
rs767284605 Chr.11: 123058819-123058819 on Build GRCh38
rs766521413 Chr.11: 123058984-123058984 on Build GRCh38
rs879252511 Chr.11: 123058630-123058630 on Build GRCh38
rs189446113 Chr.11: 123058942-123058942 on Build GRCh38
rs767621093 Chr.11: 123058611-123058611 on Build GRCh38
rs370076742 Chr.11: 123059010-123059010 on Build GRCh38
rs986477868 Chr.11: 123058717-123058717 on Build GRCh38
rs773253340 Chr.11: 123058757-123058757 on Build GRCh38
rs114271013 Chr.11: 123058889-123058889 on Build GRCh38
rs746613946 Chr.11: 123058582-123058582 on Build GRCh38
rs772264576 Chr.11: 123058714-123058714 on Build GRCh38
rs1042085144 Chr.11: 123058758-123058758 on Build GRCh38
rs771005222 Chr.11: 123058871-123058871 on Build GRCh38
rs377525683 Chr.11: 123058618-123058618 on Build GRCh38
rs545075365 Chr.11: 123058926-123058926 on Build GRCh38
rs912266434 Chr.11: 123058726-123058726 on Build GRCh38
rs140962304 Chr.11: 123058845-123058845 on Build GRCh38
rs199504970 Chr.11: 123058705-123058705 on Build GRCh38
rs34958717 Chr.11: 123058630-123058631 on Build GRCh38
rs111774985 Chr.11: 123058648-123058648 on Build GRCh38
rs1381245 Chr.11: 123058946-123058946 on Build GRCh38
rs371118607 Chr.11: 123058836-123058836 on Build GRCh38
rs1025424865 Chr.11: 123058587-123058587 on Build GRCh38
rs778373831 Chr.11: 123058687-123058687 on Build GRCh38
rs773535665 Chr.11: 123058589-123058589 on Build GRCh38
rs945045071 Chr.11: 123058744-123058744 on Build GRCh38
rs183933120 Chr.11: 123058838-123058838 on Build GRCh38
rs1064585 Chr.11: 123058699-123058699 on Build GRCh38
rs115611816 Chr.11: 123058936-123058936 on Build GRCh38
rs760810506 Chr.11: 123058729-123058729 on Build GRCh38
rs777064602 Chr.11: 123058899-123058899 on Build GRCh38
rs746441724 Chr.11: 123058770-123058770 on Build GRCh38
rs774576037 Chr.11: 123058608-123058608 on Build GRCh38
rs75793565 Chr.11: 123059019-123059019 on Build GRCh38
rs775629110 Chr.11: 123058720-123058720 on Build GRCh38
rs925044940 Chr.11: 123058764-123058764 on Build GRCh38
rs749707173 Chr.11: 123058694-123058694 on Build GRCh38
rs553450668 Chr.11: 123058804-123058804 on Build GRCh38
rs954984664 Chr.11: 123058911-123058911 on Build GRCh38
rs763172186 Chr.11: 123058931-123058931 on Build GRCh38
rs780908558 Chr.11: 123058655-123058655 on Build GRCh38
rs767370308 Chr.11: 123058876-123058876 on Build GRCh38
rs763725484 Chr.11: 123059011-123059011 on Build GRCh38

View Full Product Details