Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31281817-31282353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM60A
Entrez Gene ID
58516
Gene Name
family with sequence similarity 60 member A
Gene Aliases
C12orf14, L4, TERA
UniGene
Hs.505154
Gene Chromosome Location
Chr.12: 31280584-31326225 on Build GRCh38
Transcripts
NM_001135811.1 NM_001135812.1 NM_021238.2 XM_011520803.2 XM_017019758.1 XM_017019759.1 XM_017019760.1 XM_017019761.1 XM_017019762.1

SNP Detail

SNP ID Chromosome Location
rs59295865 Chr.12: 31282127-31282130 on Build GRCh38
rs116478715 Chr.12: 31282262-31282262 on Build GRCh38
rs763242562 Chr.12: 31282288-31282288 on Build GRCh38
rs532816843 Chr.12: 31282039-31282039 on Build GRCh38
rs560323170 Chr.12: 31282160-31282160 on Build GRCh38
rs549205070 Chr.12: 31282280-31282280 on Build GRCh38
rs199836281 Chr.12: 31282132-31282135 on Build GRCh38
rs564119818 Chr.12: 31281849-31281849 on Build GRCh38
rs916001732 Chr.12: 31282025-31282025 on Build GRCh38
rs960017485 Chr.12: 31281888-31281888 on Build GRCh38
rs4015571 Chr.12: 31282109-31282109 on Build GRCh38
rs4015569 Chr.12: 31281992-31281992 on Build GRCh38
rs547820920 Chr.12: 31282135-31282135 on Build GRCh38
rs543816150 Chr.12: 31281843-31281843 on Build GRCh38
rs773562471 Chr.12: 31282204-31282204 on Build GRCh38
rs187800117 Chr.12: 31281858-31281858 on Build GRCh38
rs1007823719 Chr.12: 31281845-31281845 on Build GRCh38
rs4015568 Chr.12: 31281961-31281961 on Build GRCh38
rs540351409 Chr.12: 31281942-31281942 on Build GRCh38
rs1007274277 Chr.12: 31282308-31282308 on Build GRCh38
rs992816030 Chr.12: 31282013-31282013 on Build GRCh38
rs3953534 Chr.12: 31281879-31281879 on Build GRCh38
rs4015570 Chr.12: 31282065-31282066 on Build GRCh38
rs968628284 Chr.12: 31281851-31281851 on Build GRCh38
rs888865394 Chr.12: 31282291-31282292 on Build GRCh38
rs397850727 Chr.12: 31282215-31282215 on Build GRCh38
rs35128222 Chr.12: 31282204-31282204 on Build GRCh38
rs948795594 Chr.12: 31282061-31282061 on Build GRCh38
rs769944589 Chr.12: 31282101-31282101 on Build GRCh38
rs1031715113 Chr.12: 31281861-31281861 on Build GRCh38
rs940317662 Chr.12: 31282186-31282186 on Build GRCh38
rs796747474 Chr.12: 31282131-31282134 on Build GRCh38
rs897523286 Chr.12: 31282222-31282222 on Build GRCh38

View Full Product Details