Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31671055-31671570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_006719054.3 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

SNP Detail

SNP ID Chromosome Location
rs957898982 Chr.12: 31671195-31671195 on Build GRCh38
rs576755234 Chr.12: 31671366-31671366 on Build GRCh38
rs887566317 Chr.12: 31671170-31671170 on Build GRCh38
rs927269180 Chr.12: 31671444-31671444 on Build GRCh38
rs530020665 Chr.12: 31671119-31671119 on Build GRCh38
rs988149122 Chr.12: 31671368-31671368 on Build GRCh38
rs958124229 Chr.12: 31671202-31671202 on Build GRCh38
rs116212412 Chr.12: 31671242-31671242 on Build GRCh38
rs1032645254 Chr.12: 31671187-31671187 on Build GRCh38
rs1021071564 Chr.12: 31671256-31671256 on Build GRCh38
rs34907876 Chr.12: 31671447-31671448 on Build GRCh38
rs187409271 Chr.12: 31671176-31671176 on Build GRCh38
rs181857429 Chr.12: 31671359-31671359 on Build GRCh38
rs773867145 Chr.12: 31671514-31671514 on Build GRCh38
rs1032300018 Chr.12: 31671335-31671335 on Build GRCh38
rs1020557273 Chr.12: 31671377-31671377 on Build GRCh38
rs901980130 Chr.12: 31671222-31671222 on Build GRCh38
rs34535151 Chr.12: 31671368-31671369 on Build GRCh38
rs566089868 Chr.12: 31671332-31671332 on Build GRCh38
rs554877785 Chr.12: 31671379-31671379 on Build GRCh38
rs1029269606 Chr.12: 31671275-31671275 on Build GRCh38
rs886857397 Chr.12: 31671083-31671083 on Build GRCh38
rs147797962 Chr.12: 31671446-31671447 on Build GRCh38
rs543718985 Chr.12: 31671470-31671470 on Build GRCh38
rs555802425 Chr.12: 31671342-31671342 on Build GRCh38
rs149726060 Chr.12: 31671132-31671132 on Build GRCh38
rs574660572 Chr.12: 31671431-31671431 on Build GRCh38
rs926642371 Chr.12: 31671330-31671331 on Build GRCh38
rs929532538 Chr.12: 31671408-31671408 on Build GRCh38
rs557263071 Chr.12: 31671539-31671539 on Build GRCh38
rs982100503 Chr.12: 31671324-31671324 on Build GRCh38
rs180687572 Chr.12: 31671103-31671103 on Build GRCh38
rs12698 Chr.12: 31671510-31671510 on Build GRCh38
rs1036947096 Chr.12: 31671193-31671193 on Build GRCh38
rs35682650 Chr.12: 31671336-31671337 on Build GRCh38
rs1007932381 Chr.12: 31671084-31671084 on Build GRCh38
rs937021518 Chr.12: 31671104-31671104 on Build GRCh38
rs956789555 Chr.12: 31671339-31671339 on Build GRCh38
rs943751653 Chr.12: 31671469-31671469 on Build GRCh38
rs971280869 Chr.12: 31671380-31671380 on Build GRCh38
rs3534 Chr.12: 31671319-31671319 on Build GRCh38

View Full Product Details