Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31697227-31697763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_006719054.3 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4041409 c.582G>T p.K194N Chr.12: 31697370-31697370 on Build GRCh38
3359705 c.591+5T>C p.? Chr.12: 31697356-31697356 on Build GRCh38
2205648 c.559C>T p.L187F Chr.12: 31697393-31697393 on Build GRCh38
4571927 c.587T>C p.L196S Chr.12: 31697365-31697365 on Build GRCh38
3398667 c.578C>T p.A193V Chr.12: 31697374-31697374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74744848 Chr.12: 31697349-31697349 on Build GRCh38
rs1047719275 Chr.12: 31697642-31697642 on Build GRCh38
rs776324728 Chr.12: 31697712-31697712 on Build GRCh38
rs751774634 Chr.12: 31697319-31697319 on Build GRCh38
rs974380260 Chr.12: 31697327-31697327 on Build GRCh38
rs751678036 Chr.12: 31697349-31697349 on Build GRCh38
rs545282301 Chr.12: 31697576-31697576 on Build GRCh38
rs12822865 Chr.12: 31697646-31697646 on Build GRCh38
rs771352182 Chr.12: 31697394-31697394 on Build GRCh38
rs372596524 Chr.12: 31697461-31697461 on Build GRCh38
rs377293296 Chr.12: 31697426-31697426 on Build GRCh38
rs749266255 Chr.12: 31697334-31697334 on Build GRCh38
rs1028930997 Chr.12: 31697331-31697331 on Build GRCh38
rs776820957 Chr.12: 31697409-31697409 on Build GRCh38
rs1025777802 Chr.12: 31697277-31697277 on Build GRCh38
rs937823140 Chr.12: 31697528-31697528 on Build GRCh38
rs369865604 Chr.12: 31697435-31697435 on Build GRCh38
rs776165334 Chr.12: 31697546-31697546 on Build GRCh38
rs374889400 Chr.12: 31697302-31697302 on Build GRCh38
rs12822978 Chr.12: 31697597-31697597 on Build GRCh38
rs887266905 Chr.12: 31697679-31697679 on Build GRCh38
rs372572233 Chr.12: 31697438-31697438 on Build GRCh38
rs763466711 Chr.12: 31697702-31697703 on Build GRCh38
rs769427717 Chr.12: 31697676-31697676 on Build GRCh38
rs74085158 Chr.12: 31697682-31697682 on Build GRCh38
rs771707923 Chr.12: 31697374-31697374 on Build GRCh38
rs36119592 Chr.12: 31697260-31697261 on Build GRCh38
rs375411486 Chr.12: 31697697-31697697 on Build GRCh38
rs775980430 Chr.12: 31697719-31697719 on Build GRCh38
rs773005290 Chr.12: 31697375-31697375 on Build GRCh38
rs770192827 Chr.12: 31697715-31697715 on Build GRCh38
rs770531585 Chr.12: 31697708-31697708 on Build GRCh38
rs531780557 Chr.12: 31697262-31697262 on Build GRCh38
rs763085975 Chr.12: 31697422-31697422 on Build GRCh38
rs375845901 Chr.12: 31697510-31697510 on Build GRCh38
rs941236292 Chr.12: 31697585-31697585 on Build GRCh38
rs957548979 Chr.12: 31697724-31697724 on Build GRCh38
rs755159363 Chr.12: 31697698-31697700 on Build GRCh38
rs748034645 Chr.12: 31697462-31697462 on Build GRCh38
rs746278300 Chr.12: 31697713-31697713 on Build GRCh38
rs1030252945 Chr.12: 31697287-31697287 on Build GRCh38
rs927787198 Chr.12: 31697487-31697487 on Build GRCh38
rs12824497 Chr.12: 31697661-31697661 on Build GRCh38
rs369791800 Chr.12: 31697586-31697587 on Build GRCh38
rs755906346 Chr.12: 31697323-31697323 on Build GRCh38
rs146226968 Chr.12: 31697467-31697467 on Build GRCh38
rs571724139 Chr.12: 31697339-31697339 on Build GRCh38
rs1028627297 Chr.12: 31697718-31697718 on Build GRCh38
rs957269210 Chr.12: 31697337-31697337 on Build GRCh38
rs1007136260 Chr.12: 31697340-31697340 on Build GRCh38
rs747706961 Chr.12: 31697373-31697373 on Build GRCh38
rs753292243 Chr.12: 31697705-31697706 on Build GRCh38
rs564124621 Chr.12: 31697271-31697271 on Build GRCh38
rs371548307 Chr.12: 31697415-31697415 on Build GRCh38
rs564162932 Chr.12: 31697727-31697727 on Build GRCh38
rs370433531 Chr.12: 31697427-31697427 on Build GRCh38
rs199712749 Chr.12: 31697412-31697412 on Build GRCh38
rs772724232 Chr.12: 31697730-31697730 on Build GRCh38
rs12824502 Chr.12: 31697663-31697663 on Build GRCh38
rs759914221 Chr.12: 31697410-31697410 on Build GRCh38
rs2388926 Chr.12: 31697286-31697286 on Build GRCh38
rs1055683382 Chr.12: 31697485-31697488 on Build GRCh38
rs928668004 Chr.12: 31697459-31697459 on Build GRCh38
rs779656149 Chr.12: 31697324-31697324 on Build GRCh38
rs777374003 Chr.12: 31697694-31697694 on Build GRCh38
rs774374018 Chr.12: 31697737-31697737 on Build GRCh38
rs779927408 Chr.12: 31697439-31697439 on Build GRCh38
rs746693846 Chr.12: 31697696-31697696 on Build GRCh38
rs753612065 Chr.12: 31697448-31697448 on Build GRCh38
rs909783786 Chr.12: 31697583-31697583 on Build GRCh38
rs760350187 Chr.12: 31697377-31697377 on Build GRCh38
rs988753645 Chr.12: 31697381-31697381 on Build GRCh38

View Full Product Details