Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31701783-31702281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_006719054.3 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259411 c.257A>C p.K86T Chr.12: 31701922-31701922 on Build GRCh38
7178834 c.254G>C p.R85T Chr.12: 31701925-31701925 on Build GRCh38
5589627 c.206G>A p.R69Q Chr.12: 31701973-31701973 on Build GRCh38
3772605 c.196C>A p.L66I Chr.12: 31701983-31701983 on Build GRCh38
5551335 c.213C>T p.C71C Chr.12: 31701966-31701966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920505604 Chr.12: 31702097-31702097 on Build GRCh38
rs980045371 Chr.12: 31702090-31702090 on Build GRCh38
rs778123903 Chr.12: 31701996-31701996 on Build GRCh38
rs779736452 Chr.12: 31701938-31701938 on Build GRCh38
rs371094062 Chr.12: 31701881-31701881 on Build GRCh38
rs771131665 Chr.12: 31702008-31702008 on Build GRCh38
rs772678124 Chr.12: 31701974-31701974 on Build GRCh38
rs192752736 Chr.12: 31702087-31702087 on Build GRCh38
rs576811405 Chr.12: 31702220-31702220 on Build GRCh38
rs751464143 Chr.12: 31701883-31701883 on Build GRCh38
rs1013640795 Chr.12: 31702028-31702028 on Build GRCh38
rs942438911 Chr.12: 31701945-31701945 on Build GRCh38
rs779728234 Chr.12: 31702025-31702025 on Build GRCh38
rs564606840 Chr.12: 31702165-31702165 on Build GRCh38
rs903499293 Chr.12: 31701962-31701962 on Build GRCh38
rs370799416 Chr.12: 31701966-31701966 on Build GRCh38
rs753482154 Chr.12: 31701944-31701944 on Build GRCh38
rs761759978 Chr.12: 31701884-31701884 on Build GRCh38
rs776981108 Chr.12: 31701875-31701875 on Build GRCh38
rs745447593 Chr.12: 31701907-31701907 on Build GRCh38
rs969872125 Chr.12: 31702066-31702066 on Build GRCh38
rs774154404 Chr.12: 31702037-31702037 on Build GRCh38
rs200630208 Chr.12: 31701995-31701996 on Build GRCh38
rs1025221017 Chr.12: 31702126-31702126 on Build GRCh38
rs759776349 Chr.12: 31702036-31702036 on Build GRCh38
rs758053673 Chr.12: 31701932-31701933 on Build GRCh38
rs952031980 Chr.12: 31702123-31702123 on Build GRCh38
rs200109528 Chr.12: 31702010-31702010 on Build GRCh38
rs1052316686 Chr.12: 31702012-31702012 on Build GRCh38
rs182849179 Chr.12: 31701845-31701845 on Build GRCh38
rs972295551 Chr.12: 31701811-31701811 on Build GRCh38
rs539441663 Chr.12: 31701840-31701840 on Build GRCh38
rs761480681 Chr.12: 31702038-31702038 on Build GRCh38
rs905315302 Chr.12: 31702170-31702170 on Build GRCh38
rs576031973 Chr.12: 31702049-31702049 on Build GRCh38
rs762909823 Chr.12: 31701878-31701878 on Build GRCh38
rs767538647 Chr.12: 31702040-31702040 on Build GRCh38
rs755338140 Chr.12: 31701965-31701965 on Build GRCh38
rs768991865 Chr.12: 31701815-31701815 on Build GRCh38
rs1002678719 Chr.12: 31702191-31702191 on Build GRCh38
rs187141583 Chr.12: 31701920-31701920 on Build GRCh38
rs771328077 Chr.12: 31701853-31701853 on Build GRCh38
rs375183160 Chr.12: 31701894-31701894 on Build GRCh38
rs761500478 Chr.12: 31702041-31702042 on Build GRCh38
rs536067868 Chr.12: 31701997-31701997 on Build GRCh38
rs749989333 Chr.12: 31701904-31701904 on Build GRCh38
rs907500080 Chr.12: 31702226-31702226 on Build GRCh38
rs377565397 Chr.12: 31701847-31701847 on Build GRCh38
rs1024146974 Chr.12: 31702063-31702063 on Build GRCh38
rs375140557 Chr.12: 31701973-31701973 on Build GRCh38
rs184753033 Chr.12: 31702243-31702243 on Build GRCh38
rs983154145 Chr.12: 31702154-31702154 on Build GRCh38
rs766628214 Chr.12: 31702228-31702228 on Build GRCh38
rs200816879 Chr.12: 31701818-31701818 on Build GRCh38
rs1039410968 Chr.12: 31701949-31701949 on Build GRCh38

View Full Product Details