Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31708849-31709395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981002 c.131T>C p.M44T Chr.12: 31709333-31709333 on Build GRCh38
5913320 c.100C>T p.P34S Chr.12: 31709364-31709364 on Build GRCh38
431026 c.118C>T p.L40L Chr.12: 31709346-31709346 on Build GRCh38
375188 c.159A>C p.S53S Chr.12: 31709305-31709305 on Build GRCh38
3811878 c.75T>A p.Y25* Chr.12: 31709389-31709389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014522844 Chr.12: 31709036-31709036 on Build GRCh38
rs184987545 Chr.12: 31708968-31708968 on Build GRCh38
rs986581049 Chr.12: 31709164-31709164 on Build GRCh38
rs745993535 Chr.12: 31709265-31709265 on Build GRCh38
rs753754186 Chr.12: 31708976-31708976 on Build GRCh38
rs949481035 Chr.12: 31709005-31709006 on Build GRCh38
rs765806612 Chr.12: 31708929-31708929 on Build GRCh38
rs772953536 Chr.12: 31709330-31709330 on Build GRCh38
rs983154675 Chr.12: 31708948-31708948 on Build GRCh38
rs935437586 Chr.12: 31708999-31708999 on Build GRCh38
rs953800205 Chr.12: 31709293-31709293 on Build GRCh38
rs778412336 Chr.12: 31709004-31709004 on Build GRCh38
rs776540263 Chr.12: 31709346-31709346 on Build GRCh38
rs752287347 Chr.12: 31709194-31709194 on Build GRCh38
rs1027238131 Chr.12: 31708906-31708906 on Build GRCh38
rs913745434 Chr.12: 31709347-31709347 on Build GRCh38
rs1001615427 Chr.12: 31708902-31708902 on Build GRCh38
rs925240140 Chr.12: 31709193-31709195 on Build GRCh38
rs181908240 Chr.12: 31709050-31709050 on Build GRCh38
rs371807376 Chr.12: 31709276-31709276 on Build GRCh38
rs757151824 Chr.12: 31708995-31708995 on Build GRCh38
rs771756139 Chr.12: 31709117-31709117 on Build GRCh38
rs759028691 Chr.12: 31709187-31709187 on Build GRCh38
rs770907515 Chr.12: 31709344-31709344 on Build GRCh38
rs374840560 Chr.12: 31709332-31709332 on Build GRCh38
rs371429545 Chr.12: 31709174-31709174 on Build GRCh38
rs752137212 Chr.12: 31709214-31709214 on Build GRCh38
rs758676835 Chr.12: 31708975-31708975 on Build GRCh38
rs553487461 Chr.12: 31708982-31708982 on Build GRCh38
rs1004610541 Chr.12: 31709165-31709165 on Build GRCh38
rs760545510 Chr.12: 31709152-31709152 on Build GRCh38
rs376908206 Chr.12: 31709158-31709158 on Build GRCh38
rs1045534088 Chr.12: 31709044-31709044 on Build GRCh38
rs773135331 Chr.12: 31709135-31709135 on Build GRCh38
rs117817433 Chr.12: 31709190-31709190 on Build GRCh38
rs750589644 Chr.12: 31709106-31709106 on Build GRCh38
rs539576705 Chr.12: 31708972-31708972 on Build GRCh38
rs757797225 Chr.12: 31709244-31709244 on Build GRCh38
rs749400512 Chr.12: 31709319-31709319 on Build GRCh38
rs771911909 Chr.12: 31709328-31709328 on Build GRCh38
rs376308150 Chr.12: 31709054-31709054 on Build GRCh38
rs139260347 Chr.12: 31709098-31709098 on Build GRCh38
rs143329862 Chr.12: 31708949-31708949 on Build GRCh38
rs1043751204 Chr.12: 31708989-31708989 on Build GRCh38
rs1014642777 Chr.12: 31709170-31709170 on Build GRCh38
rs764308579 Chr.12: 31709212-31709212 on Build GRCh38
rs769407607 Chr.12: 31709198-31709199 on Build GRCh38
rs1052509538 Chr.12: 31709003-31709003 on Build GRCh38
rs372876258 Chr.12: 31709156-31709156 on Build GRCh38
rs150816425 Chr.12: 31709064-31709064 on Build GRCh38
rs770094749 Chr.12: 31709102-31709102 on Build GRCh38
rs781754450 Chr.12: 31709264-31709264 on Build GRCh38
rs912217390 Chr.12: 31708942-31708942 on Build GRCh38
rs575804101 Chr.12: 31709107-31709107 on Build GRCh38
rs773053288 Chr.12: 31709256-31709277 on Build GRCh38
rs748129190 Chr.12: 31709174-31709175 on Build GRCh38
rs535092181 Chr.12: 31709173-31709174 on Build GRCh38
rs776162388 Chr.12: 31709157-31709157 on Build GRCh38
rs79750888 Chr.12: 31709173-31709173 on Build GRCh38
rs757428528 Chr.12: 31709076-31709076 on Build GRCh38
rs994111318 Chr.12: 31709122-31709122 on Build GRCh38
rs568013065 Chr.12: 31708940-31708940 on Build GRCh38
rs758794930 Chr.12: 31709204-31709204 on Build GRCh38
rs189587319 Chr.12: 31708969-31708969 on Build GRCh38
rs951837043 Chr.12: 31708907-31708907 on Build GRCh38
rs781370456 Chr.12: 31709078-31709078 on Build GRCh38
rs777650355 Chr.12: 31709056-31709056 on Build GRCh38
rs756134843 Chr.12: 31709275-31709275 on Build GRCh38

View Full Product Details