Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 45331154-45331678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
UniGene
Hs.505339
Gene Chromosome Location
Chr.12: 45215987-45440404 on Build GRCh38
Transcripts
NM_001025356.2 NM_001142678.1 NM_001142679.1 NM_001204803.1 XM_005268706.4 XM_005268707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404640 c.115C>A p.P39T Chr.12: 45331313-45331313 on Build GRCh38
4826780 c.203G>A p.R68K Chr.12: 45331347-45331347 on Build GRCh38
4827037 c.190G>A p.D64N Chr.12: 45331334-45331334 on Build GRCh38
136058 c.200G>A p.R67Q Chr.12: 45331344-45331344 on Build GRCh38
5620274 c.252T>G p.N84K Chr.12: 45331396-45331396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745938678 Chr.12: 45331300-45331300 on Build GRCh38
rs994885541 Chr.12: 45331340-45331340 on Build GRCh38
rs762235354 Chr.12: 45331467-45331467 on Build GRCh38
rs767881630 Chr.12: 45331472-45331472 on Build GRCh38
rs750571136 Chr.12: 45331395-45331395 on Build GRCh38
rs778960751 Chr.12: 45331445-45331448 on Build GRCh38
rs746802648 Chr.12: 45331424-45331424 on Build GRCh38
rs1019176391 Chr.12: 45331385-45331385 on Build GRCh38
rs533103339 Chr.12: 45331303-45331303 on Build GRCh38
rs1012093275 Chr.12: 45331367-45331367 on Build GRCh38
rs772699401 Chr.12: 45331244-45331244 on Build GRCh38
rs140147341 Chr.12: 45331420-45331420 on Build GRCh38
rs771964303 Chr.12: 45331524-45331526 on Build GRCh38
rs928898360 Chr.12: 45331576-45331576 on Build GRCh38
rs567258249 Chr.12: 45331532-45331532 on Build GRCh38
rs770623217 Chr.12: 45331427-45331427 on Build GRCh38
rs772928472 Chr.12: 45331335-45331335 on Build GRCh38
rs994936280 Chr.12: 45331615-45331615 on Build GRCh38
rs963221918 Chr.12: 45331500-45331500 on Build GRCh38
rs371446949 Chr.12: 45331288-45331288 on Build GRCh38
rs759028735 Chr.12: 45331356-45331356 on Build GRCh38
rs144030718 Chr.12: 45331559-45331559 on Build GRCh38
rs1042867090 Chr.12: 45331196-45331196 on Build GRCh38
rs774638142 Chr.12: 45331456-45331456 on Build GRCh38
rs938863086 Chr.12: 45331290-45331290 on Build GRCh38
rs777084769 Chr.12: 45331419-45331419 on Build GRCh38
rs751811799 Chr.12: 45331378-45331378 on Build GRCh38
rs373168816 Chr.12: 45331408-45331408 on Build GRCh38
rs200699542 Chr.12: 45331245-45331245 on Build GRCh38
rs755797821 Chr.12: 45331344-45331344 on Build GRCh38
rs748813968 Chr.12: 45331428-45331430 on Build GRCh38
rs779014168 Chr.12: 45331404-45331404 on Build GRCh38
rs546662602 Chr.12: 45331641-45331641 on Build GRCh38
rs138413111 Chr.12: 45331375-45331375 on Build GRCh38
rs770411031 Chr.12: 45331436-45331437 on Build GRCh38
rs776239220 Chr.12: 45331428-45331428 on Build GRCh38
rs770562151 Chr.12: 45331343-45331343 on Build GRCh38
rs559214409 Chr.12: 45331214-45331214 on Build GRCh38
rs925802449 Chr.12: 45331464-45331464 on Build GRCh38
rs745846535 Chr.12: 45331450-45331453 on Build GRCh38
rs753106949 Chr.12: 45331365-45331365 on Build GRCh38
rs559278675 Chr.12: 45331587-45331587 on Build GRCh38
rs200785570 Chr.12: 45331305-45331305 on Build GRCh38
rs879130765 Chr.12: 45331444-45331444 on Build GRCh38
rs938894435 Chr.12: 45331595-45331595 on Build GRCh38
rs967103327 Chr.12: 45331444-45331444 on Build GRCh38
rs868450490 Chr.12: 45331298-45331298 on Build GRCh38
rs549016070 Chr.12: 45331527-45331527 on Build GRCh38
rs778556960 Chr.12: 45331392-45331392 on Build GRCh38
rs1055606390 Chr.12: 45331284-45331284 on Build GRCh38
rs917159592 Chr.12: 45331544-45331544 on Build GRCh38
rs945902720 Chr.12: 45331186-45331186 on Build GRCh38
rs907449419 Chr.12: 45331273-45331276 on Build GRCh38
rs373816443 Chr.12: 45331202-45331202 on Build GRCh38
rs145969134 Chr.12: 45331372-45331372 on Build GRCh38
rs897786305 Chr.12: 45331610-45331610 on Build GRCh38
rs1055660079 Chr.12: 45331529-45331529 on Build GRCh38
rs781542223 Chr.12: 45331388-45331388 on Build GRCh38
rs776079108 Chr.12: 45331436-45331436 on Build GRCh38
rs369239470 Chr.12: 45331383-45331383 on Build GRCh38
rs1036881670 Chr.12: 45331329-45331329 on Build GRCh38
rs765775901 Chr.12: 45331361-45331361 on Build GRCh38
rs1036369357 Chr.12: 45331586-45331586 on Build GRCh38
rs769249049 Chr.12: 45331432-45331432 on Build GRCh38
rs777491349 Chr.12: 45331422-45331422 on Build GRCh38
rs765814948 Chr.12: 45331254-45331254 on Build GRCh38
rs765723046 Chr.12: 45331266-45331266 on Build GRCh38
rs757374418 Chr.12: 45331293-45331293 on Build GRCh38
rs760099596 Chr.12: 45331262-45331262 on Build GRCh38
rs760174767 Chr.12: 45331341-45331341 on Build GRCh38
rs748004511 Chr.12: 45331312-45331312 on Build GRCh38
rs751685422 Chr.12: 45331292-45331292 on Build GRCh38
rs112025010 Chr.12: 45331558-45331558 on Build GRCh38
rs142459456 Chr.12: 45331369-45331369 on Build GRCh38
rs753113905 Chr.12: 45331269-45331269 on Build GRCh38
rs756221549 Chr.12: 45331398-45331398 on Build GRCh38
rs759081955 Chr.12: 45331430-45331430 on Build GRCh38
rs771713040 Chr.12: 45331333-45331333 on Build GRCh38
rs781354389 Chr.12: 45331297-45331297 on Build GRCh38
rs754343416 Chr.12: 45331256-45331256 on Build GRCh38

View Full Product Details