Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75275955-75276485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
UniGene
Hs.733835
Gene Chromosome Location
Chr.12: 75275979-75390941 on Build GRCh38
Transcripts
NM_001286547.1 NM_032606.3 XM_005269192.1 XM_005269194.3 XM_006719648.2 XM_006719649.2 XM_011538878.1 XM_011538886.2 XM_011538887.2 XM_011538888.1 XM_011538889.2 XM_011538892.1 XM_017020034.1 XM_017020035.1 XM_017020036.1 XM_017020037.1 XM_017020038.1 XM_017020039.1 XM_017020040.1 XM_017020041.1 XM_017020042.1 XM_017020043.1 XM_017020044.1 XM_017020045.1 XM_017020046.1 XM_017020047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3688442 c.1670G>T p.G557V Chr.12: 75276247-75276247 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1019331236 Chr.12: 75275984-75275984 on Build GRCh38
rs772507522 Chr.12: 75276203-75276203 on Build GRCh38
rs570427212 Chr.12: 75276007-75276007 on Build GRCh38
rs982372700 Chr.12: 75276045-75276045 on Build GRCh38
rs752103388 Chr.12: 75276228-75276228 on Build GRCh38
rs916493429 Chr.12: 75276013-75276013 on Build GRCh38
rs367693630 Chr.12: 75276235-75276235 on Build GRCh38
rs973408618 Chr.12: 75276023-75276023 on Build GRCh38
rs542782966 Chr.12: 75276288-75276288 on Build GRCh38
rs754599424 Chr.12: 75276208-75276208 on Build GRCh38
rs148211827 Chr.12: 75276059-75276059 on Build GRCh38
rs752821051 Chr.12: 75276291-75276291 on Build GRCh38
rs889384812 Chr.12: 75276164-75276164 on Build GRCh38
rs868054489 Chr.12: 75276459-75276459 on Build GRCh38
rs190158948 Chr.12: 75276266-75276266 on Build GRCh38
rs893998114 Chr.12: 75276101-75276101 on Build GRCh38
rs938186933 Chr.12: 75276261-75276261 on Build GRCh38
rs545886290 Chr.12: 75276377-75276377 on Build GRCh38
rs539012394 Chr.12: 75276139-75276139 on Build GRCh38
rs999189821 Chr.12: 75276121-75276121 on Build GRCh38
rs768673802 Chr.12: 75276175-75276175 on Build GRCh38
rs986637093 Chr.12: 75275992-75275992 on Build GRCh38
rs865951186 Chr.12: 75276227-75276227 on Build GRCh38
rs182103961 Chr.12: 75276419-75276419 on Build GRCh38
rs902833140 Chr.12: 75276117-75276117 on Build GRCh38
rs915411618 Chr.12: 75276362-75276362 on Build GRCh38
rs755490966 Chr.12: 75276072-75276072 on Build GRCh38
rs375947473 Chr.12: 75276233-75276233 on Build GRCh38
rs769849033 Chr.12: 75276078-75276085 on Build GRCh38
rs758468771 Chr.12: 75276414-75276414 on Build GRCh38
rs969349699 Chr.12: 75276418-75276418 on Build GRCh38
rs952558059 Chr.12: 75276294-75276294 on Build GRCh38
rs554617536 Chr.12: 75276228-75276228 on Build GRCh38
rs368613161 Chr.12: 75276127-75276127 on Build GRCh38
rs991022220 Chr.12: 75276342-75276342 on Build GRCh38
rs143072850 Chr.12: 75276048-75276048 on Build GRCh38
rs751405137 Chr.12: 75276271-75276271 on Build GRCh38
rs927678306 Chr.12: 75276461-75276465 on Build GRCh38
rs768983049 Chr.12: 75276229-75276229 on Build GRCh38
rs1042784926 Chr.12: 75276100-75276100 on Build GRCh38
rs371460773 Chr.12: 75276214-75276214 on Build GRCh38
rs539388213 Chr.12: 75276263-75276263 on Build GRCh38
rs919256859 Chr.12: 75276027-75276027 on Build GRCh38
rs749397841 Chr.12: 75276067-75276067 on Build GRCh38
rs767074309 Chr.12: 75276207-75276208 on Build GRCh38
rs1007312901 Chr.12: 75276170-75276170 on Build GRCh38
rs929570819 Chr.12: 75276037-75276037 on Build GRCh38
rs558902738 Chr.12: 75276154-75276154 on Build GRCh38
rs765705987 Chr.12: 75276132-75276132 on Build GRCh38
rs746412154 Chr.12: 75276202-75276202 on Build GRCh38
rs984076708 Chr.12: 75276012-75276012 on Build GRCh38
rs963354622 Chr.12: 75276008-75276008 on Build GRCh38
rs1017382979 Chr.12: 75276188-75276188 on Build GRCh38
rs987483768 Chr.12: 75275979-75275979 on Build GRCh38
rs567131508 Chr.12: 75276068-75276068 on Build GRCh38
rs567391660 Chr.12: 75276038-75276038 on Build GRCh38
rs575197268 Chr.12: 75276201-75276201 on Build GRCh38
rs1009526455 Chr.12: 75276166-75276166 on Build GRCh38
rs950718579 Chr.12: 75276219-75276219 on Build GRCh38
rs144236844 Chr.12: 75275986-75275987 on Build GRCh38

View Full Product Details