Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 77021446-77021984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F7
Entrez Gene ID
144455
Gene Name
E2F transcription factor 7
Gene Aliases
-
UniGene
Hs.416375
Gene Chromosome Location
Chr.12: 77021246-77065580 on Build GRCh38
Transcripts
NM_203394.2 XM_011537966.2 XM_011537969.2

SNP Detail

SNP ID Chromosome Location
rs528224805 Chr.12: 77021541-77021541 on Build GRCh38
rs769579666 Chr.12: 77021704-77021704 on Build GRCh38
rs955703241 Chr.12: 77021770-77021770 on Build GRCh38
rs758545767 Chr.12: 77021496-77021496 on Build GRCh38
rs565188177 Chr.12: 77021470-77021474 on Build GRCh38
rs1048429476 Chr.12: 77021521-77021521 on Build GRCh38
rs190831591 Chr.12: 77021790-77021790 on Build GRCh38
rs1004146718 Chr.12: 77021545-77021545 on Build GRCh38
rs920430942 Chr.12: 77021890-77021890 on Build GRCh38
rs891311540 Chr.12: 77021688-77021690 on Build GRCh38
rs1035960572 Chr.12: 77021595-77021595 on Build GRCh38
rs186429868 Chr.12: 77021596-77021596 on Build GRCh38
rs917663112 Chr.12: 77021473-77021473 on Build GRCh38
rs868707080 Chr.12: 77021808-77021808 on Build GRCh38
rs778057199 Chr.12: 77021591-77021591 on Build GRCh38
rs772672738 Chr.12: 77021659-77021659 on Build GRCh38
rs1012064791 Chr.12: 77021876-77021876 on Build GRCh38
rs374077853 Chr.12: 77021645-77021645 on Build GRCh38
rs923579758 Chr.12: 77021499-77021499 on Build GRCh38
rs569095686 Chr.12: 77021655-77021655 on Build GRCh38
rs936344016 Chr.12: 77021500-77021500 on Build GRCh38
rs528965796 Chr.12: 77021479-77021479 on Build GRCh38
rs557345040 Chr.12: 77021898-77021898 on Build GRCh38
rs943276716 Chr.12: 77021843-77021843 on Build GRCh38
rs895085504 Chr.12: 77021812-77021812 on Build GRCh38
rs1025206339 Chr.12: 77021889-77021890 on Build GRCh38
rs180888550 Chr.12: 77021497-77021497 on Build GRCh38
rs1050369783 Chr.12: 77021674-77021674 on Build GRCh38
rs563298972 Chr.12: 77021474-77021474 on Build GRCh38
rs1026897390 Chr.12: 77021789-77021789 on Build GRCh38
rs987119939 Chr.12: 77021773-77021773 on Build GRCh38
rs748980363 Chr.12: 77021944-77021944 on Build GRCh38
rs149345223 Chr.12: 77021929-77021929 on Build GRCh38
rs1038901282 Chr.12: 77021878-77021878 on Build GRCh38
rs887111948 Chr.12: 77021536-77021536 on Build GRCh38
rs914841728 Chr.12: 77021570-77021570 on Build GRCh38
rs970907647 Chr.12: 77021484-77021484 on Build GRCh38
rs551168053 Chr.12: 77021579-77021579 on Build GRCh38
rs995548992 Chr.12: 77021771-77021771 on Build GRCh38
rs978516340 Chr.12: 77021703-77021703 on Build GRCh38
rs1037124078 Chr.12: 77021764-77021764 on Build GRCh38
rs559495839 Chr.12: 77021520-77021520 on Build GRCh38
rs1023554391 Chr.12: 77021652-77021652 on Build GRCh38

View Full Product Details