Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 77022492-77023004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F7
Entrez Gene ID
144455
Gene Name
E2F transcription factor 7
Gene Aliases
-
UniGene
Hs.416375
Gene Chromosome Location
Chr.12: 77021246-77065580 on Build GRCh38
Transcripts
NM_203394.2 XM_011537966.2 XM_011537969.2

SNP Detail

SNP ID Chromosome Location
rs900236601 Chr.12: 77022911-77022911 on Build GRCh38
rs993325768 Chr.12: 77022728-77022728 on Build GRCh38
rs544774904 Chr.12: 77022912-77022912 on Build GRCh38
rs972999860 Chr.12: 77022816-77022816 on Build GRCh38
rs936228846 Chr.12: 77022858-77022858 on Build GRCh38
rs996268440 Chr.12: 77022941-77022941 on Build GRCh38
rs1018656825 Chr.12: 77022576-77022576 on Build GRCh38
rs965644773 Chr.12: 77022809-77022809 on Build GRCh38
rs964749628 Chr.12: 77022588-77022588 on Build GRCh38
rs1009043649 Chr.12: 77022863-77022863 on Build GRCh38
rs1027558149 Chr.12: 77022603-77022603 on Build GRCh38
rs556985601 Chr.12: 77022622-77022622 on Build GRCh38
rs537774681 Chr.12: 77022542-77022542 on Build GRCh38
rs1040540833 Chr.12: 77022909-77022909 on Build GRCh38
rs34974675 Chr.12: 77022970-77022971 on Build GRCh38
rs573601873 Chr.12: 77022635-77022635 on Build GRCh38
rs948082924 Chr.12: 77022528-77022528 on Build GRCh38
rs983288717 Chr.12: 77022679-77022679 on Build GRCh38
rs1053328488 Chr.12: 77022861-77022861 on Build GRCh38
rs772642088 Chr.12: 77022559-77022559 on Build GRCh38
rs904812896 Chr.12: 77022845-77022845 on Build GRCh38
rs906489126 Chr.12: 77022548-77022548 on Build GRCh38
rs948744804 Chr.12: 77022768-77022768 on Build GRCh38
rs141787382 Chr.12: 77022862-77022862 on Build GRCh38
rs559141667 Chr.12: 77022810-77022810 on Build GRCh38
rs1044397409 Chr.12: 77022815-77022815 on Build GRCh38
rs917318862 Chr.12: 77022767-77022767 on Build GRCh38
rs939166001 Chr.12: 77022720-77022720 on Build GRCh38
rs754380581 Chr.12: 77022977-77022977 on Build GRCh38
rs202108048 Chr.12: 77022944-77022945 on Build GRCh38
rs974405477 Chr.12: 77022597-77022597 on Build GRCh38
rs375256444 Chr.12: 77022855-77022855 on Build GRCh38
rs533893231 Chr.12: 77022661-77022661 on Build GRCh38
rs893651930 Chr.12: 77022642-77022642 on Build GRCh38
rs1052310779 Chr.12: 77022965-77022965 on Build GRCh38
rs927986344 Chr.12: 77022910-77022910 on Build GRCh38
rs571223578 Chr.12: 77022541-77022541 on Build GRCh38
rs948035994 Chr.12: 77022895-77022895 on Build GRCh38
rs1019867078 Chr.12: 77022790-77022790 on Build GRCh38
rs952918080 Chr.12: 77022856-77022856 on Build GRCh38
rs766742868 Chr.12: 77022716-77022716 on Build GRCh38
rs148058630 Chr.12: 77022712-77022712 on Build GRCh38
rs1027275931 Chr.12: 77022948-77022948 on Build GRCh38

View Full Product Details