Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119998628-119999132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL1
Entrez Gene ID
92558
Gene Name
BICD family like cargo adaptor 1
Gene Aliases
BICDR-1, BICDR1, CCDC64, H_267D11.1
UniGene
Hs.369763
Gene Chromosome Location
Chr.12: 119989806-120094495 on Build GRCh38
Transcripts
NM_207311.2 XM_005253991.4 XM_006719694.3 XM_006719696.3 XM_006719697.3 XM_011538999.2 XM_011539001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883594 c.610G>T p.E204* Chr.12: 119998701-119998701 on Build GRCh38
6007923 c.538G>A p.E180K Chr.12: 119998629-119998629 on Build GRCh38
268850 c.589C>T p.R197W Chr.12: 119998680-119998680 on Build GRCh38
4146711 c.639C>A p.L213L Chr.12: 119998730-119998730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758726230 Chr.12: 119998742-119998742 on Build GRCh38
rs34221846 Chr.12: 119998732-119998732 on Build GRCh38
rs1015203529 Chr.12: 119999060-119999060 on Build GRCh38
rs376594740 Chr.12: 119998816-119998816 on Build GRCh38
rs10659159 Chr.12: 119999062-119999063 on Build GRCh38
rs756276898 Chr.12: 119999063-119999063 on Build GRCh38
rs960064035 Chr.12: 119999057-119999057 on Build GRCh38
rs376923599 Chr.12: 119998761-119998761 on Build GRCh38
rs962260603 Chr.12: 119998809-119998809 on Build GRCh38
rs775368911 Chr.12: 119998749-119998756 on Build GRCh38
rs530503047 Chr.12: 119998827-119998827 on Build GRCh38
rs540886788 Chr.12: 119999082-119999083 on Build GRCh38
rs753498748 Chr.12: 119998718-119998718 on Build GRCh38
rs979345656 Chr.12: 119998853-119998853 on Build GRCh38
rs903552221 Chr.12: 119998668-119998668 on Build GRCh38
rs925084179 Chr.12: 119999103-119999103 on Build GRCh38
rs1042312610 Chr.12: 119998926-119998926 on Build GRCh38
rs7958402 Chr.12: 119998895-119998895 on Build GRCh38
rs771746776 Chr.12: 119998768-119998768 on Build GRCh38
rs895240360 Chr.12: 119998963-119998963 on Build GRCh38
rs1016419757 Chr.12: 119998968-119998968 on Build GRCh38
rs773932042 Chr.12: 119998747-119998747 on Build GRCh38
rs992006708 Chr.12: 119999060-119999061 on Build GRCh38
rs1052059264 Chr.12: 119998872-119998872 on Build GRCh38
rs543448351 Chr.12: 119998797-119998797 on Build GRCh38
rs1005120421 Chr.12: 119999055-119999055 on Build GRCh38
rs748129703 Chr.12: 119998700-119998700 on Build GRCh38
rs775164109 Chr.12: 119998777-119998777 on Build GRCh38
rs928613102 Chr.12: 119998815-119998815 on Build GRCh38
rs548526347 Chr.12: 119998898-119998898 on Build GRCh38
rs777500780 Chr.12: 119998705-119998705 on Build GRCh38
rs561217277 Chr.12: 119998730-119998730 on Build GRCh38
rs755119778 Chr.12: 119998683-119998683 on Build GRCh38
rs901502343 Chr.12: 119998882-119998882 on Build GRCh38
rs1052020571 Chr.12: 119998726-119998726 on Build GRCh38
rs144705742 Chr.12: 119998934-119998934 on Build GRCh38
rs902502585 Chr.12: 119998927-119998927 on Build GRCh38
rs574768591 Chr.12: 119998795-119998795 on Build GRCh38
rs933105546 Chr.12: 119998893-119998893 on Build GRCh38
rs953572381 Chr.12: 119998800-119998800 on Build GRCh38
rs77211221 Chr.12: 119998854-119998854 on Build GRCh38
rs766524053 Chr.12: 119998669-119998669 on Build GRCh38
rs979252321 Chr.12: 119999097-119999097 on Build GRCh38
rs550642099 Chr.12: 119998969-119998969 on Build GRCh38
rs1012789228 Chr.12: 119998980-119998980 on Build GRCh38
rs968744046 Chr.12: 119999084-119999084 on Build GRCh38
rs10658480 Chr.12: 119999083-119999083 on Build GRCh38
rs973742962 Chr.12: 119999083-119999083 on Build GRCh38
rs1016513297 Chr.12: 119998807-119998807 on Build GRCh38
rs943963703 Chr.12: 119998911-119998911 on Build GRCh38
rs1006504305 Chr.12: 119998801-119998801 on Build GRCh38
rs758580208 Chr.12: 119998657-119998657 on Build GRCh38
rs781079649 Chr.12: 119998699-119998699 on Build GRCh38
rs568393035 Chr.12: 119998902-119998902 on Build GRCh38
rs901594151 Chr.12: 119998774-119998774 on Build GRCh38
rs552596368 Chr.12: 119998826-119998826 on Build GRCh38
rs765649959 Chr.12: 119998652-119998652 on Build GRCh38
rs371966029 Chr.12: 119998650-119998650 on Build GRCh38
rs750785448 Chr.12: 119998653-119998653 on Build GRCh38
rs201157328 Chr.12: 119998681-119998681 on Build GRCh38
rs570381036 Chr.12: 119998993-119998993 on Build GRCh38
rs936691178 Chr.12: 119998930-119998930 on Build GRCh38
rs1025614387 Chr.12: 119998796-119998796 on Build GRCh38
rs539167549 Chr.12: 119999025-119999025 on Build GRCh38
rs919699972 Chr.12: 119998863-119998863 on Build GRCh38
rs906727393 Chr.12: 119999034-119999034 on Build GRCh38
rs1036132533 Chr.12: 119999043-119999044 on Build GRCh38
rs201124505 Chr.12: 119998806-119998806 on Build GRCh38

View Full Product Details