Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120461405-120461953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATC
Entrez Gene ID
283459
Gene Name
glutamyl-tRNA amidotransferase subunit C
Gene Aliases
15E1.2
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120446438-120463753 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

Gene Symbol
SRSF9
Entrez Gene ID
8683
Gene Name
serine and arginine rich splicing factor 9
Gene Aliases
SFRS9, SRp30c
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120461668-120469755 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

SNP Detail

SNP ID Chromosome Location
rs1046396799 Chr.12: 120461707-120461707 on Build GRCh38
rs114832191 Chr.12: 120461718-120461718 on Build GRCh38
rs746425066 Chr.12: 120461490-120461490 on Build GRCh38
rs532871032 Chr.12: 120461621-120461621 on Build GRCh38
rs897377310 Chr.12: 120461514-120461514 on Build GRCh38
rs899313320 Chr.12: 120461764-120461764 on Build GRCh38
rs939573151 Chr.12: 120461800-120461800 on Build GRCh38
rs1048163653 Chr.12: 120461790-120461790 on Build GRCh38
rs529010748 Chr.12: 120461699-120461699 on Build GRCh38
rs1011848660 Chr.12: 120461884-120461885 on Build GRCh38
rs544420072 Chr.12: 120461477-120461477 on Build GRCh38
rs142287809 Chr.12: 120461809-120461809 on Build GRCh38
rs772181930 Chr.12: 120461497-120461497 on Build GRCh38
rs941457515 Chr.12: 120461775-120461775 on Build GRCh38
rs959028031 Chr.12: 120461571-120461571 on Build GRCh38
rs957891280 Chr.12: 120461567-120461567 on Build GRCh38
rs921880364 Chr.12: 120461716-120461716 on Build GRCh38
rs780067333 Chr.12: 120461569-120461569 on Build GRCh38
rs746547284 Chr.12: 120461763-120461764 on Build GRCh38
rs1036507635 Chr.12: 120461733-120461733 on Build GRCh38
rs909947307 Chr.12: 120461742-120461742 on Build GRCh38
rs976402933 Chr.12: 120461712-120461712 on Build GRCh38
rs1004212552 Chr.12: 120461532-120461532 on Build GRCh38
rs747113501 Chr.12: 120461632-120461632 on Build GRCh38
rs1037170307 Chr.12: 120461787-120461787 on Build GRCh38
rs895429060 Chr.12: 120461862-120461862 on Build GRCh38
rs1031264784 Chr.12: 120461669-120461669 on Build GRCh38
rs928172161 Chr.12: 120461708-120461708 on Build GRCh38
rs977927741 Chr.12: 120461665-120461666 on Build GRCh38
rs1021935175 Chr.12: 120461900-120461900 on Build GRCh38
rs916699186 Chr.12: 120461642-120461642 on Build GRCh38
rs528770912 Chr.12: 120461914-120461914 on Build GRCh38
rs779395567 Chr.12: 120461500-120461501 on Build GRCh38
rs995031663 Chr.12: 120461788-120461788 on Build GRCh38
rs967794647 Chr.12: 120461650-120461650 on Build GRCh38
rs575378839 Chr.12: 120461465-120461465 on Build GRCh38
rs34136998 Chr.12: 120461899-120461900 on Build GRCh38
rs985483306 Chr.12: 120461729-120461729 on Build GRCh38
rs775160638 Chr.12: 120461711-120461711 on Build GRCh38
rs770142743 Chr.12: 120461500-120461500 on Build GRCh38
rs989511402 Chr.12: 120461625-120461625 on Build GRCh38
rs200337069 Chr.12: 120461899-120461899 on Build GRCh38
rs563807114 Chr.12: 120461597-120461597 on Build GRCh38

View Full Product Details