Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120463562-120464061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATC
Entrez Gene ID
283459
Gene Name
glutamyl-tRNA amidotransferase subunit C
Gene Aliases
15E1.2
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120446438-120463753 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

Gene Symbol
SRSF9
Entrez Gene ID
8683
Gene Name
serine and arginine rich splicing factor 9
Gene Aliases
SFRS9, SRp30c
UniGene
Hs.706889
Gene Chromosome Location
Chr.12: 120461668-120469755 on Build GRCh38
Transcripts
NM_176818.2 NR_033684.1 NM_003769.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5439249 c.520G>A p.E174K Chr.12: 120463952-120463952 on Build GRCh38
6318677 c.471C>A p.D157E Chr.12: 120464001-120464001 on Build GRCh38
5374386 c.443G>A p.G148E Chr.12: 120464029-120464029 on Build GRCh38
7186710 c.422A>C p.D141A Chr.12: 120464050-120464050 on Build GRCh38
5962259 c.421G>A p.D141N Chr.12: 120464051-120464051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182039749 Chr.12: 120463624-120463624 on Build GRCh38
rs1057378870 Chr.12: 120463641-120463641 on Build GRCh38
rs1021761345 Chr.12: 120463818-120463818 on Build GRCh38
rs763744277 Chr.12: 120463921-120463921 on Build GRCh38
rs917084654 Chr.12: 120463858-120463858 on Build GRCh38
rs1048935391 Chr.12: 120463701-120463701 on Build GRCh38
rs980908628 Chr.12: 120463843-120463843 on Build GRCh38
rs892520598 Chr.12: 120464034-120464034 on Build GRCh38
rs183513157 Chr.12: 120463913-120463913 on Build GRCh38
rs905201415 Chr.12: 120463668-120463668 on Build GRCh38
rs753813982 Chr.12: 120463935-120463935 on Build GRCh38
rs144718122 Chr.12: 120463750-120463750 on Build GRCh38
rs769288084 Chr.12: 120463986-120463986 on Build GRCh38
rs141506018 Chr.12: 120463937-120463937 on Build GRCh38
rs767909905 Chr.12: 120464033-120464033 on Build GRCh38
rs752335285 Chr.12: 120463787-120463787 on Build GRCh38
rs767105261 Chr.12: 120463740-120463740 on Build GRCh38
rs892049714 Chr.12: 120463774-120463774 on Build GRCh38
rs187091912 Chr.12: 120463693-120463693 on Build GRCh38
rs919216501 Chr.12: 120463687-120463687 on Build GRCh38
rs929223890 Chr.12: 120463979-120463979 on Build GRCh38
rs973174581 Chr.12: 120463597-120463597 on Build GRCh38
rs748433319 Chr.12: 120463924-120463926 on Build GRCh38
rs963068555 Chr.12: 120463779-120463779 on Build GRCh38
rs760390002 Chr.12: 120463795-120463795 on Build GRCh38
rs559786056 Chr.12: 120463648-120463648 on Build GRCh38
rs750732977 Chr.12: 120463932-120463932 on Build GRCh38
rs555710364 Chr.12: 120463749-120463752 on Build GRCh38
rs1045007258 Chr.12: 120463657-120463657 on Build GRCh38
rs926298191 Chr.12: 120463631-120463631 on Build GRCh38
rs1001310494 Chr.12: 120463670-120463670 on Build GRCh38
rs1044284951 Chr.12: 120463906-120463906 on Build GRCh38
rs745799567 Chr.12: 120463944-120463944 on Build GRCh38
rs569271980 Chr.12: 120463908-120463908 on Build GRCh38
rs147887689 Chr.12: 120463891-120463891 on Build GRCh38
rs191671910 Chr.12: 120463751-120463751 on Build GRCh38
rs774709001 Chr.12: 120463912-120463912 on Build GRCh38
rs1009712233 Chr.12: 120463723-120463723 on Build GRCh38
rs776216215 Chr.12: 120463971-120463971 on Build GRCh38
rs374865867 Chr.12: 120463960-120463960 on Build GRCh38
rs997561691 Chr.12: 120463765-120463765 on Build GRCh38
rs968209535 Chr.12: 120463585-120463585 on Build GRCh38
rs758330684 Chr.12: 120463956-120463956 on Build GRCh38
rs760013988 Chr.12: 120464025-120464025 on Build GRCh38
rs759271026 Chr.12: 120463866-120463866 on Build GRCh38
rs764266978 Chr.12: 120463917-120463917 on Build GRCh38
rs148751549 Chr.12: 120463656-120463656 on Build GRCh38
rs774378465 Chr.12: 120463643-120463643 on Build GRCh38
rs758939719 Chr.12: 120463729-120463729 on Build GRCh38
rs145299974 Chr.12: 120463962-120463962 on Build GRCh38
rs766811801 Chr.12: 120463900-120463900 on Build GRCh38
rs967796872 Chr.12: 120463832-120463832 on Build GRCh38
rs918990109 Chr.12: 120463805-120463805 on Build GRCh38
rs1031881120 Chr.12: 120463769-120463769 on Build GRCh38
rs919034286 Chr.12: 120463608-120463608 on Build GRCh38
rs774700766 Chr.12: 120463905-120463905 on Build GRCh38
rs370660252 Chr.12: 120463730-120463730 on Build GRCh38
rs780382015 Chr.12: 120464022-120464022 on Build GRCh38
rs756259723 Chr.12: 120463915-120463915 on Build GRCh38
rs752594381 Chr.12: 120463995-120463995 on Build GRCh38
rs529394906 Chr.12: 120463834-120463834 on Build GRCh38
rs566814907 Chr.12: 120463754-120463754 on Build GRCh38
rs950533684 Chr.12: 120463841-120463841 on Build GRCh38
rs941720175 Chr.12: 120463587-120463587 on Build GRCh38
rs980433393 Chr.12: 120463616-120463616 on Build GRCh38
rs767854700 Chr.12: 120463910-120463910 on Build GRCh38
rs909022001 Chr.12: 120464011-120464011 on Build GRCh38
rs201631930 Chr.12: 120463961-120463961 on Build GRCh38
rs973506050 Chr.12: 120463791-120463791 on Build GRCh38

View Full Product Details