Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121420161-121420656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
UniGene
Hs.524800
Gene Chromosome Location
Chr.12: 121402289-121582268 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Gene Symbol
RNF34
Entrez Gene ID
80196
Gene Name
ring finger protein 34
Gene Aliases
CARP-1, CARP1, RFI, RIF, RIFF, hRFI
UniGene
Hs.292804
Gene Chromosome Location
Chr.12: 121400083-121424353 on Build GRCh38
Transcripts
XM_011538867.2 NM_001256858.1 NM_025126.3 NM_194271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1224003 c.701A>C p.D234A Chr.12: 121420306-121420306 on Build GRCh38
6767923 c.794T>C p.V265A Chr.12: 121420641-121420641 on Build GRCh38
4039454 c.684C>T p.D228D Chr.12: 121420289-121420289 on Build GRCh38
7313414 c.655G>T p.A219S Chr.12: 121420263-121420263 on Build GRCh38
1359466 c.678C>T p.D226D Chr.12: 121420286-121420286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782340848 Chr.12: 121420287-121420287 on Build GRCh38
rs544697639 Chr.12: 121420322-121420322 on Build GRCh38
rs3751134 Chr.12: 121420409-121420409 on Build GRCh38
rs552464462 Chr.12: 121420466-121420466 on Build GRCh38
rs782676250 Chr.12: 121420333-121420333 on Build GRCh38
rs1036122546 Chr.12: 121420200-121420200 on Build GRCh38
rs782205285 Chr.12: 121420475-121420475 on Build GRCh38
rs3751135 Chr.12: 121420405-121420405 on Build GRCh38
rs782302427 Chr.12: 121420226-121420226 on Build GRCh38
rs782718487 Chr.12: 121420292-121420292 on Build GRCh38
rs1057207380 Chr.12: 121420438-121420438 on Build GRCh38
rs782753057 Chr.12: 121420298-121420299 on Build GRCh38
rs994143133 Chr.12: 121420451-121420451 on Build GRCh38
rs782304541 Chr.12: 121420536-121420536 on Build GRCh38
rs782212928 Chr.12: 121420244-121420244 on Build GRCh38
rs913462391 Chr.12: 121420367-121420367 on Build GRCh38
rs1001656623 Chr.12: 121420419-121420419 on Build GRCh38
rs782583004 Chr.12: 121420265-121420265 on Build GRCh38
rs781904957 Chr.12: 121420623-121420624 on Build GRCh38
rs370594485 Chr.12: 121420374-121420374 on Build GRCh38
rs1053784242 Chr.12: 121420346-121420346 on Build GRCh38
rs377017432 Chr.12: 121420289-121420289 on Build GRCh38
rs964455463 Chr.12: 121420302-121420302 on Build GRCh38
rs781822735 Chr.12: 121420625-121420625 on Build GRCh38
rs781893046 Chr.12: 121420295-121420295 on Build GRCh38
rs782167758 Chr.12: 121420298-121420298 on Build GRCh38
rs782782316 Chr.12: 121420582-121420582 on Build GRCh38
rs190921881 Chr.12: 121420283-121420283 on Build GRCh38
rs964507743 Chr.12: 121420601-121420601 on Build GRCh38
rs368005152 Chr.12: 121420323-121420323 on Build GRCh38
rs895812360 Chr.12: 121420444-121420444 on Build GRCh38
rs1026668052 Chr.12: 121420458-121420458 on Build GRCh38
rs781943892 Chr.12: 121420540-121420540 on Build GRCh38
rs782279180 Chr.12: 121420270-121420270 on Build GRCh38
rs782055171 Chr.12: 121420580-121420580 on Build GRCh38
rs61751317 Chr.12: 121420313-121420313 on Build GRCh38
rs782688135 Chr.12: 121420565-121420566 on Build GRCh38
rs532006940 Chr.12: 121420518-121420518 on Build GRCh38
rs574924801 Chr.12: 121420340-121420340 on Build GRCh38
rs573048643 Chr.12: 121420274-121420274 on Build GRCh38
rs202132357 Chr.12: 121420286-121420286 on Build GRCh38
rs61739898 Chr.12: 121420310-121420310 on Build GRCh38
rs781802564 Chr.12: 121420326-121420326 on Build GRCh38
rs558958221 Chr.12: 121420290-121420290 on Build GRCh38
rs138199363 Chr.12: 121420247-121420247 on Build GRCh38
rs190400029 Chr.12: 121420567-121420567 on Build GRCh38
rs139288912 Chr.12: 121420602-121420602 on Build GRCh38
rs147092139 Chr.12: 121420227-121420227 on Build GRCh38
rs782675008 Chr.12: 121420215-121420215 on Build GRCh38
rs782642706 Chr.12: 121420609-121420609 on Build GRCh38
rs111466028 Chr.12: 121420236-121420236 on Build GRCh38
rs536489502 Chr.12: 121420211-121420211 on Build GRCh38
rs782771959 Chr.12: 121420311-121420311 on Build GRCh38
rs782643365 Chr.12: 121420233-121420233 on Build GRCh38
rs782737126 Chr.12: 121420565-121420565 on Build GRCh38
rs782314946 Chr.12: 121420214-121420214 on Build GRCh38
rs1043275053 Chr.12: 121420408-121420408 on Build GRCh38
rs527945351 Chr.12: 121420412-121420412 on Build GRCh38
rs1026196601 Chr.12: 121420254-121420254 on Build GRCh38
rs782224901 Chr.12: 121420386-121420386 on Build GRCh38
rs782141810 Chr.12: 121420557-121420557 on Build GRCh38
rs782545231 Chr.12: 121420480-121420480 on Build GRCh38
rs782575372 Chr.12: 121420417-121420417 on Build GRCh38
rs564509403 Chr.12: 121420490-121420490 on Build GRCh38
rs371660693 Chr.12: 121420555-121420555 on Build GRCh38
rs992517032 Chr.12: 121420230-121420230 on Build GRCh38
rs782600448 Chr.12: 121420354-121420354 on Build GRCh38

View Full Product Details