Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121429019-121429529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
UniGene
Hs.524800
Gene Chromosome Location
Chr.12: 121402289-121582268 on Build GRCh38
Transcripts
NM_001005366.1 NM_032590.4 XM_005253955.3 XM_005253956.3 XM_005253961.4 XM_011538867.2 XM_011538868.2 XM_011538869.2 XM_011538875.2

SNP Detail

SNP ID Chromosome Location
rs1049853167 Chr.12: 121429065-121429065 on Build GRCh38
rs902477566 Chr.12: 121429125-121429125 on Build GRCh38
rs782603684 Chr.12: 121429400-121429400 on Build GRCh38
rs1012785463 Chr.12: 121429477-121429477 on Build GRCh38
rs144247530 Chr.12: 121429145-121429145 on Build GRCh38
rs1033317585 Chr.12: 121429151-121429151 on Build GRCh38
rs151292455 Chr.12: 121429043-121429043 on Build GRCh38
rs555859658 Chr.12: 121429395-121429395 on Build GRCh38
rs1012730311 Chr.12: 121429181-121429181 on Build GRCh38
rs782524246 Chr.12: 121429397-121429397 on Build GRCh38
rs11065570 Chr.12: 121429418-121429418 on Build GRCh38
rs782805130 Chr.12: 121429209-121429209 on Build GRCh38
rs140610678 Chr.12: 121429058-121429058 on Build GRCh38
rs781789212 Chr.12: 121429067-121429067 on Build GRCh38
rs936638065 Chr.12: 121429456-121429456 on Build GRCh38
rs1042826480 Chr.12: 121429448-121429448 on Build GRCh38
rs188577056 Chr.12: 121429463-121429463 on Build GRCh38
rs192122366 Chr.12: 121429480-121429480 on Build GRCh38
rs539691677 Chr.12: 121429226-121429226 on Build GRCh38
rs1022733798 Chr.12: 121429193-121429193 on Build GRCh38
rs117682549 Chr.12: 121429102-121429102 on Build GRCh38
rs574515803 Chr.12: 121429421-121429421 on Build GRCh38
rs537766315 Chr.12: 121429332-121429332 on Build GRCh38
rs184170052 Chr.12: 121429487-121429487 on Build GRCh38
rs944101516 Chr.12: 121429070-121429070 on Build GRCh38
rs974370815 Chr.12: 121429376-121429376 on Build GRCh38
rs13754 Chr.12: 121429454-121429454 on Build GRCh38
rs944111635 Chr.12: 121429430-121429430 on Build GRCh38
rs146560042 Chr.12: 121429182-121429182 on Build GRCh38
rs543611631 Chr.12: 121429294-121429294 on Build GRCh38
rs1015269791 Chr.12: 121429281-121429281 on Build GRCh38
rs558335807 Chr.12: 121429228-121429228 on Build GRCh38
rs892983260 Chr.12: 121429163-121429163 on Build GRCh38
rs533073295 Chr.12: 121429285-121429285 on Build GRCh38
rs971161285 Chr.12: 121429196-121429196 on Build GRCh38
rs535527878 Chr.12: 121429071-121429071 on Build GRCh38
rs1053778243 Chr.12: 121429458-121429458 on Build GRCh38
rs998785012 Chr.12: 121429148-121429148 on Build GRCh38

View Full Product Details