Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122278877-122279390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP1
Entrez Gene ID
6249
Gene Name
CAP-Gly domain containing linker protein 1
Gene Aliases
CLIP, CLIP-170, CLIP170, CYLN1, RSN
UniGene
Hs.524809
Gene Chromosome Location
Chr.12: 122271432-122422956 on Build GRCh38
Transcripts
NM_001247997.1 NM_002956.2 NM_198240.1 XM_005253593.2 XM_006719552.3 XM_017019786.1 XM_017019787.1 XM_017019788.1 XM_017019789.1 XM_017019790.1 XM_017019791.1 XM_017019792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416565 c.3756C>T p.N1252N Chr.12: 122278919-122278919 on Build GRCh38
3811302 c.3763G>A p.E1255K Chr.12: 122279030-122279030 on Build GRCh38
4835323 c.3812C>G p.S1271* Chr.12: 122278896-122278896 on Build GRCh38
3416566 c.3789C>T p.N1263N Chr.12: 122278919-122278919 on Build GRCh38
1188639 c.3789G>T p.Q1263H Chr.12: 122278886-122278886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs5801472 Chr.12: 122279243-122279244 on Build GRCh38
rs367900505 Chr.12: 122279185-122279185 on Build GRCh38
rs377004725 Chr.12: 122278994-122278994 on Build GRCh38
rs199532478 Chr.12: 122279004-122279004 on Build GRCh38
rs375838255 Chr.12: 122279005-122279005 on Build GRCh38
rs773928829 Chr.12: 122278917-122278917 on Build GRCh38
rs190323097 Chr.12: 122279241-122279241 on Build GRCh38
rs143953208 Chr.12: 122278918-122278918 on Build GRCh38
rs139849947 Chr.12: 122279150-122279150 on Build GRCh38
rs775626536 Chr.12: 122279192-122279192 on Build GRCh38
rs747639103 Chr.12: 122279142-122279142 on Build GRCh38
rs201024409 Chr.12: 122278982-122278982 on Build GRCh38
rs776561836 Chr.12: 122279017-122279017 on Build GRCh38
rs543399003 Chr.12: 122279001-122279001 on Build GRCh38
rs991151536 Chr.12: 122279362-122279362 on Build GRCh38
rs756055262 Chr.12: 122279099-122279099 on Build GRCh38
rs759402510 Chr.12: 122279019-122279019 on Build GRCh38
rs200290353 Chr.12: 122278991-122278991 on Build GRCh38
rs778419728 Chr.12: 122278995-122278995 on Build GRCh38
rs899872308 Chr.12: 122279174-122279174 on Build GRCh38
rs760432235 Chr.12: 122278945-122278945 on Build GRCh38
rs17881033 Chr.12: 122279123-122279123 on Build GRCh38
rs368279544 Chr.12: 122279051-122279051 on Build GRCh38
rs759653407 Chr.12: 122278967-122278967 on Build GRCh38
rs533217721 Chr.12: 122279216-122279216 on Build GRCh38
rs766314040 Chr.12: 122278960-122278960 on Build GRCh38
rs997125359 Chr.12: 122279242-122279242 on Build GRCh38
rs1017351797 Chr.12: 122279222-122279222 on Build GRCh38
rs765357892 Chr.12: 122279024-122279024 on Build GRCh38
rs374016383 Chr.12: 122279183-122279183 on Build GRCh38
rs772442021 Chr.12: 122279052-122279052 on Build GRCh38
rs777395978 Chr.12: 122279169-122279169 on Build GRCh38
rs193021824 Chr.12: 122279260-122279260 on Build GRCh38
rs201175616 Chr.12: 122279010-122279010 on Build GRCh38
rs990045041 Chr.12: 122279267-122279267 on Build GRCh38
rs368064312 Chr.12: 122279002-122279002 on Build GRCh38
rs1017694955 Chr.12: 122278932-122278932 on Build GRCh38
rs888529881 Chr.12: 122279207-122279207 on Build GRCh38
rs764623561 Chr.12: 122279191-122279192 on Build GRCh38
rs775742839 Chr.12: 122279040-122279040 on Build GRCh38
rs761162814 Chr.12: 122279148-122279148 on Build GRCh38
rs202045807 Chr.12: 122278993-122278993 on Build GRCh38
rs932634233 Chr.12: 122279177-122279177 on Build GRCh38
rs1028239094 Chr.12: 122279182-122279182 on Build GRCh38
rs553942367 Chr.12: 122278916-122278916 on Build GRCh38
rs530971664 Chr.12: 122279253-122279254 on Build GRCh38
rs549451361 Chr.12: 122279263-122279263 on Build GRCh38
rs780583063 Chr.12: 122279132-122279132 on Build GRCh38
rs148653401 Chr.12: 122278919-122278919 on Build GRCh38
rs781720842 Chr.12: 122278992-122278992 on Build GRCh38
rs773025211 Chr.12: 122278928-122278928 on Build GRCh38
rs577877661 Chr.12: 122279294-122279294 on Build GRCh38
rs36024001 Chr.12: 122279210-122279210 on Build GRCh38
rs80047612 Chr.12: 122279253-122279253 on Build GRCh38
rs752907914 Chr.12: 122278974-122278974 on Build GRCh38
rs751898190 Chr.12: 122279074-122279074 on Build GRCh38
rs866529698 Chr.12: 122279270-122279270 on Build GRCh38
rs765234289 Chr.12: 122278973-122278973 on Build GRCh38
rs1004794481 Chr.12: 122279219-122279220 on Build GRCh38
rs756613341 Chr.12: 122279124-122279124 on Build GRCh38
rs1045123404 Chr.12: 122279085-122279085 on Build GRCh38
rs775894534 Chr.12: 122279147-122279148 on Build GRCh38
rs981108947 Chr.12: 122278989-122278989 on Build GRCh38
rs769822655 Chr.12: 122279190-122279190 on Build GRCh38
rs778243200 Chr.12: 122278981-122278981 on Build GRCh38
rs201689613 Chr.12: 122279013-122279013 on Build GRCh38
rs969911120 Chr.12: 122279350-122279350 on Build GRCh38
rs780216473 Chr.12: 122279253-122279255 on Build GRCh38
rs12313660 Chr.12: 122278976-122278976 on Build GRCh38
rs770699897 Chr.12: 122279184-122279184 on Build GRCh38
rs372003670 Chr.12: 122278908-122278908 on Build GRCh38
rs559303684 Chr.12: 122278962-122278962 on Build GRCh38
rs905229952 Chr.12: 122279109-122279109 on Build GRCh38
rs372241680 Chr.12: 122279165-122279165 on Build GRCh38

View Full Product Details