Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122702529-122703034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR2
Entrez Gene ID
338442
Gene Name
hydroxycarboxylic acid receptor 2
Gene Aliases
GPR109A, HCA2, HM74a, HM74b, NIACR1, PUMAG, Puma-g
UniGene
Hs.524812
Gene Chromosome Location
Chr.12: 122701293-122703357 on Build GRCh38
Transcripts
NM_177551.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
467931 c.281A>G p.K94R Chr.12: 122703003-122703003 on Build GRCh38
3457348 c.420C>T p.S140S Chr.12: 122702864-122702864 on Build GRCh38
4599552 c.536G>A p.S179N Chr.12: 122702748-122702748 on Build GRCh38
3457346 c.584T>C p.L195P Chr.12: 122702700-122702700 on Build GRCh38
5783710 c.513T>C p.N171N Chr.12: 122702771-122702771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141052038 Chr.12: 122702767-122702767 on Build GRCh38
rs676823 Chr.12: 122702692-122702692 on Build GRCh38
rs769808683 Chr.12: 122702717-122702717 on Build GRCh38
rs781064932 Chr.12: 122702712-122702712 on Build GRCh38
rs370020427 Chr.12: 122702974-122702974 on Build GRCh38
rs115977736 Chr.12: 122702960-122702960 on Build GRCh38
rs200398717 Chr.12: 122702832-122702832 on Build GRCh38
rs754954067 Chr.12: 122702861-122702861 on Build GRCh38
rs778983735 Chr.12: 122702795-122702795 on Build GRCh38
rs201162226 Chr.12: 122702936-122702936 on Build GRCh38
rs770293961 Chr.12: 122703006-122703006 on Build GRCh38
rs140267406 Chr.12: 122702881-122702881 on Build GRCh38
rs202042878 Chr.12: 122702833-122702833 on Build GRCh38
rs138859924 Chr.12: 122702632-122702632 on Build GRCh38
rs752005857 Chr.12: 122702785-122702787 on Build GRCh38
rs201174876 Chr.12: 122702681-122702681 on Build GRCh38
rs676404 Chr.12: 122702758-122702758 on Build GRCh38
rs142084430 Chr.12: 122702638-122702638 on Build GRCh38
rs199839290 Chr.12: 122702933-122702933 on Build GRCh38
rs202235765 Chr.12: 122702811-122702811 on Build GRCh38
rs764419070 Chr.12: 122702548-122702548 on Build GRCh38
rs200113893 Chr.12: 122702741-122702741 on Build GRCh38
rs200794872 Chr.12: 122702896-122702896 on Build GRCh38
rs769715682 Chr.12: 122702619-122702619 on Build GRCh38
rs147573131 Chr.12: 122702785-122702785 on Build GRCh38
rs368271708 Chr.12: 122702819-122702819 on Build GRCh38
rs760345055 Chr.12: 122702696-122702696 on Build GRCh38
rs201310632 Chr.12: 122702609-122702609 on Build GRCh38
rs760143699 Chr.12: 122702821-122702821 on Build GRCh38
rs199573054 Chr.12: 122702735-122702735 on Build GRCh38
rs747232483 Chr.12: 122702981-122702981 on Build GRCh38
rs199514271 Chr.12: 122702637-122702637 on Build GRCh38
rs200171322 Chr.12: 122702563-122702563 on Build GRCh38
rs562912587 Chr.12: 122702808-122702808 on Build GRCh38
rs200721962 Chr.12: 122702716-122702716 on Build GRCh38
rs767868840 Chr.12: 122702653-122702653 on Build GRCh38
rs202036334 Chr.12: 122702768-122702768 on Build GRCh38
rs201831072 Chr.12: 122702736-122702736 on Build GRCh38
rs749224152 Chr.12: 122702783-122702783 on Build GRCh38
rs375831072 Chr.12: 122702769-122702769 on Build GRCh38
rs755870982 Chr.12: 122702854-122702854 on Build GRCh38
rs200794525 Chr.12: 122702983-122702983 on Build GRCh38
rs758469046 Chr.12: 122702969-122702969 on Build GRCh38
rs144258873 Chr.12: 122702820-122702820 on Build GRCh38
rs774485880 Chr.12: 122702728-122702728 on Build GRCh38
rs144376493 Chr.12: 122702766-122702766 on Build GRCh38
rs752320524 Chr.12: 122702678-122702678 on Build GRCh38
rs942555261 Chr.12: 122702703-122702703 on Build GRCh38
rs773672846 Chr.12: 122702733-122702733 on Build GRCh38
rs371493470 Chr.12: 122702911-122702911 on Build GRCh38
rs150357116 Chr.12: 122702886-122702886 on Build GRCh38
rs931357507 Chr.12: 122702751-122702751 on Build GRCh38
rs377019221 Chr.12: 122702570-122702570 on Build GRCh38
rs761046567 Chr.12: 122702810-122702810 on Build GRCh38
rs202134216 Chr.12: 122702554-122702554 on Build GRCh38
rs754899604 Chr.12: 122702958-122702958 on Build GRCh38
rs748346019 Chr.12: 122702797-122702797 on Build GRCh38
rs137895931 Chr.12: 122702726-122702726 on Build GRCh38
rs200224963 Chr.12: 122702818-122702818 on Build GRCh38
rs745849991 Chr.12: 122702610-122702610 on Build GRCh38
rs369894128 Chr.12: 122702780-122702780 on Build GRCh38
rs200492929 Chr.12: 122702700-122702700 on Build GRCh38
rs777708833 Chr.12: 122702584-122702584 on Build GRCh38
rs199822101 Chr.12: 122702882-122702882 on Build GRCh38
rs200999108 Chr.12: 122702579-122702579 on Build GRCh38
rs199754768 Chr.12: 122702826-122702826 on Build GRCh38
rs375811004 Chr.12: 122702918-122702918 on Build GRCh38
rs151172149 Chr.12: 122702860-122702860 on Build GRCh38
rs770491968 Chr.12: 122702915-122702915 on Build GRCh38
rs142060832 Chr.12: 122702806-122702806 on Build GRCh38
rs200534953 Chr.12: 122702901-122702901 on Build GRCh38
rs761263431 Chr.12: 122702661-122702661 on Build GRCh38
rs201138003 Chr.12: 122702822-122702822 on Build GRCh38
rs750055800 Chr.12: 122702849-122702849 on Build GRCh38
rs200587011 Chr.12: 122702629-122702629 on Build GRCh38
rs145014727 Chr.12: 122702781-122702781 on Build GRCh38
rs964281307 Chr.12: 122702888-122702888 on Build GRCh38
rs746863234 Chr.12: 122702604-122702604 on Build GRCh38
rs571187269 Chr.12: 122702654-122702654 on Build GRCh38
rs202043479 Chr.12: 122702928-122702928 on Build GRCh38
rs774047399 Chr.12: 122702825-122702825 on Build GRCh38
rs150040336 Chr.12: 122702730-122702730 on Build GRCh38
rs676770 Chr.12: 122702724-122702724 on Build GRCh38
rs138078446 Chr.12: 122702903-122702903 on Build GRCh38
rs754317986 Chr.12: 122702760-122702760 on Build GRCh38
rs138786161 Chr.12: 122702994-122702994 on Build GRCh38
rs766114637 Chr.12: 122702859-122702859 on Build GRCh38
rs752639607 Chr.12: 122702963-122702963 on Build GRCh38
rs753670084 Chr.12: 122702953-122702953 on Build GRCh38
rs144981479 Chr.12: 122702967-122702967 on Build GRCh38
rs755391802 Chr.12: 122702980-122702983 on Build GRCh38
rs751262399 Chr.12: 122702708-122702708 on Build GRCh38
rs201756503 Chr.12: 122702902-122702902 on Build GRCh38
rs200758992 Chr.12: 122702862-122702862 on Build GRCh38
rs753094236 Chr.12: 122702952-122702952 on Build GRCh38
rs146071786 Chr.12: 122702620-122702620 on Build GRCh38
rs546599894 Chr.12: 122702642-122702642 on Build GRCh38
rs201792054 Chr.12: 122703000-122703000 on Build GRCh38
rs750950242 Chr.12: 122702569-122702569 on Build GRCh38
rs757461993 Chr.12: 122702982-122702982 on Build GRCh38
rs747704207 Chr.12: 122702571-122702571 on Build GRCh38
rs766970337 Chr.12: 122702747-122702747 on Build GRCh38
rs200705418 Chr.12: 122702919-122702919 on Build GRCh38
rs201460900 Chr.12: 122702705-122702705 on Build GRCh38
rs200369885 Chr.12: 122702671-122702671 on Build GRCh38
rs200146945 Chr.12: 122702977-122702977 on Build GRCh38
rs470923 Chr.12: 122702711-122702711 on Build GRCh38
rs761348052 Chr.12: 122702927-122702927 on Build GRCh38
rs772880430 Chr.12: 122702931-122702931 on Build GRCh38
rs758167318 Chr.12: 122702583-122702583 on Build GRCh38

View Full Product Details