Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113474086-113474628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D2
Entrez Gene ID
55208
Gene Name
defective in cullin neddylation 1 domain containing 2
Gene Aliases
C13orf17
UniGene
Hs.682987
Gene Chromosome Location
Chr.13: 113455819-113492047 on Build GRCh38
Transcripts
NM_001014283.1 XM_017020643.1 XM_017020644.1 XM_017020645.1 XM_017020646.1 XM_017020647.1 XM_017020648.1 XM_017020649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235072 c.446A>G p.K149R Chr.13: 113474198-113474198 on Build GRCh38
4413194 c.510G>C p.Q170H Chr.13: 113474134-113474134 on Build GRCh38
6235071 c.446A>G p.K149R Chr.13: 113474198-113474198 on Build GRCh38
7270284 c.413A>C p.K138T Chr.13: 113474231-113474231 on Build GRCh38
4872586 c.390-1G>T p.? Chr.13: 113474255-113474255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922799127 Chr.13: 113474290-113474290 on Build GRCh38
rs886172210 Chr.13: 113474326-113474326 on Build GRCh38
rs768276040 Chr.13: 113474170-113474170 on Build GRCh38
rs1042562597 Chr.13: 113474433-113474433 on Build GRCh38
rs995529842 Chr.13: 113474566-113474566 on Build GRCh38
rs886398140 Chr.13: 113474493-113474493 on Build GRCh38
rs191610295 Chr.13: 113474386-113474386 on Build GRCh38
rs755119638 Chr.13: 113474254-113474254 on Build GRCh38
rs978805561 Chr.13: 113474425-113474425 on Build GRCh38
rs1016092387 Chr.13: 113474291-113474291 on Build GRCh38
rs368577955 Chr.13: 113474258-113474258 on Build GRCh38
rs371126972 Chr.13: 113474259-113474259 on Build GRCh38
rs751471323 Chr.13: 113474242-113474242 on Build GRCh38
rs940421401 Chr.13: 113474332-113474332 on Build GRCh38
rs142079557 Chr.13: 113474151-113474151 on Build GRCh38
rs755031793 Chr.13: 113474122-113474122 on Build GRCh38
rs749621059 Chr.13: 113474152-113474152 on Build GRCh38
rs552619184 Chr.13: 113474391-113474391 on Build GRCh38
rs1056139075 Chr.13: 113474570-113474570 on Build GRCh38
rs770254615 Chr.13: 113474149-113474149 on Build GRCh38
rs936448688 Chr.13: 113474464-113474464 on Build GRCh38
rs765765157 Chr.13: 113474584-113474584 on Build GRCh38
rs898799674 Chr.13: 113474344-113474344 on Build GRCh38
rs924988460 Chr.13: 113474434-113474434 on Build GRCh38
rs368160004 Chr.13: 113474472-113474472 on Build GRCh38
rs759246067 Chr.13: 113474597-113474597 on Build GRCh38
rs760300930 Chr.13: 113474166-113474166 on Build GRCh38
rs748318241 Chr.13: 113474141-113474141 on Build GRCh38
rs1004753286 Chr.13: 113474285-113474285 on Build GRCh38
rs925495309 Chr.13: 113474600-113474600 on Build GRCh38
rs1054773210 Chr.13: 113474466-113474466 on Build GRCh38
rs373874285 Chr.13: 113474205-113474205 on Build GRCh38
rs771361709 Chr.13: 113474156-113474156 on Build GRCh38
rs912845110 Chr.13: 113474414-113474414 on Build GRCh38
rs781437102 Chr.13: 113474132-113474132 on Build GRCh38
rs1020244644 Chr.13: 113474583-113474583 on Build GRCh38
rs761384433 Chr.13: 113474193-113474193 on Build GRCh38
rs750919985 Chr.13: 113474265-113474266 on Build GRCh38
rs568601615 Chr.13: 113474492-113474492 on Build GRCh38
rs771389495 Chr.13: 113474272-113474272 on Build GRCh38
rs774973893 Chr.13: 113474162-113474162 on Build GRCh38
rs527719312 Chr.13: 113474390-113474390 on Build GRCh38
rs374187714 Chr.13: 113474277-113474277 on Build GRCh38
rs564574820 Chr.13: 113474407-113474407 on Build GRCh38
rs986999376 Chr.13: 113474401-113474401 on Build GRCh38
rs922865652 Chr.13: 113474428-113474428 on Build GRCh38
rs529541274 Chr.13: 113474495-113474495 on Build GRCh38
rs1037343397 Chr.13: 113474341-113474341 on Build GRCh38
rs370170005 Chr.13: 113474190-113474190 on Build GRCh38
rs531715705 Chr.13: 113474419-113474419 on Build GRCh38
rs368427815 Chr.13: 113474293-113474293 on Build GRCh38
rs146925124 Chr.13: 113474578-113474578 on Build GRCh38
rs925945992 Chr.13: 113474217-113474217 on Build GRCh38
rs1046338738 Chr.13: 113474490-113474490 on Build GRCh38
rs940452657 Chr.13: 113474491-113474491 on Build GRCh38
rs560915085 Chr.13: 113474501-113474501 on Build GRCh38
rs963093529 Chr.13: 113474565-113474565 on Build GRCh38
rs1013288265 Chr.13: 113474482-113474482 on Build GRCh38
rs1028393535 Chr.13: 113474572-113474572 on Build GRCh38
rs764878500 Chr.13: 113474194-113474194 on Build GRCh38
rs773580534 Chr.13: 113474437-113474437 on Build GRCh38
rs954037902 Chr.13: 113474573-113474573 on Build GRCh38
rs762761315 Chr.13: 113474236-113474236 on Build GRCh38
rs749660245 Chr.13: 113474271-113474271 on Build GRCh38
rs766353562 Chr.13: 113474239-113474239 on Build GRCh38
rs975651686 Chr.13: 113474234-113474234 on Build GRCh38
rs577199390 Chr.13: 113474427-113474427 on Build GRCh38
rs370446039 Chr.13: 113474529-113474529 on Build GRCh38
rs776010331 Chr.13: 113474304-113474304 on Build GRCh38
rs777999822 Chr.13: 113474270-113474270 on Build GRCh38
rs558939749 Chr.13: 113474363-113474363 on Build GRCh38
rs934185531 Chr.13: 113474450-113474450 on Build GRCh38
rs766090685 Chr.13: 113474394-113474394 on Build GRCh38
rs756317034 Chr.13: 113474268-113474268 on Build GRCh38
rs115090816 Chr.13: 113474557-113474557 on Build GRCh38
rs761347088 Chr.13: 113474306-113474306 on Build GRCh38
rs751662064 Chr.13: 113474119-113474119 on Build GRCh38
rs967465934 Chr.13: 113474424-113474424 on Build GRCh38
rs942952911 Chr.13: 113474220-113474220 on Build GRCh38
rs1034269137 Chr.13: 113474371-113474371 on Build GRCh38
rs1051489830 Chr.13: 113474299-113474299 on Build GRCh38
rs774747404 Chr.13: 113474292-113474292 on Build GRCh38
rs1033428965 Chr.13: 113474426-113474427 on Build GRCh38
rs962830321 Chr.13: 113474345-113474345 on Build GRCh38
rs2257442 Chr.13: 113474467-113474467 on Build GRCh38
rs978634842 Chr.13: 113474599-113474599 on Build GRCh38

View Full Product Details