Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113648720-113649213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP4B
Entrez Gene ID
496
Gene Name
ATPase H+/K+ transporting beta subunit
Gene Aliases
ATP6B
UniGene
Hs.434202
Gene Chromosome Location
Chr.13: 113648804-113658198 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

Gene Symbol
GRK1
Entrez Gene ID
6011
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
GPRK1, RHOK, RK
UniGene
Hs.103501
Gene Chromosome Location
Chr.13: 113645771-113735664 on Build GRCh38
Transcripts
NM_000705.3 XM_017020684.1

SNP Detail

SNP ID Chromosome Location
rs541916892 Chr.13: 113649116-113649116 on Build GRCh38
rs1020098244 Chr.13: 113649003-113649003 on Build GRCh38
rs960065667 Chr.13: 113648781-113648781 on Build GRCh38
rs955139640 Chr.13: 113649092-113649092 on Build GRCh38
rs984266536 Chr.13: 113648903-113648903 on Build GRCh38
rs746240159 Chr.13: 113649066-113649066 on Build GRCh38
rs758154504 Chr.13: 113648826-113648826 on Build GRCh38
rs752533399 Chr.13: 113648817-113648817 on Build GRCh38
rs577646030 Chr.13: 113649053-113649053 on Build GRCh38
rs910033206 Chr.13: 113648923-113648923 on Build GRCh38
rs777516873 Chr.13: 113649052-113649052 on Build GRCh38
rs879403172 Chr.13: 113649010-113649011 on Build GRCh38
rs12867415 Chr.13: 113649169-113649169 on Build GRCh38
rs1043566927 Chr.13: 113649078-113649078 on Build GRCh38
rs922301771 Chr.13: 113648963-113648963 on Build GRCh38
rs892356849 Chr.13: 113649070-113649070 on Build GRCh38
rs1034398245 Chr.13: 113649136-113649136 on Build GRCh38
rs1052313817 Chr.13: 113649044-113649044 on Build GRCh38
rs763755571 Chr.13: 113648843-113648843 on Build GRCh38
rs187605147 Chr.13: 113648754-113648754 on Build GRCh38
rs974310570 Chr.13: 113649143-113649143 on Build GRCh38
rs565116718 Chr.13: 113649100-113649100 on Build GRCh38
rs192103532 Chr.13: 113649072-113649072 on Build GRCh38
rs373204828 Chr.13: 113648936-113648936 on Build GRCh38
rs559308328 Chr.13: 113648998-113648998 on Build GRCh38
rs922349919 Chr.13: 113648755-113648755 on Build GRCh38
rs532022704 Chr.13: 113649003-113649004 on Build GRCh38
rs113895052 Chr.13: 113649002-113649003 on Build GRCh38
rs1014234667 Chr.13: 113649142-113649142 on Build GRCh38
rs951633105 Chr.13: 113649183-113649183 on Build GRCh38
rs780185934 Chr.13: 113649191-113649191 on Build GRCh38
rs755860877 Chr.13: 113648916-113648916 on Build GRCh38
rs534362591 Chr.13: 113648962-113648962 on Build GRCh38
rs905156619 Chr.13: 113648883-113648883 on Build GRCh38
rs562549692 Chr.13: 113649073-113649073 on Build GRCh38
rs941647063 Chr.13: 113648855-113648855 on Build GRCh38
rs918808031 Chr.13: 113648820-113648820 on Build GRCh38
rs1025919620 Chr.13: 113649155-113649155 on Build GRCh38
rs896556246 Chr.13: 113649190-113649190 on Build GRCh38
rs575338743 Chr.13: 113649187-113649187 on Build GRCh38
rs375576661 Chr.13: 113649003-113649003 on Build GRCh38
rs139513600 Chr.13: 113648816-113648816 on Build GRCh38
rs1029412545 Chr.13: 113649077-113649077 on Build GRCh38
rs1002063433 Chr.13: 113649117-113649117 on Build GRCh38
rs541141779 Chr.13: 113649139-113649139 on Build GRCh38

View Full Product Details